Silver -Ag
Silver Foil, 0.25mm (0.01 in.) thick, hard, Premion™, 99.998% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver Name Note: Hard CAS: 7440-22-4 Physical Form: Foil, 0.25mm (0.01 in.) thick Assay Percent Range: 99.998% (Metals basis) MDL Number: MFCD00003397
Silver Gauze, 80 mesh woven from 0.115mm (0.0045 in.) dia. wire, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Gauze Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003397
Silver Foil, 0.025mm (0.001 in.) thick, annealed, 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver Name Note: Annealed CAS: 7440-22-4 Physical Form: Foil, 0.025mm (0.001 in.) thick Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00003397
Silver Wire, 1.0mm (0.04 in.) dia., hard, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver Name Note: Hard CAS: 7440-22-4 Physical Form: Wire, 1.0mm (0.04 in.) dia. Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003397
Silver Powder, spherical, -500 mesh, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Spherical Powder Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003397
Silver Foil, 0.1mm (0.004 in.) thick, hard, Premion™, 99.998% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver Name Note: Hard CAS: 7440-22-4 Physical Form: Foil, 0.1mm (0.004 in.) thick Assay Percent Range: 99.998% (Metals basis) MDL Number: MFCD00003397
Silver Wire, 0.5mm (0.02 in.) dia., 1/2 hard, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver Name Note: 1/2 Hard CAS: 7440-22-4 Physical Form: Wire, 0.5mm (0.02 in.) dia. Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003397
Silver Foil, 2.0mm (0.08 in.) thick, Premion™, 99.9985% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Foil, 2.0mm (0.08 in.) thick Assay Percent Range: 99.9985% (Metals basis) MDL Number: MFCD00003397
Silver shot, 99.999% (metals basis), Alfa Aesar™, Premion™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Shot, 2-3mm (0.08-0.12 in.) Assay Percent Range: 99.999% (Metals basis) MDL Number: MFCD00003397
Silver foil, 99.998% (metals basis), Alfa Aesar™
Chemical Name or Material: Silver Name Note: Annealed CAS: 7440-22-4 Physical Form: Foil, 0.1mm (0.004 in.) thick Assay Percent Range: 99.998% (Metals basis) MDL Number: MFCD00003397
Silver Powder, 0.7-1.3 micron, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Powder Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003397
Silver Foil, 1.0mm (0.04 in.) thick, Premion™, 99.9985% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Foil, 1.0mm (0.04 in.) thick Assay Percent Range: 99.9985% (Metals basis) MDL Number: MFCD00003397
Silver Wire, 0.25mm (0.01 in.) dia., hard, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver Name Note: Hard CAS: 7440-22-4 Physical Form: Wire, 0.25mm (0.01 in.) dia. Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003397
Silver Foil, 0.5mm (0.02 in.) thick, annealed, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver Name Note: Annealed CAS: 7440-22-4 Physical Form: Foil, 0.5mm (0.02 in.) thick Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003397
Silver wire, 99.999% (metals basis), Alfa Aesar™, Premion™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Wire, 2mm (0.08 in.) dia. Assay Percent Range: 99.999% (Metals basis) MDL Number: MFCD00003397
Silver, 99.9%, foil, 25x25mm, 1.0mm thick, 1ea. = 6.6g, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Silver Name Note: 99.9%, Each sheet is 25x25mm, 6.6g CAS: 7440-22-4 Physical Form: Foil Assay Percent Range: 99.90% Molecular Formula: Ag Linear Formula: Ag Formula Weight: 107.87 MDL Number: MFCD00003397
Silver wire, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Silver Name Note: Annealed CAS: 7440-22-4 Physical Form: Wire, 1.0mm (0.04 in.) dia. Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003397
Silver Wire, 1.0mm (0.04 in.) dia., Premion™, 99.999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Wire, 1.0mm (0.04 in.) dia. Assay Percent Range: 99.999% (Metals basis) MDL Number: MFCD00003397
Silver wire, 99.9985% (metals basis), Alfa Aesar™, Premion™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Wire, 0.25mm (0.01 in.) dia. Assay Percent Range: 99.9985% (Metals basis) MDL Number: MFCD00003397
Silver casting grain, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Casting Grain, 1.6-6.35mm (0.063-0.25 in.) Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003397
Silver foil, 99.998% (metals basis), Alfa Aesar™, Premion™
Chemical Name or Material: Silver Name Note: Annealed CAS: 7440-22-4 Physical Form: Foil, 0.025mm (0.001 in.) thick Assay Percent Range: 99.998% (Metals basis) MDL Number: MFCD00003397
Silver wire, 99.95% (metals basis), Alfa Aesar™
Chemical Name or Material: Silver Name Note: Hard CAS: 7440-22-4 Physical Form: Wire, 0.203mm (0.008 in.) dia. Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00003397
Silver Powder, 4-7 micron, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Powder Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003397
Siver Rod, Alfa Aesar™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Rod, 5mm (0.2 in.) dia. Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00003397
Alfa Aesar™, Silver Conductive Ink
Chemical Name or Material: Silver Conductive Ink
Silver wire, Alfa Aesar™, Premion™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Wire, 1.5mm (0.06 in.) dia. Assay Percent Range: 99.998% (Metals basis) MDL Number: MFCD00003397
Silver gauze, Alfa Aesar™
Chemical Name or Material: Silver wire gauze CAS: 7440-22-4 Physical Form: Wire Cloth MDL Number: MFCD00003397
Silver, 99.9% (trace metal basis), wire, 0.25mm dia., 1ea. = 10.4g, ACROS Organics™
Chemical Name or Material: Silver Name Note: 99.9% (1ea = 10.4gr) CAS: 7440-22-4 Purity Grade: Trace Metal Basis Physical Form: Wire Assay Percent Range: 99.9% Molecular Formula: Ag Linear Formula: Ag Formula Weight: 107.87 MDL Number: MFCD00003397
Silver Wire, 0.127mm (0.005in.) dia., Annealed, 99.9% (Metal Basis), Alfa Aesar
Chemical Name or Material: Silver Name Note: Annealed CAS: 7440-22-4 Physical Form: Wire, 0.127mm (0.005 in.) dia. Assay Percent Range: 99.9%, metal basis MDL Number: MFCD00003397
Silver Shot, 1-5mm (0.04-0.2 in.), 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Silver CAS: 7440-22-4 Physical Form: Shot, 1-5mm (0.04-0.2 in.) Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003397
  spinner