Sodium -Na
Sodium molybdate(VI) dihydrate, 99+%, ACROS Organics™
Chemical Name or Material: Sodium molybdate(VI) dihydrate Name Note: 99+% CAS: 10102-40-6 Assay: ≥99.0% (Na2MoO4.2H2O) Assay Percent Range: 99+% Molecular Formula: MoNa2O4·2H2O Linear Formula: Na2MoO4·2H2O Formula Weight: 241.95 MDL Number: MFCD00149170
Sodium phosphate, monobasic monohydrate, 99.5%, Acros Organics Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium phosphate, monobasic monohydrate CAS: 10049-21-5 Purity Grade: For biochemistry Assay Percent Range: 99.5% Molecular Formula: H2NaO4P·H2O Formula Weight: 137.99 Synonym: Dihydrogen sodium phosphate monohydrate
Sodium perchlorate, anhydrous, ACS, 98.0 to 102.0%, Alfa Aesar™
Chemical Name or Material: Sodium perchlorate, anhydrous Name Note: For analysis ACS CAS: 7601-89-0 Physical Form: Crystalline Assay Percent Range: 98.0 to 102.0% Molecular Formula: NaClO4 Formula Weight: 122.46 MDL Number: MFCD00003480
Sodium hexachloroplatinate(IV) hexahydrate, Alfa Aesar™
Chemical Name or Material: Sodium hexachloroplatinate(IV) hexahydrate CAS: 19583-77-8 Physical Form: Crystalline Molecular Formula: Na2PtCl6·6H2O Formula Weight: 561.88; 453.79 (anhydrous) MDL Number: MFCD00149172 Synonym: Sodium chloroplatinate(IV)
Sodium Acetate Trihydrate (Granular/USP/FCC), Fisher Chemical
Chemical Name or Material: Sodium Acetate Trihydrate CAS: 6131-90-4 Purity Grade: FCC/USP Physical Form: Solid Assay: 99.0 to 101.0% (FCC) Molecular Formula: C2H3NaO2 · 3 H2O Linear Formula: CH3COONa·3H2O Formula Weight: 136.08 MDL Number: MFCD00071557
Sodium chloride Crystal Optic Rectangle, 38.5 x 19.5 x 4mm, Alfa Aesar™
Chemical Name or Material: Sodium chloride Crystal Optic Rectangle Name Note: (Drilled), polished both sides
Sodium hydrogen phosphate heptahydrate, ACS, 98.0-102.0%, Alfa Aesar™
Chemical Name or Material: Sodium hydrogen phosphate heptahydrate CAS: 7782-85-6 Purity Grade: ACS Grade Physical Form: Crystalline Assay Percent Range: 98.0-102.0% Molecular Formula: Na2HPO4·7H2O Formula Weight: 268.07; 141.98 (anhydrous) MDL Number: MFCD00149180 Synonym: Disodium phosphate
Sodium chlorite, 80%, pure, unstabilized, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium chlorite Name Note: Unstabilized CAS: 7758-19-2 CAS: 7647-14-5 Purity Grade: Pure Physical Form: Powder Assay: ≥75% Assay Percent Range: 80% Molecular Formula: ClNaO2 Linear Formula: NaClO2 Formula Weight: 90.44 MDL Number: MFCD00003478
Sodium metaborate tetrahydrate, 98.5%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium metaborate tetrahydrate Name Note: p.a. CAS: 10555-76-7 Purity Grade: Extra Pure Assay: ≥98.0% Assay Percent Range: 98.5% Molecular Formula: BNaO2·4H2O Linear Formula: NaBO2·4H2O Formula Weight: 137.86 MDL Number: MFCD00149205
Dodecyl sulfate sodium salt, 98%, for biochemistry, suitable for electrophoresis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dodecyl sulfate sodium salt CAS: 151-21-3 CAS: 7757-82-6 CAS: 7647-14-5 Purity Grade: For biochemistry Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C12H25NaO4S Linear Formula: CH3(CH2)11OSO3Na Formula Weight: 288.38 Synonym: Dodecyl Sodium Sulfate
Dextran sulfate sodium salt, approx. M.W. 500,000, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dextran sulfate sodium salt CAS: 9011-18-1
Sodium borohydride, 99%, VenPure™ AF caplets
Chemical Name or Material: Sodium borohydride CAS: 16940-66-2 Molecular Formula: H4BNa Formula Weight: 37.83 Synonym: SBH
Sodium Sulfate Anhydrous (Low Nitrogen), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Sulfate Anhydrous CAS: 7757-82-6 Purity Grade: Low Nitrogen Molecular Formula: Na2O4S Formula Weight: 142.04 MDL Number: MFCD00003504
Sodium Sulfite Anhydrous (Crystalline/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Sulfite Anhydrous CAS: 7757-83-7 Purity Grade: Certified ACS Grade Physical Form: Solid Assay: ≥98% Assay Percent Range: ≥98 % Molecular Formula: Na2O3S Formula Weight: 126.04 MDL Number: MFCD00003503
Sodium Sulfate Anhydrous (Powder/USP/FCC/EP/BP), Fisher Chemical
Chemical Name or Material: Sodium Sulfate Anhydrous CAS: 7757-82-6 Purity Grade: BP/EP/FCC/USP Physical Form: Solid Assay: 98.5 to 101.0% (EP) Molecular Formula: Na2O4S Formula Weight: 142.04 MDL Number: MFCD00003504
Sodium sulfate decahydrate, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium sulfate decahydrate CAS: 7727-73-3 Purity Grade: Extra Pure Assay: ≥99.0% Molecular Formula: Na2O4S·10H2O Linear Formula: Na2SO4·10H2O Formula Weight: 322.2 MDL Number: MFCD00149189 Synonym: Glauber';s salt
Sodium Carbonate Anhydrous (Calcined/Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Carbonate Anhydrous Name Note: Calcined CAS: 497-19-8 Purity Grade: Laboratory Grade Physical Form: Solid Molecular Formula: CNa2O3 Formula Weight: 105.99 MDL Number: MFCD00003494 Synonym: Soda Ash
Sodium Nitrate (Crystalline/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Nitrate CAS: 7631-99-4 Purity Grade: Certified ACS Grade Physical Form: Crystalline Solid Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: NNaO3 Formula Weight: 84.99 MDL Number: MFCD00011119 Synonym: Chili Saltpeter
Sodium chlorate, 99% min, Alfa Aesar™
Chemical Name or Material: Sodium chlorate CAS: 7775-09-9 Physical Form: Granular Assay Percent Range: 99% min. Molecular Formula: NaClO3 Formula Weight: 106.45 MDL Number: MFCD00003479
Sodium Sulfate Anhydrous (Powder/USP/FCC), Fisher Chemical
Chemical Name or Material: Sodium Sulfate Anhydrous CAS: 7757-82-6 Purity Grade: FCC/USP Physical Form: Solid Assay: 99.0 to 100.5% (FCC) Molecular Formula: Na2O4S Formula Weight: 142.04 MDL Number: MFCD00003504
Sodium hydroxide, 1.0N Standardized Solution, Alfa Aesar™
Chemical Name or Material: Sodium hydroxide CAS: 1310-73-2 Physical Form: Liquid Molecular Formula: NaOH Formula Weight: 40 MDL Number: MFCD00003548
Sodium perchlorate, 98%, extra pure, ACROS Organics™
Chemical Name or Material: Sodium perchlorate CAS: 7601-89-0 Purity Grade: Extra Pure Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: ClNaO4 Linear Formula: NaClO4 Formula Weight: 122.44 MDL Number: MFCD00003480
Sodium Thiosulfate, Anhydrous, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium thiosulfate CAS: 7772-98-7 Purity Grade: Extra Pure Assay: ≥98.0% (on dry substance) Assay Percent Range: 98.5% Molecular Formula: Na2O3S2 Linear Formula: Na2S2O3 Formula Weight: 158.1
Sodium borohydride, 98+%, powder, Acros Organics Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium borohydride CAS: 16940-66-2 Physical Form: Powder Assay: ≥98.0% Assay Percent Range: 98+% Molecular Formula: H4BNa Linear Formula: NaBH4 Formula Weight: 37.83 MDL Number: MFCD00003518 Synonym: SBH
Sodium tetraborate, anhydrous, 99.95% (metals basis), Alfa Aesar™
Chemical Name or Material: Sodium tetraborate CAS: 1330-43-4 Physical Form: Powder Assay Percent Range: 99.95% (metals basis) Molecular Formula: B4Na2O7 Formula Weight: 201.22 MDL Number: MFCD00081185 Synonym: Borax
Sodium metatungstate monohydrate, Alfa Aesar™
Chemical Name or Material: Sodium metatungstate monohydrate CAS: 12141-67-2 Physical Form: Powder Molecular Formula: Na6O39W12·H2O Formula Weight: 2986.13 (2968.12 Anhydrous) MDL Number: MFCD00061466 Synonym: Sodium polytungstate
Sodium Thiocyanate (Crystalline/Certified ACS), Fisher Chemical
Chemical Name or Material: Sodium Thiocyanate CAS: 540-72-7 Purity Grade: Certified ACS Grade Physical Form: Solid Assay: ≥98% Assay Percent Range: ≥98 % Molecular Formula: CNNaS Formula Weight: 81.07 MDL Number: MFCD00011123 Synonym: Sodium Isothiocyanate
Sodium aluminium hydride, 93%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 13770-96-2 Assay Percent Range: 93% Molecular Formula: H4AlNa Formula Weight: 54 MDL Number: MFCD00192443
Sodium Bisulfite (Powder/Certified ACS), Fisher Chemical
Chemical Name or Material: Sodium Bisulfite CAS: 7631-90-5 CAS: 7681-57-4 CAS: 7757-82-6 Purity Grade: Certified ACS Grade Physical Form: Powder/Solid Assay: ≥58.5% Molecular Formula: HNaO3S MDL Number: MFCD00003530 Synonym: Sodium Hydrogen Sulfite
Sodium azide, 99%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium azide Name Note: 99% CAS: 26628-22-8 Purity Grade: Extra Pure Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: N3Na Linear Formula: NaN3 Formula Weight: 65.01 MDL Number: MFCD00003536
  spinner