Sodium Hydroxide
Sodium hydroxide, pure, 1N Standard Solution, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium hydroxide CAS: 1310-73-2 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: HNaO Linear Formula: NaOH Formula Weight: 40 Synonym: Caustic soda
Sodium Hydroxide Solution (5N/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 5N CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: NaOH · H2O MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide (Pellets/NF/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide CAS: 1310-73-2 CAS: 497-19-8 Purity Grade: FCC/NF Physical Form: Solid Assay: 95.0 to 100.5% (NF) Molecular Formula: NaOH Formula Weight: 40 MDL Number: MFCD00003548 Synonym: Caustic Soda
Sodium hydroxide, Acros Organics
Chemical Name or Material: Sodium Hydroxide CAS: 1310-73-2 Purity Grade: Extra Pure Molecular Formula: HNaO Formula Weight: 40 Synonym: Caustic soda
Sodium Hydroxide Solution (6.25N), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 6.25N CAS: 1310-73-2 CAS: 7732-18-5 Molecular Formula: NaOH · H2O MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide (Pellets/Certified ACS), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide CAS: 1310-73-2 CAS: 497-19-8 Purity Grade: Certified ACS Grade Physical Form: Solid Assay: ≥97.0% Assay Percent Range: ≥97.0 % Molecular Formula: NaOH Formula Weight: 40 MDL Number: MFCD00003548 Synonym: Caustic Soda
Sodium hydroxide, extra pure, 50 wt% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium hydroxide Name Note: 50 wt% solution in water CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Extra Pure Assay: 49.0 to 51.0% (Acidimetry) Molecular Formula: HNaO Linear Formula: NaOH Formula Weight: 40 MDL Number: MFCD00003548 Synonym: Caustic soda
Sodium Hydroxide Solution (2N/Certified), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 2N CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: NaOH · H2O MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium hydroxide, 98%, extra pure, pellets, ACROS Organics™
Chemical Name or Material: Sodium hydroxide CAS: 1310-73-2 Purity Grade: Extra Pure Physical Form: Crystalline Powder Assay: ≥97.5% (Acidimetry) Assay Percent Range: 98% Molecular Formula: HNaO Linear Formula: NaOH Formula Weight: 40 MDL Number: MFCD00003548 Synonym: Caustic soda
Sodium hydroxide, 0.2 N standard solution, ACROS Organics™
Chemical Name or Material: Sodium hydroxide CAS: 1310-73-2 CAS: 7732-18-5 Molecular Formula: HNaO Linear Formula: NaOH Formula Weight: 39.99 MDL Number: MFCD00003548 Synonym: Caustic soda
Sodium hydroxide, pure, pellets, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium hydroxide CAS: 1310-73-2 Purity Grade: Pure Molecular Formula: HNaO Linear Formula: NaOH Formula Weight: 40 Synonym: Caustic soda
Sodium Hydroxide Solution, 10N Products for Science Education
Chemical Name or Material: Sodium Hydroxide Solution, 10N Synonym: Caustic Soda
Sodium Hydroxide Solutions, 1M/1N Products for Science Education
Chemical Name or Material: Sodium Hydroxide Solutions, 1M/1N CAS: 1310-73-2 Linear Formula: NaOH Formula Weight: 4 Synonym: Caustic Soda
Sodium Hydroxide (NF/EP/BP/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide CAS: 1310-73-2 CAS: 497-19-8 Purity Grade: BP/EP/FCC/NF Assay: 97.0 to 100.5% (EP) Molecular Formula: NaOH Formula Weight: 40 MDL Number: MFCD00003548
Sodium Hydroxide Solution Products for Science Education
Chemical Name or Material: Sodium Hydroxide Solution CAS: 1310-73-2 CAS: 7732-18-5 Physical Form: Liquid Synonym: Caustic Soda
Sodium Hydroxide Solution (1N/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 1N CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (0.25N (N/4)/Certified), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 0.25N (N/4) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: HNaO MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (0.02N (N/50)/Certified), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 0.02N (N/50) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium hydroxide, for analysis, 50% solution in water, ACROS Organics™
Chemical Name or Material: Sodium hydroxide CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: For analysis Assay: 49 to 51% Molecular Formula: HNaO Linear Formula: NaOH Formula Weight: 40 MDL Number: MFCD00003548 Synonym: Caustic soda
Sodium Hydroxide Solution (2.5N/Certified), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution, 2.5N CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: NaOH · H2O MDL Number: MFCD00003548
Sodium hydroxide monohydrate, 99.996% (metals basis), Alfa Aesar™
Chemical Name or Material: Sodium hydroxide monohydrate CAS: 12179-02-1 Physical Form: Crystals Assay Percent Range: 99.996% (metals basis) Molecular Formula: NaOH·H2O Formula Weight: 58.01 MDL Number: MFCD00003548
Sodium Hydroxide Solution (10N/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 10N CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: NaOH MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (0.5N (N/2)/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 0.5N (N/2) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (0.05N (N/20)/Certified), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 0.05N (N/20) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium hydroxide, 98%, Acros Organics
Chemical Name or Material: Sodium hydroxide CAS: 1310-73-2 Purity Grade: Pure Assay Percent Range: 98% Molecular Formula: HNaO Formula Weight: 40 Synonym: Caustic soda
Sodium hydroxide, 98.5%, for analysis, pellets, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium hydroxide CAS: 1310-73-2 Purity Grade: For analysis Physical Form: Pellets Assay: ≥98.0% Assay Percent Range: 98.5% Molecular Formula: HNaO Linear Formula: NaOH Formula Weight: 40 MDL Number: MFCD00003548 Synonym: Caustic soda
Sodium Hydroxide 10N Concentrate (Certified) ACS, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide 10N Concentrate CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade Physical Form: Liquid Molecular Formula: NaOH MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (0.1N (N/10)/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 0.1N (N/10) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium hydroxide, for analysis, micropearls, ACROS Organics™
Chemical Name or Material: Sodium hydroxide CAS: 1310-73-2 Purity Grade: For analysis Assay: ≥99.0% Molecular Formula: HNaO Linear Formula: NaOH Formula Weight: 40 MDL Number: MFCD00003548 Synonym: Caustic soda
Sodium Hydroxide (Granular/NF/FCC/EP/BP/JP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide CAS: 1310-73-2 Purity Grade: BP/EP/FCC/JP/NF Physical Form: Solid Assay: 97.0 to 100.5% (BP) Molecular Formula: NaOH Formula Weight: 40 MDL Number: MFCD00003548 Synonym: Caustic Soda
  spinner