Standard Solutions
Yttrium, AAS standard solution, Y 1000μg/mL, Alfa Aesar™, Specpure™
Chemical Name or Material: Yttrium Physical Form: Liquid Assay Percent Range: Y 1000;mug/mL Molecular Formula: Y2O3 in 5% HNO3 MDL Number: MFCD00011468
Cadmium, plasma standard solution, Cd 1000μg/mL, Alfa Aesar™, Specpure™
Chemical Name or Material: Cadmium CAS: 7697-37-2 CAS: 7440-43-9 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: Cd in 5% HNO3 MDL Number: MFCD00010914
Platinum Plasma Std. Solution, Alfa Aesar™, Specpure™
Chemical Name or Material: Platinum Physical Form: Liquid Molecular Formula: Pt in 20% HCl MDL Number: MFCD00011179
Manganese plasma standard solution, Alfa Aesar™, Specpure™
Chemical Name or Material: Manganese CAS: 7697-37-2 CAS: 7439-96-5 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: Mn in 5% HNO3 MDL Number: MFCD00011111
Bismuth plasma standard solution, Alfa Aesar™, Specpure™
Chemical Name or Material: Bismuth CAS: 7697-37-2 CAS: 7440-69-9 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: Bi in 5% HNO3 MDL Number: MFCD00134033
Vanadium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical
Chemical Name or Material: Vanadium, Reference Standard Solution Name Note: 1000ppm ±1% CAS: 7647-01-0 CAS: 1314-62-1 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00011453
Sulfur in Crude Oil standard solution, 1000μg/g (0.10%), Alfa Aesar™, Specpure™
Chemical Name or Material: Sulfur Name Note: 1000μg/g (0.10%) Physical Form: Liquid Formula Weight: 3350 MDL Number: MFCD00197838
Sulfuric acid, 3.0N Standardized Solution, Alfa Aesar™
Chemical Name or Material: Sulfuric acid CAS: 7664-93-9 Physical Form: Liquid Molecular Formula: H2SO4 Formula Weight: 98.08 MDL Number: MFCD00064589
Silver, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical
Chemical Name or Material: Silver, Reference Standard Solution Name Note: 1000ppm ±1% CAS: 7761-88-8 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00003397
Platinum plasma standard solution, Alfa Aesar™, Specpure™
Chemical Name or Material: Platinum CAS: 7647-01-0 CAS: 7440-06-4 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: Pt in 20% HCl MDL Number: MFCD00011179
Multi-Element QC-7 Check Standard Solution, Specpure™
Chemical Name or Material: Multi-Element QC-7 Check Standard Solution Name Note: Standard Solution Purity Grade: Specpure™ Molecular Formula: 5% HNO3/tr· F? MDL Number: MFCD00210654 UN Number: UN3264
Lactic acid, 1.0N Standardized Solution, Alfa Aesar™
Chemical Name or Material: Lactic acid Name Note: Lactic acid: 9.0%; Water: 91.0% CAS: 50-21-5 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: C3H6O3 Formula Weight: 90.08 MDL Number: MFCD00004520
Magnesium plasma standard solution, Alfa Aesar™, Specpure™
Chemical Name or Material: Magnesium CAS: 7697-37-2 CAS: 7439-95-4 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: Mg in 5% HNO3 MDL Number: MFCD00085308
Potassium, plasma standard solutionμg/mL, Alfa Aesar™, Specpure™
Chemical Name or Material: Potassium CAS: 7697-37-2 CAS: 7757-79-1 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: KNO3 in 5% HNO3 MDL Number: MFCD00133776
Rhodium, plasma standard solution, Rh 1000μg/mL, Alfa Aesar™, Specpure™
Chemical Name or Material: Rhodium Name Note: Rh 1000μg/mL Physical Form: Liquid Molecular Formula: RhCl3 in 20% HCl MDL Number: MFCD00011201
Silver plasma standard solution, Alfa Aesar™, Specpure™
Chemical Name or Material: Silver Name Note: plasma standard solution Physical Form: Liquid Molecular Formula: Ag in 5% HNO3 MDL Number: MFCD00003397
Copper, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical
Chemical Name or Material: Copper, Reference Standard Solution Name Note: 1000ppm ±1% CAS: 19004-19-4 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00010965
Strontium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical
Chemical Name or Material: Strontium, Reference Standard Solution Name Note: 1000ppm ±1% CAS: 1633-05-2 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00134060
Rubidium, plasma standard solution, Rb 1000μg/mL, Alfa Aesar™, Specpure™
Chemical Name or Material: Rubidium CAS: 7697-37-2 CAS: 584-09-8 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: Rb2CO3 in 5% HNO3 MDL Number: MFCD00134055
Aluminium plasma standard solution, Alfa Aesar™, Specpure™
Chemical Name or Material: Aluminium Name Note: plasma standard solution, Al 1000μg/mL CAS: 7697-37-2 CAS: 7784-27-2 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: Al(NO3)3 in 5% HNO3 MDL Number: MFCD00134029
Boron plasma standard solution, Alfa Aesar™, Specpure™
Chemical Name or Material: Boron Name Note: plasma standard solution, B 1000μg/mL Physical Form: Liquid Molecular Formula: H3BO3 in H2O MDL Number: MFCD00134034
Barium, Reference Standard Solution (1000ppm ± 1%/Certified), Fisher Chemical
Chemical Name or Material: Barium, Reference Standard Solution Name Note: 1000ppm ± 1% CAS: 10361-37-2 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00134031
Gallium, Plasma standard solution, Ga 1000μg/mL, Alfa Aesar™, Specpure™
Chemical Name or Material: Gallium CAS: 7697-37-2 CAS: 7440-55-3 CAS: 7647-01-0 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: Ga in 5% HNO3/tr· HCl MDL Number: MFCD00134045
Antimony, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical
Chemical Name or Material: Antimony, Reference Standard Solution Name Note: 1000ppm ±1% CAS: 10025-91-9 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00134030
Lead plasma standard solution, Alfa Aesar™, Specpure™
Chemical Name or Material: Lead CAS: 7697-37-2 CAS: 7439-92-1 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: Pb in 5% HNO3 MDL Number: MFCD00134050
Molybdenum, plasma standard solution, Mo 1000μg/mL, Alfa Aesar™, Specpure™
Chemical Name or Material: Molybdenum Name Note: Mo 1000μg/mL Physical Form: Liquid Molecular Formula: Mo in 5% HNO3/tr· HF MDL Number: MFCD00003465
Hydrochloric acid, 1.0N Standardized Solution, Alfa Aesar™
Chemical Name or Material: Hydrochloric acid CAS: 7647-01-0 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: HCl Formula Weight: 36.46 MDL Number: MFCD00011324
Sodium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical
Chemical Name or Material: Sodium, Reference Standard Solution Name Note: 1000ppm ±1% CAS: 7647-14-5 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: NaCl MDL Number: MFCD00085307
Gold Plasma Std. Solution, Alfa Aesar™, Specpure™
Chemical Name or Material: Gold Physical Form: Liquid Molecular Formula: Au in 20% HCl MDL Number: MFCD00003436
Lutetium, AAS standard solution, Lu 1000μg/mL, Alfa Aesar™, Specpure™
Chemical Name or Material: Lutetium CAS: 7697-37-2 CAS: 12032-20-1 CAS: 7732-18-5 Purity Grade: AAS Standard Physical Form: Liquid Molecular Formula: Lu2O3 in 5% HNO3 MDL Number: MFCD00011098
  spinner