Strontium -Sr
Strontium nitrate, anhydrous, 98%, Alfa Aesar™
Name Note: Anhydrous CAS: 10042-76-9 Physical Form: Crystalline or Powder Assay Percent Range: 98% Molecular Formula: N2O6Sr Formula Weight: 211.63 MDL Number: MFCD00011248
Strontium carbonate, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Strontium carbonate CAS: 1633-05-2 Physical Form: Powder Assay Percent Range: 99.99% (metals basis) Molecular Formula: SrCO3 Formula Weight: 147.63 MDL Number: MFCD00011250
Strontium chloride, ultra dry, 99.995% (metals basis), Alfa Aesar™
Chemical Name or Material: Strontium chloride CAS: 10476-85-4 Physical Form: -10 Mesh Beads, Ampouled Under Argon Assay Percent Range: 99.995% (metals basis) Molecular Formula: SrCl2 Formula Weight: 158.53 MDL Number: MFCD00011249
Strontium hydroxide octahydrate, 99% (metals basis), Alfa Aesar™
Chemical Name or Material: Strontium hydroxide octahydrate CAS: 1311-10-0 Physical Form: Crystalline Assay Percent Range: 99% (metals basis) Molecular Formula: Sr(OH)2·8H2O Formula Weight: 265.76; (121.60 Anhydrous) MDL Number: MFCD00150395
Strontium molybdenum oxide, 99.9% (excluding Ba), Ba <0.3%; Alfa Aesar™
Name Note: Ba < 0.3% CAS: 13470-04-7 Physical Form: Powder Assay Percent Range: 99.9% (excluding Ba) Molecular Formula: SrMoO4 Formula Weight: 247.56 MDL Number: MFCD00049550 Synonym: Strontium molybdate
Alfa Aesar™ Strontium Titanium Oxide Substrate, 5 x 5 x 1mm, Polished One Side, 100 Orientation, Alfa Aesar™
Chemical Name or Material: Strontium Titanium Oxide Substrate CAS: 12060-59-2 Name Note: Polished One Side Molecular Formula: SrTiO3 MDL Number: MFCD00049554
Strontium aluminate, 99.5% (metals basis), Alfa Aesar™
Chemical Name or Material: Strontium aluminate CAS: 12004-37-4 Physical Form: -100 Mesh Powder Assay Percent Range: 99.5% (metals basis) Molecular Formula: SrAl2O4 Formula Weight: 205.58 MDL Number: MFCD00054054
Strontium Titanium Oxide Substrate, 6.25 x 6.25 x 0.5mm, Polished One Side, 110 Orientation, Alfa Aesar™
Chemical Name or Material: Strontium Titanium Oxide Substrate Name Note: Polished One Side CAS: 12060-59-2 Physical Form: Substrate Molecular Formula: SrTiO3 MDL Number: MFCD00049554
Alfa Aesar™ Strontium Titanium Oxide Substrate, 10 x 10 x 1mm, Polished One Side, 110 Orientation, Alfa Aesar™
Chemical Name or Material: Strontium Titanium Oxide Substrate CAS: 12060-59-2 Name Note: Polished One Side Molecular Formula: SrTiO3 MDL Number: MFCD00049554
Alfa Aesar™ Strontium Titanium Oxide Substrate, 10 x 10 x 1mm, Polished One Side, 100 Orientation, Alfa Aesar™
Chemical Name or Material: Strontium Titanium Oxide Substrate CAS: 12060-59-2 Name Note: Polished One Side Molecular Formula: SrTiO3 MDL Number: MFCD00049554
Strontium Nitrate Products for Science Education
Chemical Name or Material: Strontium Nitrate CAS: 10042-76-9 Purity Grade: Laboratory Grade Assay Percent Range: >95% Linear Formula: Sr(NO3)2 Formula Weight: 211.63
Strontium perchlorate trihydrate, 98%, Alfa Aesar™
Chemical Name or Material: Strontium perchlorate trihydrate CAS: 15650-09-6 Physical Form: Crystalline Assay Percent Range: 98% Molecular Formula: Cl2O8Sr·3H2O Formula Weight: 340.54 (286.52 Anhydrous) MDL Number: MFCD09039117
Strontium oxide, 99.5% (metals basis), SrO ≈97%, Alfa Aesar™
Chemical Name or Material: Strontium oxide CAS: 1314-11-0 Physical Form: -100 Mesh Powder Assay Percent Range: 99.5% (metals basis) Molecular Formula: SrO Formula Weight: 103.62 MDL Number: MFCD00049558
Alfa Aesar™ Strontium Titanium Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, 100 Orientation, Alfa Aesar™
Chemical Name or Material: Strontium Titanium Oxide Substrate CAS: 12060-59-2 Name Note: Polished One Side Molecular Formula: SrTiO3 MDL Number: MFCD00049554
Alfa Aesar™ Strontium Titanium Oxide Substrate, 5 x 5 x 1mm, Polished One Side, 110 Orientation, Alfa Aesar™
Chemical Name or Material: Strontium Titanium Oxide Substrate CAS: 12060-59-2 Name Note: Polished One Side Molecular Formula: SrTiO3 MDL Number: MFCD00049554
Strontium Titanium Oxide Substrate, 5 x 5 x 1mm, Polished Two Sides, 100 Orientation, Alfa Aesar™
Chemical Name or Material: Strontium Titanium Oxide Substrate Name Note: Polished Two Sides CAS: 12060-59-2 Physical Form: Substrate Molecular Formula: SrTiO3 MDL Number: MFCD00049554
Strontium nitrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Strontium nitrate CAS: 10042-76-9 Physical Form: Crystalline, Packaged Under Argon Assay Percent Range: 99.999% (metals basis) Molecular Formula: Sr(NO3)2 Formula Weight: 211.63 MDL Number: MFCD00011248
Strontium Titanium Oxide Substrate, 10 x 10 x 1mm, Polished Two Sides, 100 Orientation, Alfa Aesar™
Chemical Name or Material: Strontium Titanium Oxide Substrate Name Note: Polished Two Sides CAS: 12060-59-2 Physical Form: Substrate Molecular Formula: SrTiO3 MDL Number: MFCD00049554
Strontium nitrate, ACS, 99.0% min., Alfa Aesar™
Chemical Name or Material: Strontium nitrate Name Note: For analysis ACS CAS: 10042-76-9 Physical Form: Crystalline Assay Percent Range: 99.0% min. Molecular Formula: Sr(NO3)2 Formula Weight: 211.63 MDL Number: MFCD00011248
Strontium nitrate, 99+%, ACS reagent, ACROS Organics™
Chemical Name or Material: Strontium nitrate Name Note: 99+% CAS: 10042-76-9 Purity Grade: ACS Reagent Grade Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: N2O6Sr Linear Formula: Sr(NO3)2 Formula Weight: 211.63 MDL Number: MFCD00011248
Strontium Chloride, Anhydrous, 99.5% (Metal Basis), Alfa Aesar™
Chemical Name or Material: Strontium Chloride CAS: 10476-85-4 Physical Form: -40 Mesh Powder Assay Percent Range: 99.5%, metal basis Molecular Formula: Cl2Sr Formula Weight: 158.53 MDL Number: MFCD00011249
Strontium Fluoride, 99.99% (Metal Basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Strontium Fluoride CAS: 7783-48-4 Physical Form: Powder Assay Percent Range: 99.99%, metal basis Molecular Formula: SrF2 Formula Weight: 125.62 MDL Number: MFCD00011251
Strontium carbonate, 99%, Alfa Aesar™
Chemical Name or Material: Strontium carbonate, 99%, Ba 1% CAS: 1633-05-2 Physical Form: -325 Mesh, Typically 1 Micron Powder Assay Percent Range: 99% Molecular Formula: SrCO3 Formula Weight: 147.63 MDL Number: MFCD00011250
Alfa Aesar™ Strontium Titanium Oxide Substrate, 5 x 5 x 0.5mm, Polished One Side, 100 Orientation, Alfa Aesar™
Chemical Name or Material: Strontium Titanium Oxide Substrate CAS: 12060-59-2 Name Note: Polished One Side Molecular Formula: SrTiO3 MDL Number: MFCD00049554
Strontium nitrate, 99+%, for analysis, ACROS Organics™
Chemical Name or Material: Strontium nitrate Name Note: p.a. CAS: 10042-76-9 Purity Grade: For analysis Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: N2O6Sr Linear Formula: Sr(NO3)2 Formula Weight: 211.63 MDL Number: MFCD00011248
Alfa Aesar™ Strontium Titanium Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, 111 Orientation, Alfa Aesar™
Chemical Name or Material: Strontium Titanium Oxide Substrate CAS: 12060-59-2 Name Note: Polished One Side Molecular Formula: SrTiO3 MDL Number: MFCD00049554
Strontium Chloride, Hexahydrate Products for Science Education
Chemical Name or Material: Strontium Chloride, Hexahydrate CAS: 10025-70-4 Purity Grade: Laboratory Grade Assay Percent Range: >99% Linear Formula: SrCl2·6H2O Formula Weight: 266.62
Strontium carbonate, 97.5%, Alfa Aesar™
Chemical Name or Material: Strontium carbonate CAS: 1633-05-2 Physical Form: Powder Assay Percent Range: 97.5% Molecular Formula: SrCO3 Formula Weight: 147.63 MDL Number: MFCD00011250
Strontium Chloride Hexahydrate (Crystalline/Certified ACS), Fisher Chemical
Chemical Name or Material: Strontium Chloride Hexahydrate CAS: 10025-70-4 Purity Grade: Certified ACS Grade Physical Form: Solid Assay: 99 to 103% Assay Percent Range: 99 to 103 % Molecular Formula: Cl2Sr · 6 H2O Formula Weight: 266.62 MDL Number: MFCD00149865
  spinner