Succinic Acid
Succinic Acid 99%, ACROS Organics™
Chemical Name or Material: Succinic acid Name Note: 99% CAS: 110-15-6 Assay Percent Range: 99% Molecular Formula: C4H6O4 Linear Formula: HO2CCH2CH2CO2H Formula Weight: 118.09 MDL Number: MFCD00002789 Synonym: Butanedioic acid
Succinic acid, disodium salt hexahydrate, 97%, ACROS Organics™
Chemical Name or Material: Succinic acid, disodium salt hexahydrate Name Note: 97% CAS: 6106-21-4 Assay: 96.0 to 103.0% (Total base) Assay Percent Range: 97% Molecular Formula: C4H4Na2O4·6H2O Formula Weight: 270.13 Synonym: Disodium succinate hexahydrate
Succinic acid, ACS reagent, ACROS Organics™
Chemical Name or Material: Succinic acid Name Note: p.a. CAS: 110-15-6 Purity Grade: ACS Reagent Grade Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C4H6O4 Linear Formula: HO2CCH2CH2CO2H Formula Weight: 118.09 MDL Number: MFCD00002789 Synonym: Butanedioic acid
Succinic Acid Products for Science Education
Chemical Name or Material: Succinic Acid CAS: 110-15-6 Linear Formula: (CH2COOH)2 Formula Weight: 118.09 Synonym: Butanedioic Acid
Succinic acid, disodium salt, 99%, anhydrous, ACROS Organics™
Chemical Name or Material: Succinic acid, disodium salt Name Note: 99% CAS: 150-90-3 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C4H4Na2O4 Linear Formula: NaO2CCH2CH2CO2Na Formula Weight: 162.05 MDL Number: MFCD00002790 Synonym: Disodium succinate, anhydrous
Succinic Acid (Crystalline/Certified), Fisher Chemical
Chemical Name or Material: Succinic Acid CAS: 110-15-6 Purity Grade: Certified Grade Physical Form: Powder/Solid Assay: 99.0 to 100.3% Assay Percent Range: 99.0 to 100.3 % Molecular Formula: C4H6O4 Linear Formula: HOOCCH2CH2COOH Formula Weight: 118.09 MDL Number: MFCD00002789 Synonym: Amber Acid
Succinic Acid (White Crystals), Fisher BioReagents
Chemical Name or Material: Succinic Acid CAS: 110-15-6 Physical Form: Powder/Solid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C4H6O4 Linear Formula: HOOCCH2CH2COOH Formula Weight: 118.09 Synonym: Dihydrofumaric Acid
Succinic Acid ACS AR Granular, Macron Fine Chemicals™
Chemical Name or Material: Succinic Acid CAS: 110-15-6 Purity Grade: ACS Grade Physical Form: Granular Molecular Formula: HOCOCH2CH2COOH Formula Weight: 118.09
Succinic acid, ACS, ≥99%, MP Biomedicals™
Chemical Name or Material: Succinic acid CAS: 110-15-6 Physical Form: Powder Assay Percent Range: ≥99% Molecular Formula: C4H6O4 Formula Weight: 118.09 Synonym: Amber acid
Succinic Acid, MP Biomedicals
Chemical Name or Material: Succinic Acid CAS: 110-15-6 Formula Weight: 118.09
Succinic Acid, Disodium Salt, hexahydrate, 98% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Succinic Acid, Disodium Salt, hexahydrate Name Note: For Synthesis CAS: 6106-21-4 Physical Form: Crystals Assay Percent Range: 98% min. Molecular Formula: NaOOCCH2CH2COONa·6H2O Formula Weight: 27.15 Synonym: Butanedioic acid
  spinner