Sulfamic Acid
Sulfamic Acid, 99.3 to 100.3%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sulfamic Acid Name Note: For Synthesis CAS: 5329-14-6 Physical Form: Crystals Assay Percent Range: 99.3 to 100.3% Molecular Formula: H3NO3S Linear Formula: HCH2NSOCH3H Formula Weight: 97.8 Synonym: Amidosulfonic Acid
Sulfamic Acid, Practical grade, 97% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sulfamic Acid CAS: 5329-14-6 Purity Grade: Practical Grade Physical Form: Crystalline Assay Percent Range: 97%, min. Molecular Formula: NH2SO3H Linear Formula: NH2SO3H Formula Weight: 97.9 Synonym: Amidosulfonic Acid
Sulfamic Acid, 99.3 to 100.3%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sulfamic Acid CAS: 5329-14-6 Name Note: For Synthesis Molecular Formula: H3NO3S Linear Formula: HCH2NSOCH3H Formula Weight: 97.8 Synonym: Amidosulfonic acid
  spinner