Surfactants and Wetting Agents
Hexadecyltrimethylammonium bromide, 99+%, ACROS Organics™
Chemical Name or Material: Hexadecyltrimethylammonium Bromide Assay: ≥99.0% (Total base) Assay Percent Range: 99+% Molecular Formula: C19H42BrN Linear Formula: CH3(CH2)15N(CH3)3Br Formula Weight: 364.45 Synonym: CTABr
Aerosol™ OT Solution, 75% (w/w), Fisher Chemical
Chemical Name or Material: Aerosol™ OT Solution Name Note: 75% (w/w) CAS: 64-17-5 MDL Number: MFCD00012455
Sarkosyl, 95%, ACROS Organics™
Chemical Name or Material: Sarkosyl CAS: 137-16-6 Assay: ≥95% Assay Percent Range: 95% Molecular Formula: C15H28NNaO3 Linear Formula: NaCO2CH2N(CH3)CO(CH2)10CH3 Formula Weight: 293.39 MDL Number: MFCD00042728
Tween™ 85, ACROS Organics™
Chemical Name or Material: Tween™ 85 CAS: 9005-70-3 Synonym: Polyoxyethylene (20) Sorbitan Trioleate
Triton™ X-100, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Triton™ X-100 CAS: 9002-93-1 Molecular Formula: C34H62O11 Formula Weight: 646.85 MDL Number: MFCD00132505 Synonym: Polyoxyethylene(10) octylphenyl ether
Brij™ 58, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Brij™ 58 CAS: 9004-95-9 Molecular Formula: C56H114O21 Linear Formula: C16H33(OCHCH)0OH Formula Weight: 1123.5 MDL Number: MFCD00080892 Synonym: Polyoxyethylene(20) cetyl ether
Dimethylethylhexadecylammonium bromide, 99%, ACROS Organics™
Chemical Name or Material: Dimethylethylhexadecylammonium bromide Name Note: 99% CAS: 124-03-8 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C20H44BrN Linear Formula: CH3(CH2)15N(CH3)2(C2H5)Br Formula Weight: 378.48 MDL Number: MFCD00011774 Synonym: Cetyldimethylethylammonium bromide
Brij™ 35, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Brij™ 35 CAS: 9002-92-0 Molecular Formula: C58H118O24 Linear Formula: C12H25(OCH2CH2)23OH Formula Weight: 1199.56 Synonym: Polyoxyethylene(23)lauryl ether
Detergent, Neutral Reagent, Fisher Chemical
Chemical Name or Material: Detergent, Neutral Reagent CAS: 7732-18-5 CAS: 151-21-3 CAS: 6381-92-6 CAS: 112-27-6 CAS: 1303-96-4 MDL Number: MFCD00283948
Tween™ 20, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tween™ 20 CAS: 9005-64-5 MDL Number: MFCD00165986 Synonym: Polyoxyethylene(20)sorbitan monolaurate
Triton X-405, 70% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Triton™ X-405 Name Note: 70% Solution in Water CAS: 9002-93-1 CAS: 7732-18-5 Assay: ≥65% Molecular Formula: C94H182O41 Formula Weight: 1968.47 MDL Number: MFCD01779734
Tween™ 40, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tween™ 40 CAS: 9005-66-7 MDL Number: MFCD00165345 Synonym: Polyoxyethylene (20) Sorbitan Monopalmitate
Dodecyl sulfate sodium salt, 99%, for biochemistry, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dodecyl Sulfate Sodium Salt Name Note: 99% CAS: 151-21-3, 7757-82-6, 7647-14-5 CAS: 7757-82-6 CAS: 7647-14-5 Purity Grade: For biochemistry Assay: ≥98.5% (GC) Assay Percent Range: 99% Molecular Formula: C12H25NaO4S Linear Formula: CH3(CH2)11OSO3Na Formula Weight: 288.38 MDL Number: MFCD00036175 Synonym: Dodecyl Sodium Sulfate
Tween™ 80, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tween™ 80 CAS: 9005-65-6 MDL Number: MFCD00082107 Synonym: Polyoxyethylene(20)sorbitan monooleate
Brij™ 30, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Brij™ 30 CAS: 9002-92-0 Molecular Formula: C20H42O5 Linear Formula: C12H25(OCH2CH2)4OH Formula Weight: 362.54 MDL Number: MFCD00043063 Synonym: Polyoxyethylene(4)lauryl ether
Acid Detergent Solution, Fisher Chemical
Chemical Name or Material: Acid Detergent Solution CAS: 7664-93-9 CAS: 57-09-0 CAS: 7732-18-5 MDL Number: MFCD00282010
Sodium Lauryl Sulfate (Powder/NF/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Lauryl Sulfate CAS: 151-21-3 Purity Grade: FCC/NF Physical Form: Solid Molecular Formula: C12H25NaO4S Linear Formula: CH3(CH2)10CH2OSO3Na MDL Number: MFCD00036175 Synonym: Dodecylsulfuric Acid Sodium Salt
CHAPS, ACROS Organics™
Chemical Name or Material: CHAPS CAS: 75621-03-3 Molecular Formula: C32H58N2O7S Formula Weight: 614.9 MDL Number: MFCD00012116 Synonym: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
Triton X-100, Alfa Aesar
Chemical Name or Material: Triton X-100 CAS: 9002-93-1
Brij™ 35, 30 w/v % solution in water, ACROS Organics™
Chemical Name or Material: Brij™ 35 CAS: 9002-92-0 CAS: 7732-18-5 Assay: 29 to 31% Molecular Formula: C58H118O24 Linear Formula: C12H25(OCH2CH2)23OH Formula Weight: 1199.56 MDL Number: MFCD00080891
Aerosol™ OT, Anhydrous Solid, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Aerosol™ OT, Solid Anhydrous CAS: 577-11-7 Physical Form: Solid Molecular Formula: C20H37O7S · Na Linear Formula: CH3(CH2)3CH(C2H5)CH2O2CCH2CH(SO3Na)CO2CH2CH(C2H5)(CH2)3CH3 Formula Weight: 444 MDL Number: MFCD00012455 Synonym: AOT
Dodecyl sulfate sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dodecyl sulfate sodium salt CAS: 151-21-3 CAS: 7757-82-6 CAS: 7647-14-5 Purity Grade: For molecular biology Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C12H25NaO4S Linear Formula: CH3(CH2)11OSO3Na Formula Weight: 288.38 MDL Number: MFCD00036175 Synonym: Dodecyl Sodium Sulfate
Dodecyl Sulfate, Sodium Salt 85%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dodecyl sulfate sodium salt CAS: 151-21-3 CAS: 7757-82-6 CAS: 7647-14-5 Purity Grade: Pure Assay: ≥80% (C12H25NaO4S) Assay Percent Range: 85% Molecular Formula: C12H25NaO4S Linear Formula: CH3(CH2)11OSO3Na Formula Weight: 288.38 Synonym: Dodecyl Sodium Sulfate
Triton™ X-100, 98%, for molecular biology, DNAse, RNAse and Protease free, ACROS Organics™
Chemical Name or Material: Triton™ X-100 Name Note: For molecular biology, DNAse, RNAse and Protease free CAS: 9002-93-1 Purity Grade: For molecular biology Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C34H62O11 Formula Weight: 646.85 MDL Number: MFCD00132505 Synonym: Polyethylene glycol mono [4-(1,1,3,3-tetramethylbutyl)phenyl] ether
Lithium dodecyl sulfate, +99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lithium dodecyl sulfate Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C12H25LiO4S Linear Formula: CH3(CH2)11OSO3Li Formula Weight: 272.33 MDL Number: MFCD00007467 Synonym: Dodecyl lithium sulfate
Triton™ X-114, ACROS Organics™
Chemical Name or Material: Triton™ X-114 CAS: 9002-93-1 Molecular Formula: 7·5 (C2H4O) Formula Weight: 537 MDL Number: MFCD01778132 Synonym: Polyoxyethylene(8) octylphenyl ether
Brij™ 98, ACROS Organics™
Chemical Name or Material: Brij™ 98 CAS: 9004-98-2 Molecular Formula: C58H116O21 Linear Formula: C18H35(OCHCH)nOH Formula Weight: 1149.56 MDL Number: MFCD00074369 Synonym: Polyoxyethylene(20) oleyl ether
Soap, Castile Products for Science Education
Chemical Name or Material: Soap, Castile Assay Percent Range: 100% w/v
Tween™ 60, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tween™ 60 CAS: 9005-67-8 MDL Number: MFCD01780110
Aerosol™ OT Solution, 10% (w/w), Fisher Chemical
Chemical Name or Material: Aerosol™ OT Solution, 10% (w/w) Name Note: 10% (w/w) CAS: 577-11-7 CAS: 111-76-2 CAS: 7732-18-5 CAS: 64-17-5 CAS: 104-76-7 Assay Percent Range: 10% w/w MDL Number: MFCD00012455
  spinner