Tantalum -Ta
Tantalum(V) Chloride, 99.99% (Metal Basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Tantalum(V) Chloride Name Note: Ampouled under argon CAS: 7721-01-9 Physical Form: Solid, Resublimed, Crystalline Assay Percent Range: 99.99%, metal basis Molecular Formula: TaCl5 Formula Weight: 358.21 MDL Number: MFCD00011253
Tantalum foil, 99.95% (metals basis excluding Nb), Alfa Aesar™
Chemical Name or Material: Tantalum Name Note: Annealed CAS: 7440-25-7 Physical Form: Foil, 0.5mm (0.02 in.) thick Assay Percent Range: 99.95% (Metals basis excluding Nb) MDL Number: MFCD00011252
Tantalum Powder, -22 mesh, Puratronic™, 99.98% (Metals basis, Nb 50ppm), Alfa Aesar™
Chemical Name or Material: Tantalum CAS: 7440-25-7 Physical Form: Powder Assay Percent Range: 99.98% (Metals basis, Nb 50ppm) MDL Number: MFCD00011252
Tantalum Wire, 0.025mm (0.001 in.) dia., hard, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Tantalum Name Note: Hard CAS: 7440-25-7 Physical Form: Wire, 0.025mm (0.001 in.) dia. Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00011252
Tantalum powder, 99.98% (metal basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Tantalum CAS: 7440-25-7 Physical Form: Powder Assay Percent Range: 99.98% (Metals basis) MDL Number: MFCD00011252
Tantalum(V) chloride, 99.99%, (trace metal basis), ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tantalum(V) chloride CAS: 7721-01-9 Purity Grade: Trace Metal Basis Assay Percent Range: 99.99% Molecular Formula: Cl5Ta Linear Formula: TaCl5 Formula Weight: 358.2 MDL Number: MFCD00011253
Tantalum wire, annealed, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Tantalum Name Note: Annealed CAS: 7440-25-7 Physical Form: Wire, 0.05mm (0.002 in.) dia. Assay Percent Range: 99.9% (Metals basis, Nb 50ppm) MDL Number: MFCD00011252
Tantalum Foil, 0.127mm (0.005 in.) thick, annealed, 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Tantalum Name Note: Annealed CAS: 7440-25-7 Physical Form: Foil, 0.127mm (0.005 in.) thick Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011252
Tantalum powder, 99.97% (metal basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Tantalum CAS: 7440-25-7 Physical Form: Powder Assay Percent Range: 99.97% (Metals basis, Nb 50ppm) MDL Number: MFCD00011252
Tantalum(V) Fluoride, 99.9% (Metal Basis), Alfa Aesar™
Chemical Name or Material: Tantalum(V) Fluoride CAS: 7783-71-3 Physical Form: Powder Assay Percent Range: 99.9%, metal basis Molecular Formula: TaF5 Formula Weight: 275.94 MDL Number: MFCD00042542 Synonym: Tantalum pentafluoride
Tantalum Powder, -325 mesh, 99.9% (Metals basis, 99.6% purity includes H, N, C, O), Alfa Aesar™
Chemical Name or Material: Tantalum CAS: 7440-25-7 Physical Form: Powder Assay Percent Range: 99.9% (Metals basis, 99.6% purity includes H, N, C, O) MDL Number: MFCD00011252
Tantalum(V) oxide, 99% (metal basis), Alfa Aesar™
Chemical Name or Material: Tantalum(V) oxide CAS: 1314-61-0 Physical Form: Powder Assay Percent Range: 99% (metals basis) Molecular Formula: Ta2O5 Formula Weight: 441.89 MDL Number: MFCD00011254 Synonym: Tantalum pentoxide
Tantalum(V) oxide, 99.85% (metals basis), Alfa Aesar™
Chemical Name or Material: Tantalum(V) oxide CAS: 1314-61-0 Physical Form: -325 Mesh Powder Assay Percent Range: 99.85% (metals basis) Molecular Formula: Ta2O5 Formula Weight: 441.89 MDL Number: MFCD00011254
Tantalum wire, 99.95% (metals basis), Alfa Aesar™
Chemical Name or Material: Tantalum Name Note: Annealed CAS: 7440-25-7 Physical Form: Wire, 0.5mm (0.02 in.) dia. Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011252
Tantalum Thinfoil, 99.95% (metals basis), Alfa Aesar™
Chemical Name or Material: Tantalum Thinfoil CAS: 7440-25-7 Physical Form: ∽2.1g/100X100mm, Certified Pinhole Free Assay Percent Range: 99.95% (metals basis) MDL Number: MFCD00011252
Tantalum Rod, 2.8mm (0.11 in.) dia., 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Tantalum CAS: 7440-25-7 Physical Form: Rod, 2.8mm (0.11 in.) dia. Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011252
Tantalum Foil, 1.0mm (0.04 in.) thick, 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Tantalum CAS: 7440-25-7 Physical Form: Foil, 1.0mm (0.04 in.) thick Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011252
Tantalum Foil, 0.127mm (0.005 in.) thick, hard, 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Tantalum Name Note: Hard CAS: 7440-25-7 Physical Form: Foil, 0.127mm (0.005 in.) thick Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011252
Tantalum foil, 99.95% (metals basis), Alfa Aesar™
Chemical Name or Material: Tantalum Name Note: Hard CAS: 7440-25-7 Physical Form: Foil, 0.025mm (0.001 in.) thick Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011252
Tantalum(V) oxide, 99.99%, (trace metal basis), ACROS Organics™
Chemical Name or Material: Tantalum(V) oxide CAS: 1314-61-0 Purity Grade: For trace metal basis analysis Assay Percent Range: 99.99% Molecular Formula: O5Ta2 Linear Formula: Ta2O5 Formula Weight: 441.89 MDL Number: MFCD00011254
Tantalum slug, 99.95% (metals basis), Alfa Aesar™
Chemical Name or Material: Tantalum CAS: 7440-25-7 Physical Form: Slug, 3.175mm (0.125 in.) dia. X 3.175mm (0.125 in.) length Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011252
Tantalum(V) Fluoride 98%, Acros Organics
Chemical Name or Material: Tantalum(V) fluoride Name Note: 98% CAS: 7783-71-3 Assay: ≥97.5% (ex F) Molecular Formula: TaF5 Linear Formula: TaF5 Formula Weight: 275.94 MDL Number: MFCD00042542
  spinner