Tartaric Acid
L-(+)-Tartaric acid dipotassium salt, 97%, Acros Organics
Chemical Name or Material: L-(+)-Tartaric acid dipotassium salt Assay Percent Range: 97% Molecular Formula: C4H4K2O6 Formula Weight: 226.27
DL-Tartaric acid, 99.5%, ACROS Organics™
Chemical Name or Material: DL-Tartaric acid Assay: ≥99.4% Assay Percent Range: 99.50% Molecular Formula: C4H6O6 Linear Formula: HO2CCH(OH)CH(OH)CO2H Formula Weight: 150.09
L(+)-Tartaric acid, 99+%, Acros Organics Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L(+)-Tartaric acid Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C4H6O6 Linear Formula: HO2CCH(OH)CH(OH)CO2H Formula Weight: 150.09 Synonym: L(+)-Dihydroxysuccinic acid
L(+)-Tartaric acid diammonium salt, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L(+)-Tartaric acid diammonium salt Name Note: 99% CAS: 3164-29-2 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C4H12N2O6 Linear Formula: [-CH(OH)CO2NH4]2 Formula Weight: 184.15 MDL Number: MFCD00013073 Synonym: Ammonium L-tartrate
D(-)-Tartaric Acid (unnatural Tartaric Acid) 99%, ACROS Organics™
Chemical Name or Material: D(-)-Tartaric acid Name Note: 99% CAS: 147-71-7 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C4H6O6 Linear Formula: HO2CCH(OH)CH(OH)CO2H Formula Weight: 150.09 MDL Number: MFCD00004238 Synonym: D(-)-Dihydroxysuccinic acid
L-(+)-Tartaric Acid (Powder/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-(+)-Tartaric Acid CAS: 87-69-4 Purity Grade: Certified ACS Grade Physical Form: Solid Assay: ≥99.0% Assay Percent Range: ≥99.0 % Molecular Formula: C4H6O6 Linear Formula: HOOC(CHOH)2COOH Formula Weight: 150.09 MDL Number: MFCD00064207 Synonym: 2, 3-Dihydroxybutanedioic Acid
Tartaric Acid Products for Science Education
Chemical Name or Material: Tartaric Acid CAS: 87-69-4 Purity Grade: Laboratory Grade Assay Percent Range: >99% w/v Linear Formula: HO2CCH(OH)CH(OH)CO2H Formula Weight: 150.09 Synonym: Threaric Acid
L-(+)-Tartaric Acid (Fine Granular/NF/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-(+)-Tartaric Acid CAS: 87-69-4 Purity Grade: FCC/NF Physical Form: Solid Assay: 99.7 to 100.5% (NF) Molecular Formula: C4H6O6 Linear Formula: HOOC(CHOH)2COOH Formula Weight: 150.09 MDL Number: MFCD00064207 Synonym: 2, 3-Dihydroxybutanedioic Acid
L-(+)-Tartaric Acid (Granular/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-(+)-Tartaric Acid CAS: 87-69-4 Purity Grade: Certified ACS Grade Physical Form: Solid Assay: ≥99.0% Assay Percent Range: ≥99.0 % Molecular Formula: C4H6O6 Linear Formula: HOOC(CHOH)2COOH Formula Weight: 150.09 MDL Number: MFCD00064207 Synonym: 2, 3-Dihydroxybutanedioic Acid
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L+-Tartaric acid, disodium salt, dihydrate CAS: 6106-24-7 Purity Grade: ACS Reagent Grade Assay: 99.0 to 101.0% Assay Percent Range: 99+% Molecular Formula: C4H4Na2O6·2H2O Linear Formula: (CHOHCOONa)2·2H2O Formula Weight: 230.08 MDL Number: MFCD00150035 Synonym: Disodium tartrate
L-(+)-Tartaric acid, ACS
Chemical Name or Material: L-(+)-Tartaric Acid CAS: 87-69-4 Purity Grade: ACS Grade Molecular Formula: HOOC(CHOH)2COOH Formula Weight: 150.09 MDL Number: MFCD00064207 Beilstein: 1725147 Merck Index: 14,9070
L(+)-Tartaric acid, ACS reagent, Acros Organics
Chemical Name or Material: L(+)-Tartaric acid Name Note: 99+% CAS: 87-69-4 Purity Grade: ACS Reagent Grade Assay: ≥99.0% Molecular Formula: C4H6O6 Linear Formula: HO2CCH(OH)CH(OH)CO2H Formula Weight: 150.09 MDL Number: MFCD00064207 Synonym: L(+)-Dihydroxysuccinic acid
Tartaric Acid NF/FCC Fine Granular, Macron Fine Chemicals™
Chemical Name or Material: Tartaric Acid CAS: 87-69-4 Purity Grade: FCC/NF Physical Form: Fine Granular Molecular Formula: HOCO(CHOH)2COOH Formula Weight: 150.09
Tartaric Acid AR Granular, Macron Fine Chemicals™
Chemical Name or Material: Tartaric Acid CAS: 87-69-4 Physical Form: Granular Molecular Formula: HOCO(CHOH)2COOH Formula Weight: 150.09
Tartaric Acid, 2% (w/v) Aqueous Solution, Certified APHA Reagent, Ricca Chemical
Chemical Name or Material: Tartaric Acid Name Note: 2% (w/v) Aqueous Solution CAS: 7732-18-5 CAS: 87-69-4 Physical Form: Liquid
Tartaric Acid, 50% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Tartaric Acid Name Note: Aqueous Solution CAS: 7732-18-5 CAS: 87-69-4 Physical Form: Liquid Assay Percent Range: 50% (w/v)
Tartaric Acid ACS AR Powder, Macron Fine Chemicals™
Chemical Name or Material: Tartaric Acid CAS: 87-69-4 Purity Grade: ACS Grade Physical Form: Powder Molecular Formula: HOCO(CHOH)2COOH Formula Weight: 150.09
Tartaric Acid ACS, Ricca Chemical
Chemical Name or Material: Tartaric Acid CAS: 87-69-4 CAS Min %: 98.0 CAS Max %: 100.0 Purity Grade: ACS Grade
Tartaric Acid Aqueous Solution, 50% (w/v), Ricca Chemical
CAS: 7732-18-5 Physical Form: Solution Assay: 50% Assay Percent Range: 50% (w/v)
L-(+)-Tartaric Acid, ≥97%, MP Biomedicals™
Chemical Name or Material: L-(+)-Tartaric Acid CAS: 87-69-4 Physical Form: Powder Assay Percent Range: ≥97% Molecular Formula: C4H6O6 Formula Weight: 150.1 Synonym: L-2, 3-Dihydroxybutanedioic acid
L-(+)-Tartaric Acid, GR, ACS, EMD Millipore
Chemical Name or Material: L-(+)-Tartaric Acid Name Note: For ACS analysis CAS: 87-69-4 Physical Form: Solid Molecular Formula: C4H6O6 Formula Weight: 150.09 Synonym: L-2, 3-Dihydroxybutanedioic acid
  spinner