Tartronic Acid
Tartronic acid, 98%
Chemical Name or Material: Tartronic acid CAS: 80-69-3 Molecular Formula: C3H4O5 Formula Weight: 120.06g/mol MDL Number: MFCD00004237 Beilstein: 1209791 Merck Index: 14,9073
  spinner