Tetrahydrofuran
Tetrahydrofuran, 99.8%, Acros Organics
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 CAS: 128-37-0 Purity Grade: For biochemistry Assay Percent Range: 99.8% Molecular Formula: C4H8O Formula Weight: 72.11 Synonym: THF
Tetrahydrofuran, 99.9%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 CAS: 128-37-0 Purity Grade: Extra Dry Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C4 H8 O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran Name Note: Stabilized, water < 50ppm CAS: 109-99-9 CAS: 128-37-0 Purity Grade: Extra Dry Purity Grade Notes: Water <50ppm Assay: ≥99.80% (GC) Assay Percent Range: 99.85% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, 99.9%, Extra Dry, Stabilized, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran Name Note: Stabilized, water < 50ppm CAS: 109-99-9 CAS: 128-37-0 Purity Grade: Extra Dry Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C4 H8 O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, UV, >99.7% min, HPLC Grade, Alfa Aesar™
Chemical Name or Material: Tetrahydrofuran Name Note: Unstable CAS: 109-99-9 Purity Grade: HPLC, UV Physical Form: Liquid Assay Percent Range: >99.7% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356
Tetrahydrofuran, anhydrous, unstable, 99.8+%, Alfa Aesar™
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356
Tetrahydrofuran, 99.9%, Extra Dry, stabilized, anhydrous, SC, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran Name Note: Stabilized CAS: 109-99-9 CAS: 128-37-0 Purity Grade: Extra Dry Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran (Certified), Fisher Chemical
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 CAS: 128-37-0 Purity Grade: Certified Grade Purity Grade Notes: Contains about 0.025% butylated hydroxytoluene as a preservative Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356
Tetrahydrofuran, Spectrophotometric grade, >99.7%, ChemSeal, Alfa Aesar™
Chemical Name or Material: Tetrahydrofuran Name Note: Unstable CAS: 109-99-9 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >99.7% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356
Tetrahydrofuran, 99.6%, ACS reagent, stabilized with BHT, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran Name Note: p.a. CAS: 109-99-9 CAS: 128-37-0 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran Name Note: Anhydrous, Stabilized CAS: 109-99-9 CAS: 128-37-0 Purity Grade: Extra Pure Assay: ≥99.85% Assay Percent Range: 99.9% Molecular Formula: C4 H8 O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 CAS: 128-37-0 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, ACROS Organics™
Chemical Name or Material: Tetrahydrofuran Name Note: Water < 50ppm CAS: 109-99-9 CAS: 128-37-0 Purity Grade: Extra Dry Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C4 H8 O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, 99.9%, for HPLC, unstabilized, ACROS Organics™
Chemical Name or Material: Tetrahydrofuran Name Note: 99.9% CAS: 109-99-9 CAS: 128-37-0 Purity Grade: For HPLC Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C4 H8 O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran Name Note: Water <50 ppm, stabilized CAS: 109-99-9 CAS: 128-37-0 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99.5% (GC) Assay Percent Range: 99.5% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, Acros Organics™
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, 99.8%, for HPLC, unstabilized, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 Purity Grade: For HPLC Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: C4 H8 O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 Purity Grade: OPTIMA™ Purity Grade Notes: High purity mobile phase for HPLC Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: Diethylene Oxide
Tetrahydrofuran (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: C4H8O Linear Formula: CH2(CH2)2CH2O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran (Histological), Fisher Chemical
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 CAS: 128-37-0 Purity Grade: Histological Grade Molecular Formula: C4H8O Linear Formula: CH2(CH2)2CH2O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, non-UV, HPLC Grade, >99.7%, stable, Alfa Aesar™
Chemical Name or Material: Tetrahydrofuran Name Note: With 250ppm BHT CAS: 109-99-9 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: >99.7% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356
Tetrahydrofuran, Spectrophotometric Grade 99.5+%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 CAS: 128-37-0 Purity Grade: For spectroscopy Assay: ≥99.5% Assay Percent Range: 99.5+% Molecular Formula: C4H8O Formula Weight: 72.11 Synonym: THF
Tetrahydrofuran, Spectrophotometric Grade, >99.7%, Alfa Aesar™
Chemical Name or Material: Tetrahydrofuran Name Note: Unstable CAS: 109-99-9 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >99.7% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, package of 4x25ML bottl, ACROS Organics™
Chemical Name or Material: Tetrahydrofuran Name Note: Stabilized CAS: 109-99-9 CAS: 128-37-0 Purity Grade: Extra Dry Assay: ≥99.80% (GC) Assay Percent Range: 99.85% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, ACS, >99%, Alfa Aesar™
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356
Tetrahydrofuran (Spectranalyzed™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 Purity Grade: Spectranalyzed™ Assay: ≥99.9% Assay Percent Range: ≥99.9 % Molecular Formula: C4H8O Linear Formula: CH2(CH2)2CH2O Formula Weight: 72.11 MDL Number: MFCD00005356 Synonym: THF
Tetrahydrofuran, anhydrous, 99.8+%, unstab., packaged under Argon in resealable ChemSeal™ bottles
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 Name Note: Anhydrous, UnStabilized Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356 UN Number: UN2056 Beilstein: 102391 Merck Index: 14,9211
Tetrahydrofuran Products for Science Education
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 Assay Percent Range: >95% Linear Formula: C4H8O Formula Weight: 72.11 Synonym: THF
Tetrahydrofuran, anhydrous, >99.8%%, Alfa Aesar™
Chemical Name or Material: Tetrahydrofuran Name Note: Water <100ppm CAS: 109-99-9 Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00005356
Tetrahydrofuran ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Tetrahydrofuran CAS: 109-99-9 Purity Grade: ACS Reagent Grade Molecular Formula: C4H8O Formula Weight: 72.11
  spinner