Thallium -Tl
Thallium(I) nitrate, 99.999%, (trace metal basis), ACROS Organics™
Chemical Name or Material: Thallium(I) nitrate CAS: 10102-45-1 Purity Grade: For trace metal basis analysis Assay: 76.5 to 77% (Tl) Assay Percent Range: 100.00% Molecular Formula: NO3Tl Linear Formula: TlNO3 Formula Weight: 266.38
Thallic Acetate 98%, ACROS Organics™
Chemical Name or Material: Thallic acetate Name Note: 98% CAS: 2570-63-0 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C6H9O6Tl Linear Formula: (CH3CO2)3Tl Formula Weight: 381.51 MDL Number: MFCD00008698 Synonym: Acetic acid, thallium(III) salt
Thallium(I) sulfate, 99.5% min (metals basis), Alfa Aesar™
Chemical Name or Material: Thallium(I) sulfate CAS: 7446-18-6 Physical Form: Crystalline Assay Percent Range: 99.5% min (metals basis) Molecular Formula: Tl2SO4 Formula Weight: 504.8 MDL Number: MFCD00011278 Synonym: Thallous sulfate
Thallium(I) nitrate, 99.5%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Thallium(I) nitrate Assay Percent Range: 99.5% Molecular Formula: NO3Tl Linear Formula: TlNO3 Formula Weight: 266.38 MDL Number: MFCD00011280
Thallium (I) Acetate (Crystalline/Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Thallium (I) Acetate CAS: 563-68-8 Purity Grade: Laboratory Grade Physical Form: Powder/Solid Molecular Formula: C2H3O2Tl Formula Weight: 263.43 MDL Number: MFCD00013045 Synonym: Acetic Acid Thallium Salt
Thallic trifluoroacetate, ACROS Organics™
Chemical Name or Material: Thallic trifluoroacetate CAS: 23586-53-0 Purity Grade: Technical Grade Molecular Formula: C6F9O6Tl Linear Formula: (CF3COO)3Tl Formula Weight: 543.42 MDL Number: MFCD00000414 Synonym: TTFA
Thallium(I) iodide, ultra dry, 99.999% (metals basis), Alfa Aesar™
Name Note: ultra dry CAS: 7790-30-9 Physical Form: Beads Assay Percent Range: 99.999% metal basis Molecular Formula: TlI Formula Weight: 331.27 MDL Number: MFCD00011279 Synonym: Thallous iodide
Thallium(I) sulfate, 99.9+%, (trace metal basis), ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Thallium(I) sulfate Name Note: 99.9+% CAS: 7446-18-6 Purity Grade: Trace Metal Basis Assay: 80.0 to 84.0% (Tl) Assay Percent Range: 99.9+% Molecular Formula: O4STl2 Linear Formula: TI2SO4 Formula Weight: 504.79 MDL Number: MFCD00011278
Thallium(I) chloride, 99%, ACROS Organics™
Chemical Name or Material: Thallium(I) chloride Name Note: 99% CAS: 7791-12-0 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: ClTl Linear Formula: TICl Formula Weight: 239.82 MDL Number: MFCD00011274 Synonym: Thallous chloride
Thallium Rod, 12.7mm (0.5in) dia., 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Thallium CAS: 7440-28-0 Physical Form: Rod, 12.7mm (0.5 in.) dia. Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00134063
Thallium(I) fluoride, 99%, ACROS Organics™
Chemical Name or Material: Thallium(I) fluoride Name Note: 99% CAS: 7789-27-7 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: FTl Linear Formula: FTl Formula Weight: 223.37 MDL Number: MFCD00049607
Thallium Iodide (Ous), 99.6%, MP Biomedicals
Chemical Name or Material: Thallium Iodide CAS: 7790-30-9 Formula Weight: 331.31
Thallium(III) nitrate trihydrate, 99.5%, ACROS Organics™
Chemical Name or Material: Thallium(III) nitrate trihydrate Name Note: 99.5% CAS: 13453-38-8 Assay Percent Range: 99.50% Molecular Formula: N3O9Tl·3H2O Linear Formula: Tl(NO3)3·3H2O Formula Weight: 444.42 MDL Number: MFCD00149874 Synonym: TTN
  spinner