Thiosalicylic Acid
Thiosalicylic acid, 98%, ACROS Organics™
Chemical Name or Material: Thiosalicylic acid Name Note: 98% CAS: 147-93-3 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C7H6O2S Linear Formula: HSC6H4CO2H Formula Weight: 154.18 MDL Number: MFCD00004836 Synonym: 2-Mercaptobenzoic acid
  spinner