Threonine -Thr and Derivatives
L-Threonine, 98%, ACROS Organics™
Chemical Name or Material: L-Threonine Assay: 97.5 to 102.5% (on dried substance) (Total base) Assay Percent Range: 98% Molecular Formula: C4H9NO3 Linear Formula: CH3CH(OH)CH(NH2)CO2H Formula Weight: 119.12 MDL Number: MFCD00064270 Synonym: L-2-Amino-3-hydroxybutyric acid
L(+)-allo-Threonine, 99%, ACROS Organics™
Chemical Name or Material: L(+)-allo-Threonine Name Note: 99% CAS: 28954-12-3 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C4H9NO3 Linear Formula: CH3CH(OH)CH(NH2)CO2H Formula Weight: 119.12 MDL Number: MFCD00064268 Synonym: (2S
D-Threonine, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: D-Threonine Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C4H9NO3 Linear Formula: CH3CH(OH)CH(NH)COH Formula Weight: 119.12 MDL Number: MFCD00064269 Synonym: (2R, 3S)-2-Amino-3-hydroxybutyric acid
DL-Threonine, 99.5%, ACROS Organics™
Chemical Name or Material: DL-Threonine Assay: 98.0 to 101.0% (on dried substance) Assay Percent Range: 99.5% Molecular Formula: C4H9NO3 Linear Formula: CH3CH(OH)CH(NH2)CO2H Formula Weight: 119.12 MDL Number: MFCD00063722
L-threonine methyleester hydrochloride, +98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-Threonine methyleester hydrochloride Assay: ≥98.0% ((on dry substance)) Assay Percent Range: 98+% Molecular Formula: C5H11NO3·HCl Linear Formula: CH3CH(OH)CH(NH2)C(O)OCH3·HCl Formula Weight: 169.61 MDL Number: MFCD00037677
L-Threonine (White Crystals or Crystalline Powder), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-Threonine CAS: 72-19-5 Physical Form: Powder/Solid Assay: 99.0 to 101.0% Assay Percent Range: 99.0 to 101.0 % Molecular Formula: C4H9NO3 Formula Weight: 119.12
DL-Threonine, >99%, MP Biomedicals™
Chemical Name or Material: DL-Threonine CAS: 80-68-2 Physical Form: Powder Molecular Formula: C4H9NO3 Formula Weight: 119.1 Synonym: DL-2-Amino-3-hydroxybutyric acid
L-Threonine Allo-free base, 990%, MP Biomedicals™
Chemical Name or Material: L-Threonine Name Note: Allo Free CAS: 72-19-5 Physical Form: Crystalline Powder Assay Percent Range: 99% Molecular Formula: C4H9NO3 Formula Weight: 119.1 MDL Number: MFCD00064270 Synonym: (2S, 3R)-2-Amino-3-hydroxybutyric acid
  spinner