Trichloroacetic Acid
Trichloroacetic acid, 99+%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trichloroacetic acid Name Note: 99+% CAS: 76-03-9 Purity Grade: ACS Reagent Grade Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C2HCl3O2 Linear Formula: CCl3CO2H Formula Weight: 163.39 MDL Number: MFCD00004177 Synonym: TCA
Trichloroacetic acid, sodium salt, 97%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trichloroacetic acid, sodium salt Assay: ≥96.0% Assay Percent Range: 97% Molecular Formula: C2Cl3NaO2 Linear Formula: CCl3CO2Na Formula Weight: 185.37 MDL Number: MFCD00064198 Synonym: Sodium trichloroacetate
Trichloroacetic Acid Solution (20% w/v), Fisher Chemical
Chemical Name or Material: Trichloroacetic Acid Solution Name Note: 20% (w/v) CAS: 76-03-9 CAS: 7732-18-5 Assay: 20 to 21% Assay Percent Range: 20 to 21 % Molecular Formula: Cl3CCOOH MDL Number: MFCD00004177
Trichloroacetic Acid Products for Science Education
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Purity Grade: Laboratory Grade Assay Percent Range: >95% Linear Formula: CCl3COOH Formula Weight: 163.39 Synonym: Tca
Trichloroacetic Acid (Crystalline/Certified), Fisher Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Purity Grade: Certified Grade Physical Form: Solid Assay: ≥99.0% Assay Percent Range: ≥99.0 % Molecular Formula: C2HCl3O2 Linear Formula: CCl3COOH Formula Weight: 163.39 MDL Number: MFCD00004177 Synonym: TCA
Trichloroacetic Acid (Crystalline/Certified ACS), Fisher Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Purity Grade: Certified ACS Grade Physical Form: Solid Assay: ≥99.0% Assay Percent Range: ≥99.0 % Molecular Formula: C2HCl3O2 Linear Formula: CCl3COOH Formula Weight: 163.39 MDL Number: MFCD00004177 Synonym: TCA
Trichloroacetic Acid, Fisher Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 CAS: 7732-18-5 Assay: 99.5 to 101.0% Assay Percent Range: 99.5 to 101.0 % Molecular Formula: Cl3CCOOH Formula Weight: 163.39 MDL Number: MFCD00004177
Trichloroacetic Acid, 15% w/v (AOAC), Fisher Chemical
Chemical Name or Material: Trichloroacetic Acid Name Note: 15% (w/v) CAS: 7732-18-5 CAS Min %: 85.0 CAS Max %: 89.0 CAS: 76-03-9 CAS Min %: 12.77 CAS Max %: 13.29 CAS: 7440-44-0 CAS Min %: 0.17 CAS Max %: 0.18
Trichloroacetic Acid, 10% w/v (AOAC), Fisher Chemical
Chemical Name or Material: Trichloroacetic Acid Name Note: 10% (w/v) CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 93.0 CAS: 76-03-9 CAS Min %: 8.9 CAS Max %: 9.26 CAS: 7440-44-0 CAS Min %: 0.18 CAS Max %: 0.19
Trichloroacetic Acid (Flakes or Crystals), Fisher BioReagents
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Physical Form: Solid Assay: ≥99.0% Assay Percent Range: ≥99.0 % Molecular Formula: C2HCl3O2 Linear Formula: CCl3COOH Formula Weight: 163.39 Synonym: TCA
Trichloroacetic Acid, 33% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Min %: 74.0 CAS Max %: 77.0 CAS: 76-03-9 CAS Min %: 24.29 CAS Max %: 25.28 CAS: 7440-44-0 CAS Min %: 0.15 CAS Max %: 0.15 Physical Form: Liquid
Trichloroacetic Acid, 80% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Min %: 54.0 CAS Max %: 57.0 CAS: 76-03-9 CAS Min %: 43.52 CAS Max %: 45.3 CAS: 7440-44-0 CAS Min %: 0.11 CAS Max %: 0.11 Physical Form: Liquid
Trichloroacetic Acid, 3% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid
Trichloroacetic acid, 15% (w/v) aqueous solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic acid CAS: 7732-18-5 CAS: 76-03-9
Trichloroacetic Acid, 10% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 93.0 CAS: 76-03-9 CAS Min %: 8.9 CAS Max %: 9.26 CAS: 7440-44-0 CAS Min %: 0.18 CAS Max %: 0.19
Trichloroacetic Acid ACS AR Crystal, Macron Fine Chemicals™
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Purity Grade: ACS Grade Physical Form: Crystalline Molecular Formula: Cl3CCOOH Formula Weight: 163.39
Trichloroacetic Acid, 5% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid
Trichloroacetic Acid, 30% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Min %: 75.0 CAS Max %: 78.0 CAS: 76-03-9 CAS Min %: 22.59 CAS Max %: 23.51 CAS: 7440-44-0 CAS Min %: 0.15 CAS Max %: 0.16 Physical Form: Liquid
Trichloroacetic Acid, 24% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Min %: 79.0 CAS Max %: 82.0 CAS: 76-03-9 CAS Min %: 18.95 CAS Max %: 19.72 CAS: 7440-44-0 CAS Min %: 0.16 CAS Max %: 0.16 Physical Form: Liquid
Trichloroacetic acid, 20% (w/v) aqueous solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic acid CAS: 7732-18-5 CAS: 76-03-9
Trichloroacetic acid, ACS, ≥99%, MP Biomedicals™
Chemical Name or Material: Trichloroacetic acid CAS: 76-03-9 Physical Form: Crystalline Powder Assay Percent Range: ≥99% Molecular Formula: C2HCl3O2 Formula Weight: 163.4 Synonym: TCA
Trichloroacetic acid, 99.6%, MP Biomedicals™
Chemical Name or Material: Trichloroacetic acid CAS: 76-03-9 Physical Form: Crystalline Powder Assay Percent Range: 0.996 Molecular Formula: C2HCl3O2 Formula Weight: 163.4 Synonym: TCA
Trichloroacetic Acid Solution (6.1N (100% w/v))
Chemical Name or Material: Trichloroacetic Acid Solution Name Note: 6.1N CAS: 76-03-9 Formula Weight: 163.38
Trichloroacetic Acid, 99.5% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trichloroacetic Acid Name Note: For ACS analysis CAS: 76-03-9 Physical Form: Deliquescent Crystals Assay Percent Range: 99.5% min. Molecular Formula: CCl3COOH Formula Weight: 163.39
Trichloroacetic Acid ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Purity Grade: ACS Grade Linear Formula: CCl3COOH Formula Weight: 163.39
  spinner