Tyrosine -Tyr and Derivatives
L-Tyrosine disodium salt, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-Tyrosine disodium salt Assay Percent Range: 98% Molecular Formula: C9H9NNa2O3 Linear Formula: NaOC6H4CH2CH(NH2)CO2Na Formula Weight: 225.15 MDL Number: MFCD00037203
DL-m-Tyrosine, 98.50%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: DL-m-Tyrosine Assay: ≥98.0% Assay Percent Range: 98.50% Molecular Formula: C9H11NO3 Linear Formula: HOC6H4CH2CH(NH2)CO2H Formula Weight: 181.19 MDL Number: MFCD00002597 Synonym: 3-(3-Hydroxyphenyl)-DL-alanine
O-Methyl-L-tyrosine, 98%, ACROS Organics™
Chemical Name or Material: O-Methyl-L-tyrosine CAS: 1581806 CAS Min %: 97.5 CAS Max %: 100.0 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C10H13NO3 Linear Formula: CH3OC6H4CH2CH(NH2)CO2H Formula Weight: 195.22 MDL Number: MFCD00002604 Synonym: p-Methoxy-L-phenylalanine
BOC-L-Tyrosine, 99+%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: BOC-L-Tyrosine Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C14H19NO5 Linear Formula: HOC6H4CH2CH(COOH)nHCO2C(CH3)3 Formula Weight: 281.3 MDL Number: MFCD00037179 Synonym: N-(tert-Butoxycarbonyl)-L-tyrosine
D-Tyrosine, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: D-Tyrosine Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C9H11NO3 Linear Formula: 4-(HO)C6H4CH2CH(NH2)CO2H Formula Weight: 181.19 MDL Number: MFCD00063073
3-Iodo-L-tyrosine, 97%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 3-Iodo-L-tyrosine CAS: 70-78-0 CAS Min %: 96.0 CAS Max %: 100.0 Assay: ≥96.0% Assay Percent Range: 97% Molecular Formula: C9H10INO3 Formula Weight: 307.08 MDL Number: MFCD00002608
GW 1929 hydrochloride, Tocris
Chemical Name or Material: GW 1929 hydrochloride Assay Percent Range: >98% Molecular Formula: C30H29N3O4·HCl Formula Weight: 532.03 Synonym: N-(2-Benzoylphenyl)-O-[2-(methyl-2-pyridinylamino)ethyl]-L-tyrosine hydrochloride
L-Tyrosine [Free Base], MP Biomedicals
Chemical Name or Material: L-Tyrosine [Free Base] CAS: 60-18-4 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystalline Molecular Formula: C9H11NO3 Formula Weight: 181.2 Synonym: β-p-Hydroxyphenylalanine
3-Iodo-L-Tyrosine, MP Biomedicals™
Chemical Name or Material: 3-Iodo-L-Tyrosine CAS: 70-78-0 Physical Form: Solid Assay Percent Range: 99% Molecular Formula: C9H10INO3 Formula Weight: 307.1 Synonym: S(-)-3-Iodo-4-Hydroxy-phenylamine
Tyr-Gly, MP Biomedicals™
Chemical Name or Material: Tyr-Gly CAS: 673-08-5 Physical Form: Powder Molecular Formula: C11H14N2O4 Formula Weight: 238.2 Synonym: L-Tyrosylglycine
L-Tyrosine Disodium Salt, MP Biomedicals
Chemical Name or Material: L-Tyrosine Disodium Salt CAS: 122666-87-9 Molecular Formula: C9H9NO3Na2 Formula Weight: 225.2 Synonym: Crystalline
L-Tyrosine, MP Biomedicals™
Chemical Name or Material: L-Tyrosine CAS: 60-18-4 Physical Form: Powder Assay Percent Range: 0.97 Molecular Formula: C9H11NO3 Formula Weight: 181.19 MDL Number: MFCD00002606 Synonym: β-p-Hydroxyphenylalanine
DL-Tyrosine, 99%, MP Biomedicals™
Chemical Name or Material: DL-Tyrosine CAS: 556-03-6 Physical Form: Crystalline Assay Percent Range: 99% Molecular Formula: C9H11NO3 Formula Weight: 181.2 Synonym: β-(p-Hydroxyphenyl)alanine
N-Acetyl-L-Tyrosine, MP Biomedicals™
Chemical Name or Material: N-Acetyl-L-Tyrosine CAS: 537-55-3 Physical Form: Crystalline Molecular Formula: C11H13NO4 Formula Weight: 223.2 Synonym: α-(Acetamido)-β-(4-Hydroxyphenyl)-propionic Acid
DL-Tyrosine, 99%, ACROS Organics™
Chemical Name or Material: DL-Tyrosine Name Note: 99% CAS: 556-03-6 Assay: 98.5 to 101.5% Assay Percent Range: 99% Molecular Formula: C9H11NO3 Linear Formula: 4-(HO)C6H4CH2CH(NH2)CO2H Formula Weight: 181.19 MDL Number: MFCD00063074 Synonym: 3-(4-Hydroxyphenyl)-DL-alanine
BOC-D-Tyrosine, 99%, ACROS Organics™
Chemical Name or Material: BOC-D-Tyrosine Name Note: 99% CAS: 70642-86-3 Assay Percent Range: 99% Molecular Formula: C14H19NO5 Linear Formula: HOC6H4CH2CH(COOH)nHCO2C(CH3)3 Formula Weight: 281.3 MDL Number: MFCD00063030 Synonym: N-(tert-Butoxycarbonyl)-D-tyrosine
L-Tyrosine (White Crystals or Crystalline Powder), Fisher BioReagents
Chemical Name or Material: L-Tyrosine CAS: 60-18-4 Physical Form: Solid Assay: 98.5 to 100.5% Assay Percent Range: 98.5 to 100.5 % Molecular Formula: C9H11NO3 Formula Weight: 181.19
L-Tyrosine Disodium Salt, (Crystalline),MP Biomedicals
Chemical Name or Material: L-Tyrosine Disodium Salt CAS: 122666-87-9 Physical Form: Crystalline Molecular Formula: C9H9NO3Na2 Formula Weight: 225.2
D-Tyrosine, >98.5%, MP Biomedicals™
Chemical Name or Material: D-Tyrosine CAS: 556-02-5 Physical Form: Crystalline Assay Percent Range: >98.5% Molecular Formula: C9H11NO3 Formula Weight: 181.2 Synonym: 3-(4-Hydroxyphenyl)-D-alanine
  spinner