Valeric Acid
Valerenic acid, Tocris
Chemical Name or Material: Valerenic acid CAS: 3569-10-6 Assay Percent Range: >99% Molecular Formula: C15H22O2 Formula Weight: 234.33g/mol Synonym: (2E)-3-[(4S,7R,7aR)-2,4,5,6,7,7a-Hexadydro-3,7-dimethyl-1H-inden-4-yl]-2-methyl-2-propenoic acid
Valeric Acid, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Valeric acid Assay: 98.5% min. Assay Percent Range: 99% Molecular Formula: C5H10O2 Linear Formula: CH3(CH2)3CO2H Formula Weight: 102.13g/mol MDL Number: MFCD00004413 Synonym: 1-Butanecarboxylic acid
  spinner