Valine -Val and Derivatives
L-Valine, 99%
Chemical Name or Material: L-Valine CAS: 72-18-4 Molecular Formula: C5H11NO2 Formula Weight: 117.15 MDL Number: MFCD00064220 Beilstein: 1721136 Merck Index: 14,9909
L(+)-Norvaline, 99%, ACROS Organics™
Chemical Name or Material: L(+)-Norvaline CAS: 6600-40-4 Assay: 98.5 to 101.5% Assay Percent Range: 99% Molecular Formula: C5H11NO2 Formula Weight: 117.15 MDL Number: MFCD00064421 Synonym: L(+)-2-Aminovaleric acid
D-Valine, +98%, ACROS Organics™
Chemical Name or Material: D-Valine Name Note: 98+% CAS: 640-68-6 Assay: 98.0 to 102.0% Assay Percent Range: 98+% Molecular Formula: C5H11NO2 Linear Formula: (CH3)2CHCH(NH2)CO2H Formula Weight: 117.15 MDL Number: MFCD00064219 Synonym: D-2-Amino-3-methylbutyric acid
DL-Valine, Acros Organics
Chemical Name or Material: DL-Valine CAS: 516-06-3 Purity Grade: Extra Pure Molecular Formula: C5H11NO2 Formula Weight: 117.15 Synonym: DL-2-Amino-3-methylbutyric acid
DL-Valine, +99%, ACROS Organics™
Chemical Name or Material: DL-Valine Name Note: 99+% CAS: 516-06-3 Purity Grade: For biochemistry Assay: 98.0 to 101.0% Assay Percent Range: 99+% Molecular Formula: C5H11NO2 Linear Formula: (CH3)2CHCH(NH2)CO2H Formula Weight: 117.15 MDL Number: MFCD00004267 Synonym: DL-2-Amino-3-methylbutyric acid
BOC-L-Valine, 99+%, ACROS Organics™
Chemical Name or Material: BOC-L-Valine Name Note: 99+% CAS: 13734-41-3 Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C10H19NO4 Linear Formula: (CH3)2CHCHCOOHNHCOOC(CH3)3 Formula Weight: 217.27 MDL Number: MFCD00065605 Synonym: N-(tert-Butoxycarbonyl)-L-valine
L-Valine 99%, ACROS Organics™
Chemical Name or Material: L-Valine Name Note: 99% CAS: 72-18-4 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C5H11NO2 Linear Formula: (CH3)2CHCH(NH2)CO2H Formula Weight: 117.15 MDL Number: MFCD00064220 Synonym: L-2-Amino-3-methylbutyric acid
D-Valine methyleester hydrochloride, 99%, ACROS Organics™
Chemical Name or Material: D-Valine methyleester hydrochloride Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C6H13NO2·HCl Linear Formula: (CH3)2CHCH(NH2)COOCH3·HCl Formula Weight: 167.63 MDL Number: MFCD00237309
PD 166793, Tocris
Chemical Name or Material: PD 166793 Assay Percent Range: >99% Molecular Formula: C17H18BrNO4S Formula Weight: 412.3 Synonym: N-[(4'-Bromo[1, 1'-biphenyl]-4-yl)sulfonyl]-L-valine
L-Valine (White Crystals or Crystalline Powder), Fisher BioReagents
Chemical Name or Material: L-Valine CAS: 72-18-4 Physical Form: Powder/Solid Assay: 98.5 to 100.5% Assay Percent Range: 98.5 to 100.5 % Molecular Formula: C5H11NO2 Formula Weight: 117.08
D-Valine, 99%, MP Biomedicals™
Chemical Name or Material: D-Valine Assay Percent Range: 99% Molecular Formula: C5H11NO2 Formula Weight: 117.1 Synonym: D-2-Amino-3-methylbutanoic acid
L-Valine, 99%, MP Biomedicals
Chemical Name or Material: L-Valine, 99% CAS: 72-18-4 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystalline Molecular Formula: C5H11NO2 Formula Weight: 117.1 Synonym: L-2-Amino-3-methylbutanoic acid
  spinner