Vanadium -V
Vanadium(III) bromide, 99.5% (metals basis), Alfa Aesar™
Chemical Name or Material: Vanadium(III) bromide CAS: 13470-26-3 Physical Form: Crystalline Assay Percent Range: 99.5% (metals basis) Molecular Formula: VBr3 Formula Weight: 290.67 MDL Number: MFCD00049695 Synonym: Vanadium tribromide
Vanadyl(IV) sulfate hydrate, 17-23% V, ACROS Organics™
Chemical Name or Material: Vanadyl(IV) sulfate hydrate Name Note: 17-23% V CAS: 12439-96-2 Assay: 17 to 23% (V) Molecular Formula: O5SV·xH2O Linear Formula: VOSO4·xH2O Formula Weight: 163 MDL Number: MFCD00149933 Synonym: Oxysulfato vanadium (IV) hydrate
Vanadium(V) oxide, +99.6%, ACROS Organics™
Chemical Name or Material: Vanadium(V) oxide Name Note: 99.6+% CAS: 1314-62-1 Assay Percent Range: 99.6+% Molecular Formula: O5V2 Linear Formula: V2O5 Formula Weight: 181.88 MDL Number: MFCD00011457 Synonym: Vanadium pentoxide
Vanadium(IV) oxide, 99% (metals basis), Alfa Aesar™
Chemical Name or Material: Vanadium(IV) oxide CAS: 12036-21-4 Physical Form: -100 Mesh Powder Assay Percent Range: 99% (metals basis) Molecular Formula: O2V Formula Weight: 82.94 MDL Number: MFCD00049702
Vanadium(IV) chloride, 99+%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Vanadium(IV) chloride Name Note: 99+% CAS: 7632-51-1 Assay: ≥99.0% (VCl4) Assay Percent Range: 99+% Molecular Formula: Cl4V Linear Formula: VCl4 Formula Weight: 192.75 MDL Number: MFCD00061343
Vanadium(III) oxide, 95%, Alfa Aesar™
CAS: 1314-34-7 Physical Form: Powder Assay Percent Range: 95% Molecular Formula: O3V2 Formula Weight: 149.88 MDL Number: MFCD00011455 Synonym: Vanadium trioxide
Vanadium(III) chloride-tetrahydrofuran complex (1:3), 0.5M solution in methylene chloride, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Vanadium(III) chloride-tetrahydrofuran complex (1:3) CAS: 19559-06-9 CAS: 75-09-2 Molecular Formula: VCl3·C12H24O3 Linear Formula: VCl·3THF Formula Weight: 373.62
Vanadium(III) chloride, 97%, ACROS Organics™
Chemical Name or Material: Vanadium(III) chloride Name Note: 99% CAS: 7718-98-1 Assay: ≥96.0% Assay Percent Range: 97% Molecular Formula: Cl3V Linear Formula: VCl3 Formula Weight: 157.3 MDL Number: MFCD00011454 Synonym: Vanadium trichloride
Vanadium(V) oxide, 99+%, Acros Organics
Chemical Name or Material: Vanadium(V) oxide Assay Percent Range: 99+% Molecular Formula: O5V2 Formula Weight: 181.88 Synonym: Vanadium pentoxide
Vanadium powder, 99.5% (metals basis), Alfa Aesar™
Chemical Name or Material: Vanadium CAS: 7440-62-2 Physical Form: Powder Assay Percent Range: 99.5% (Metals basis) MDL Number: MFCD00011453
Vanadium foil, 99.5% (metal basis), Alfa Aesar™
Chemical Name or Material: Vanadium CAS: 7440-62-2 Physical Form: Foil, 0.25mm (0.01 in.) thick Assay Percent Range: 99.5% (Metals basis) MDL Number: MFCD00011453
Vanadium Granules, 1-3mm (0.04-0.12 in.), 99.7% (Metals basis), Alfa Aesar™
Chemical Name or Material: Vanadium CAS: 7440-62-2 Purity Grade: Vacuum Deposition Grade Physical Form: Granular Assay Percent Range: 99.7% (Metals basis) MDL Number: MFCD00011453
Vanadium(V) oxide, 99.6% min, Alfa Aesar™
Chemical Name or Material: Vanadium(V) oxide CAS: 1314-62-1 Physical Form: -10 Mesh Assay Percent Range: 99.6% min Molecular Formula: V2O5 Formula Weight: 181.88 MDL Number: MFCD00011457
Vanadium(V) oxide, +98%, ACROS Organics™
Chemical Name or Material: Vanadium(V) oxide Name Note: 98+% CAS: 1314-62-1 Assay: ≥98.0% (V2O5) Assay Percent Range: 98+% Molecular Formula: O5V2 Linear Formula: V2O5 Formula Weight: 181.88 MDL Number: MFCD00011457 Synonym: Vanadium pentoxide
Vanadium(IV) Sulfate Oxide Hydrate, 99.9% (Metal Basis), Alfa Aesar™
Chemical Name or Material: Vanadium(IV) Sulfate Oxide Hydrate CAS: 123334-20-3 Physical Form: Lumps Assay Percent Range: 99.9%, metal basis Molecular Formula: VOSO4·xH2O Formula Weight: 163 MDL Number: MFCD00149933 Synonym: Vanadium oxysulfate hydrate
Vanadium(III) chloride, 99% (metals basis), Alfa Aesar™
Chemical Name or Material: Vanadium(III) chloride CAS: 7718-98-1 Physical Form: -4 Mesh Assay Percent Range: 99% (metals basis) Molecular Formula: VCl3 Formula Weight: 157.3 MDL Number: MFCD00011454
Vanadium(III) oxide, 99.7% (metals basis), Alfa Aesar™
Chemical Name or Material: Vanadium(III) oxide CAS: 1314-34-7 Physical Form: Powder Assay Percent Range: 99.7% (metals basis) Molecular Formula: V2O3 Formula Weight: 149.88 MDL Number: MFCD00011455 Synonym: Vanadium trioxide
Vanadium(V) oxide, 99.99% (metals basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Vanadium(V) oxide CAS: 1314-62-1 Physical Form: -22 Mesh Powder Assay Percent Range: 99.99% (metals basis) Molecular Formula: V2O5 Formula Weight: 181.88 MDL Number: MFCD00011457
Vanadium foil, 99.8% (metals basis), Alfa Aesar™
Chemical Name or Material: Vanadium CAS: 7440-62-2 Physical Form: Foil, 0.127mm (0.005 in.) thick Assay Percent Range: 99.8% (Metals basis) MDL Number: MFCD00011453
Vanadium carbide, 99% (metals basis), Alfa Aesar™
CAS: 12070-10-9 Physical Form: Powder Assay Percent Range: 99% metal basis Molecular Formula: VC (VC0?88 by X-Ray) Formula Weight: 62.95 MDL Number: MFCD00049698
Vanadium Pieces, 99.7% (Metals basis), Alfa Aesar™
Chemical Name or Material: Vanadium CAS: 7440-62-2 Physical Form: Pieces Assay Percent Range: 99.7% (Metals basis) MDL Number: MFCD00011453
Vanado Molybdate Reagent, AOAC, Ricca Chemical
Chemical Name or Material: Vanado - Molybdate Reagent CAS: 7732-18-5 CAS Min %: 70.0 CAS Max %: 73.0 CAS: 7601-90-3 CAS Min %: 26.29 CAS Max %: 27.37 CAS: 12054-85-2 CAS Min %: 1.41 CAS Max %: 1.46 CAS: 7803-55-6 CAS Min %: 0.07 CAS Max %: 0.07 Purity Grade: AOAC Physical Form: Liquid
Vanadium Sulfate (Vanadyl), MP Biomedicals
Chemical Name or Material: Vanadium Sulfate (Vanadyl) CAS: 27774-13-6 Formula Weight: 163
Vanadium(V) Oxide, Extra Pure, EMD Millipore™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Vanadium(V) Oxide CAS: 1314-62-1 Purity Grade: Extra Pure Molecular Formula: V2O5 Formula Weight: 181.88 UN Number: UN 2862 Synonym: Vanadic anhydride
Vanadium Turnings, 2.5cm (1.0 in.) & down, 99.7% (Metals basis), Alfa Aesar™
Chemical Name or Material: Vanadium CAS: 7440-62-2 Physical Form: Turnings, 2.5cm (1.0 in.) and down Assay Percent Range: 99.7% (Metals basis) MDL Number: MFCD00011453
Vanadyl Sulfate (Crystalline/Laboratory), Fisher Chemical
Chemical Name or Material: Vanadyl Sulfate CAS: 27774-13-6 Purity Grade: Laboratory Grade Physical Form: Solid Molecular Formula: VOSO4 Linear Formula: VOSO4·nH2O MDL Number: MFCD00011460
  spinner