Xylene -m-
m-Xylene, 99+%, extra pure, ACROS Organics™
Chemical Name or Material: m-Xylene CAS: 108-38-3 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00008536 Synonym: 1, 3-Dimethylbenzene
m-Xylene, 99.0%, Extra Dry, anhydrous, AcroSeal™, ACROS Organics™
Chemical Name or Material: m-Xylene CAS: 108-38-3 Purity Grade: Extra Dry Assay: ≥99.0% Assay Percent Range: 99.0% Molecular Formula: C8;H10 Linear Formula: C6H4(CH3;)2 Formula Weight: 106.17 MDL Number: MFCD00008536 Synonym: 1, 3-Dimethylbenzene
m-Xylene, 99%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: m-Xylene CAS: 108-38-3 Purity Grade: Extra Dry Assay Percent Range: 99% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00008536 Synonym: 1, 3-Dimethylbenzene
m-Xylene (Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: m-Xylene CAS: 108-38-3 Purity Grade: Laboratory Grade Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.16 MDL Number: MFCD00008536
m-Xylene (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: m-Xylene CAS: 108-38-3 Purity Grade: Certified Grade Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.16 MDL Number: MFCD00008536
  spinner