Xylene -o-
o-Xylene, 96% min., HPLC Grade, Alfa Aesar™
Chemical Name or Material: o-Xylene CAS: 95-47-6 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: 96% min. Molecular Formula: (CH3)2C6H4 Formula Weight: 106.17 MDL Number: MFCD00008519
o-Xylene, 99%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: o-Xylene CAS: 95-47-6 Purity Grade: Extra Dry Assay: ≥98.5% (GC) Assay Percent Range: 99% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 Synonym: 1, 2-Dimethylbenzene
o-Xylene, 99%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: o-Xylene Name Note: Pure CAS: 95-47-6 Purity Grade: Pure Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00008519 Synonym: 1, 2-Dimethylbenzene
o-Xylene (Certified), Fisher Chemical
Chemical Name or Material: o-Xylene CAS: 95-47-6 CAS: 100-41-4 Purity Grade: Certified Grade Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.16 MDL Number: MFCD00008519
  spinner