Xylene -p-
p-Xylene, 99%, for HPLC, ACROS Organics™
Chemical Name or Material: p-Xylene Purity Grade: For HPLC Assay Percent Range: 99% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 Synonym: 1, 4-Dimethylbenzene
p-Xylene, 99%, pure, ACROS Organics™
Chemical Name or Material: p-Xylene CAS: 106-42-3 Purity Grade: Pure Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00008556 Synonym: 1, 4-Dimethylbenzene
p-Xylene, 99+%, extra pure, ACROS Organics™
Chemical Name or Material: p-Xylene Name Note: Extra Pure CAS: 106-42-3 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00008556 Synonym: 1, 4-Dimethylbenzene
p-Xylene (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: p-Xylene CAS: 106-42-3 Purity Grade: Certified Grade Assay: ≥99% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00008556
p-Xylene, Reagent, 98% min., EMD Millipore
Chemical Name or Material: p-Xylene Name Note: For Gas Chromatography CAS: 106-42-3 Physical Form: Liquid Assay Percent Range: 98% min. Molecular Formula: C8H10 Linear Formula: C6H4-1, 4-(CH3)2 Formula Weight: 16.17 Synonym: 1, 4-Dimethylbenzene
  spinner