Xylenes
Xylenes, Reagent ACS, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Xylenes CAS: 1330-20-7 CAS: 100-41-4 Purity Grade: ACS Reagent Grade Assay: ≥98.5% (xylene isomers plus ethylbenzene (ethylbenzene not to exceed 25%) Separat. techn. GC) Assay Percent Range: 99% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00077264 Synonym: Dimethylbenzene
Xylenes, extra pure, mixture of isomers, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Xylenes CAS: 1330-20-7 CAS: 100-41-4 Purity Grade: Extra Pure Assay: ≥98.0% (mixed isomers) (GC) Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 Synonym: Dimethylbenzene
Xylenes, ACS, 98.5%, Alfa Aesar
Chemical Name or Material: Xylenes CAS: 1330-20-7 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: >98.5% Molecular Formula: (CH3)2C6H4 Formula Weight: 106.17 MDL Number: MFCD00077264
Xylenes Products for Science Education
Chemical Name or Material: Xylenes CAS: 1330-20-7 CAS: 1-41-4 Assay Percent Range: 0.96 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 Synonym: Dimethylbenzene
Xylenes (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Xylenes CAS: 1330-20-7 CAS: 100-41-4 Purity Grade: Certified ACS Reagent Grade Assay: ≥98.5% Assay Percent Range: Assay: ≥98.5% Molecular Formula: C8H10 Formula Weight: 106.17 MDL Number: MFCD00077264 Synonym: Xylol, Dimethylbenzene
Xylenes, Semiconductor Grade, Alfa Aesar™
Chemical Name or Material: Xylenes CAS: 1330-20-7 Purity Grade: Semiconductor Grade Physical Form: Liquid Molecular Formula: (CH3)2C6H4 Formula Weight: 106.17 MDL Number: MFCD00077264
Xylenes, 99+%, for residue analysis, mixed, electronic use grade, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Xylenes Name Note: Residue Free CAS: 1330-20-7 CAS: 100-41-4 Purity Grade: For residue analysis Assay: ≥99% (total isomers) Assay Percent Range: 99+% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00077264 Synonym: Dimethylbenzene
Xylenes, 98+%, for analysis, mixture of isomers , ACROS Organics™
Chemical Name or Material: Xylenes CAS: 1330-20-7 CAS: 100-41-4 Purity Grade: For analysis Assay Percent Range: 98+% Molecular Formula: C8H10 Formula Weight: 106.17 Synonym: Dimethylbenzene
Xylenes (Histological), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Xylenes CAS: 1330-20-7 CAS: 100-41-4 Purity Grade: Histological Grade Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00077264 Synonym: Dimethylbenzene
Xylenes, 98%, pure, mixture of isomers, ACROS Organics™
Chemical Name or Material: Xylenes Name Note: Pure CAS: 1330-20-7 CAS: 100-41-4 Purity Grade: Pure Assay: ≥97% (isomers and ethylbenzene) Assay Percent Range: 98% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00077264 Synonym: Dimethylbenzene
Xylenes, +98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Xylenes Name Note: Mixture of isomers, Extra Dry CAS: 1330-20-7 CAS: 100-41-4 Purity Grade: Extra Dry Assay: ≥98.0% Assay Percent Range: 98+% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00077264
Xylenes, 96%, pure, mixed isomers with ethylbenzene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Xylenes Name Note: Mixed Isomers with Ethylbenzene CAS: 1330-20-7 CAS: 100-41-4 Purity Grade: Pure Assay: ≥95% Assay Percent Range: 96% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00077264 Synonym: Dimethylbenzene
Xylenes (Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Xylenes CAS: 1330-20-7 CAS: 100-41-4 Purity Grade: Laboratory Grade Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00077264 Synonym: Dimethylbenzene
Xylenes ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Xylenes CAS: 1330-20-7 Purity Grade: ACS Reagent Grade Molecular Formula: C6H4(CH3)2 Formula Weight: 106.17
Xylenes ACS Reagent Grade, Ricca Chemical
Chemical Name or Material: Xylenes CAS: 1330-20-7 CAS Min %: 98.0 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade Physical Form: Liquid
Xylenes , Macron Fine Chemicals™
Chemical Name or Material: Xylenes CAS: 1330-20-7 Molecular Formula: C6H4(CH3)2 Formula Weight: 106.17
Xylenes, 99.99%, For ACS analysis, MP Biomedicals™
Chemical Name or Material: Xylenes, ACS Name Note: For ACS analysis CAS: 1330-20-7 Physical Form: Liquid Assay Percent Range: 0.9999 Molecular Formula: C8H10 Formula Weight: 106.17 Synonym: Xylol
Xylenes, ACS Specifications, 98.5% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Xylenes Name Note: For ACS analysis CAS: 1330-20-7 Physical Form: Liquid Assay Percent Range: 98.5% min. Molecular Formula: C6H4(CH3)2 Formula Weight: 106.17 Synonym: Dimethylbenzenes
Xylenes ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Xylenes CAS: 1330-20-7 Purity Grade: ACS Reagent Grade Linear Formula: C6H4(CH3)2 Formula Weight: 106.17
Xylenes, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Xylenes CAS: 1330-20-7 Physical Form: Liquid Molecular Formula: C6H4(CH3)2 Formula Weight: 106.17
Xylenes, Histology, EMD Millipore
Chemical Name or Material: Xylenes CAS: 1330-20-7 Purity Grade: Histological Grade Synonym: Dimethylbenzenes
Xylenes, ACS Specifications, 98.5% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Xylenes Name Note: For ACS analysis CAS: 1330-20-7 Physical Form: Liquid Assay Percent Range: 98.5% min. Molecular Formula: C6H4(CH3)2 Formula Weight: 106.17 Synonym: Dimethylbenzenes
Xylenes, GMP/ACS Grade, PHARMCO-AAPER
Chemical Name or Material: Xylenes CAS: 1330-20-7 CAS: 100-41-4 Physical Form: Liquid Molecular Formula: C8H10 Formula Weight: 106.17 Synonym: Dimethylbenzene
Xylenes Laboratory, Macron™
Chemical Name or Material: Xylenes CAS: 1330-20-7 Purity Grade: Laboratory Grade Molecular Formula: C6H4(CH3)2 Formula Weight: 106.17
  spinner