Yttrium -Y
Yttrium(III) oxide, 99.9999% (REO), Alfa Aesar™ Reacton™
Chemical Name or Material: Yttrium(III) oxide CAS: 1314-36-9 Physical Form: Powder Assay Percent Range: 99.9999% (REO) Molecular Formula: Y2O3 Formula Weight: 225.81 MDL Number: MFCD00011473
Yttrium Powder, -40 mesh, 0.999, Alfa Aesar™
Chemical Name or Material: Yttrium Name Note: Rare Earth Oxides Content CAS: 7440-65-5 Physical Form: Powder Assay Percent Range: 99.90% MDL Number: MFCD00011468
Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis), ACROS Organics™
Chemical Name or Material: Yttrium(III) nitrate hexahydrate Name Note: 99.9% CAS: 13494-98-9 Purity Grade: Trace Metal Basis Assay Percent Range: 99.9% Molecular Formula: N3O9Y·6H2O Linear Formula: Y(NO3)3·6H2O Formula Weight: 383.01 MDL Number: MFCD00149940
Yttrium(III) oxide, 99.99% (REO), Alfa Aesar™, Reacton™
Chemical Name or Material: Yttrium(III) oxide CAS: 1314-36-9 Physical Form: ≪10 Micron Powder Assay Percent Range: 99.99% (REO) Molecular Formula: O3Y2 Formula Weight: 225.81 MDL Number: MFCD00011473
Yttrium(III) oxide, 99.999%, (trace metal basis), ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Yttrium(III) oxide CAS: 1314-36-9 Purity Grade: For trace metal basis analysis Assay: 99.999% (Y2O3) (typically, on TREO base) Assay Percent Range: 100.00% Molecular Formula: O3Y2 Linear Formula: Y2O3 Formula Weight: 225.81 MDL Number: MFCD00011473
Yttrium(III) nitrate hexahydrate, 99.9% (REO), Alfa Aesar™
Chemical Name or Material: Yttrium(III) nitrate hexahydrate CAS: 13494-98-9 Physical Form: Crystalline Assay Percent Range: 99.9% (REO) Molecular Formula: Y(NO3)3·6H2O Formula Weight: 383.01; 274.92 (anhydrous) MDL Number: MFCD00149940
Yttrium(III) acetate tetrahydrate, 99.99% (REO), Alfa Aesar™, Reacton™
Chemical Name or Material: Yttrium(III) acetate tetrahydrate CAS: 85949-60-6 Physical Form: Crystalline Aggregates Assay Percent Range: 99.99% (REO) Molecular Formula: C6H9O6Y·4H2O Formula Weight: 338.10; 266.04 (anhydrous) MDL Number: MFCD00013048
Yttrium(III) oxide, REacton™, 99.9% (REO)
Chemical Name or Material: Yttrium(III) oxide CAS: 1314-36-9 Purity Grade: REacton™ Molecular Formula: Y2O3 Formula Weight: 225.81 MDL Number: MFCD00011473 Merck Index: 14,10107
Yttrium(III) Chloride, Anhydrous, 99.9% (REO), Alfa Aesar™
Chemical Name or Material: Yttrium(III) Chloride Name Note: Rare Earth Oxides Content CAS: 10361-92-9 Physical Form: -20 Mesh Assay Percent Range: 99.9% Molecular Formula: Cl3Y Formula Weight: 195.26 MDL Number: MFCD00011470
Yttrium(III) oxide, 99.999% (REO), REacton™, Alfa Aesar™
CAS: 1314-36-9 Physical Form: ≪10 Micron Powder Assay Percent Range: 99.999% (REO) Molecular Formula: Y2O3 Formula Weight: 225.81 MDL Number: MFCD00011473
Yttrium Foil, 0.25mm (0.01 in.) thick, 0.999, Alfa Aesar™
Chemical Name or Material: Yttrium Name Note: Rare Earth Oxides Content CAS: 7440-65-5 Physical Form: Foil, 0.25mm (0.01 in.) thick Assay Percent Range: 99.90% MDL Number: MFCD00011468
Yttrium(III) oxide, 99.99%, (trace metal basis), ACROS Organics™
Chemical Name or Material: Yttrium(III) oxide CAS: 1314-36-9 Purity Grade: For trace metal basis analysis Assay: ≥99.99% (Y2O3) (TREO base) Assay Percent Range: 99.99% Molecular Formula: O3Y2 Linear Formula: Y2O3 Formula Weight: 225.81 MDL Number: MFCD00011473
Yttrium Foil, 0.64mm (0.025 in.) thick, 0.999, Alfa Aesar™
Chemical Name or Material: Yttrium Name Note: Rare Earth Oxides Content CAS: 7440-65-5 Physical Form: Foil, 0.64mm (0.025 in.) thick Assay Percent Range: 99.90% MDL Number: MFCD00011468
Yttrium acetate hydrate 99.9%, ACROS Organics™
Chemical Name or Material: Yttrium acetate hydrate CAS: 207801-28-3 Purity Grade: For trace metal basis analysis Assay Percent Range: 99.90% Molecular Formula: C6H9O6Y·xH2O Formula Weight: 266.04 MDL Number: MFCD00150128
Yttrium(III) chloride hexahydrate, 99.999%, (trace metal basis), ACROS Organics™
Chemical Name or Material: Yttrium(III) chloride hexahydrate Name Note: 99.999% CAS: 10025-94-2 Purity Grade: For trace metal basis analysis Assay: ≥99.999% (metals basis) Assay Percent Range: 100.00% Molecular Formula: Cl3Y·6H2O Linear Formula: YCl3·6H2O Formula Weight: 303.35 MDL Number: MFCD00149941
Yttrium(III) chloride hexahydrate, 99.9%, (trace metal basis), ACROS Organics™
Chemical Name or Material: Yttrium(III) chloride hexahydrate Name Note: 99.9% CAS: 10025-94-2 Purity Grade: For trace metal basis analysis Assay Percent Range: 99.90% Molecular Formula: Cl3Y·6H2O Linear Formula: YCl3·6H2O Formula Weight: 303.35 MDL Number: MFCD00149941
Yttrium Pieces, 0.999, Alfa Aesar™
Chemical Name or Material: Yttrium Name Note: Rare Earth Oxides Content CAS: 7440-65-5 Physical Form: Pieces Assay Percent Range: 99.90% MDL Number: MFCD00011468
Yttrium Oxide Products for Science Education
Chemical Name or Material: Yttrium Oxide CAS: 1314-36-9 Linear Formula: Y2O3 Formula Weight: 225.81 Synonym: Yttrium (III) Oxide
Ytterbium(III) chloride hexahydrate, 99.9%, (trace metal basis), ACROS Organics™
Chemical Name or Material: Ytterbium(III) chloride hexahydrate CAS: 10035-01-5 Purity Grade: For trace metal basis analysis Assay Percent Range: 99.90% Molecular Formula: Cl3Yb·6H2O Linear Formula: YbCl3·6H2O Formula Weight: 387.48 MDL Number: MFCD00149885
Yttrium Chloride, Anh., MP Biomedicals
Chemical Name or Material: Yttrium Chloride CAS: 10361-92-9 Formula Weight: 195.26
Yttrium Tungstate, 99.99%, MP Biomedicals
Chemical Name or Material: Yttrium Tungstate Formula Weight: 921.35
Yttrium Oxide, 99%+, EMD Millipore™
Chemical Name or Material: Yttrium Oxide CAS: 1314-36-9 Molecular Formula: Y2O3 Formula Weight: 225.81. (molar mass)
Yttrium(III) oxide, Aerosol Refractory Paint, 99.9% (REO), Alfa Aesar™
Chemical Name or Material: Yttrium(III) oxide Name Note: Maximum temperature use is 1900°C air, vacuum or inert Assay Percent Range: 99.9% (REO) Molecular Formula: Y2O3 MDL Number: MFCD00011473
Yttrium Nitrate Hexahydrate (Crystalline/Laboratory), Fisher Chemical
Chemical Name or Material: Yttrium Nitrate Hexahydrate CAS: 13494-98-9 Purity Grade: Laboratory Grade Physical Form: Solid Molecular Formula: Y(NO3)3 · 6 H2O Linear Formula: Y(NO3)3·6H2O Formula Weight: 383.01 MDL Number: MFCD00149940
  spinner