Zirconium -Zr
Zirconium Sponge, 25.4mm (1.0 in.) and down, 99.2% (Metals basis), Alfa Aesar™
Chemical Name or Material: Zirconium CAS: 7440-67-7 Physical Form: Sponge, 25.4mm (1.0 in.) and Down Assay Percent Range: 99.2% (Metals basis) MDL Number: MFCD00011303
Zirconium(IV) iodide, 99%, Alfa Aesar™
CAS: 13986-26-0 Physical Form: Powder Assay Percent Range: 99% Molecular Formula: I4Zr Formula Weight: 598.84 MDL Number: MFCD00049641 Synonym: Zirconium tetraiodide
Zirconium Oxide, Catalyst Support, Median pore diameter 80/400Å, Alfa Aesar™
Chemical Name or Material: Zirconium oxide, catalyst support Name Note: 1/8 in. Pellets MDL Number: MFCD00011310
Zirconium silicate, 98%, -325 mesh, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Zirconium silicate Name Note: 99%, -325 mesh CAS: 10101-52-7 Assay Percent Range: 98% Molecular Formula: SiO4Zr Linear Formula: ZrSiO4 Formula Weight: 183.3 MDL Number: MFCD00085353
Zirconium(IV) chloride, Reactor grade, >99.5% (metals basis), Alfa Aesar™
Chemical Name or Material: Zirconium(IV) chloride CAS: 10026-11-6 Physical Form: Powder Assay Percent Range: >99.5% (metals basis) Molecular Formula: ZrCl4 Formula Weight: 233.03 MDL Number: MFCD00011306 Synonym: Zirconium tetrachloride
Zirconium Oxide, Catalyst Support, Tungstated, Alfa Aesar™
Chemical Name or Material: Zirconium Oxide MDL Number: MFCD00168096
Zirconium oxide, Yttria stabilized, Grinding Pieces, 10mm diameter, Alfa Aesar™
Chemical Name or Material: Zirconium oxide MDL Number: MFCD00145546
Zirconium nitride, 99.5% (metals basis excluding Hf), Hf >3%, Alfa Aesar™
Chemical Name or Material: Zirconium nitride CAS: 25658-42-8 Physical Form: -325 Mesh Powder Assay Percent Range: 99.5% (metals basis excluding Hf) Molecular Formula: NZr Formula Weight: 105.23 MDL Number: MFCD00049642
Zirconium fluoride, 99.5%, ACROS Organics™
Chemical Name or Material: Zirconium fluoride Name Note: 99.5% CAS: 7783-64-4 Assay: ≥99.4% Assay Percent Range: 99.50% Molecular Formula: F4Zr Linear Formula: ZrF4 Formula Weight: 167.21 MDL Number: MFCD00011307
Zirconium Oxide Anhydrous (Powder/Laboratory), Fisher Chemical
Chemical Name or Material: Zirconium Oxide Anhydrous CAS: 1314-23-4 Purity Grade: Laboratory Grade Physical Form: Solid Molecular Formula: ZrO2 Formula Weight: 123.22 MDL Number: MFCD00011310 Synonym: Zirconium Dioxide
Zirconium(IV) oxide, cement, Alfa Aesar™
Chemical Name or Material: Zirconium(IV) oxide CAS: 1314-23-4 MDL Number: MFCD01868884
Zirconium Oxide, Catalyst Support, Sulfated, Alfa Aesar™
Chemical Name or Material: Zirconium Oxide MDL Number: MFCD01863639
Zirconium Foil, 0.127mm (0.005 in.) thick, annealed, 99.5% (Metals basis), Alfa Aesar™
Chemical Name or Material: Zirconium Name Note: Annealed CAS: 7440-67-7 Physical Form: Foil, 0.127mm (0.005 in.) thick Assay Percent Range: 99.5% (Metals basis) MDL Number: MFCD00011303
Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf, ACROS Organics™
Chemical Name or Material: Zirconyl(IV) nitrate hydrate Name Note: 99.5% CAS: 14985-18-3 Assay Percent Range: 99.50% Molecular Formula: N2O7Zr·xH2O Linear Formula: ZrO(NO3)2·xH2O Formula Weight: 231.23 MDL Number: MFCD00149896
Zirconium(IV) chloride, 98%, anhydrous, Acros Organics
Chemical Name or Material: Zirconium(IV) chloride Name Note: 98% CAS: 10026-11-6 Assay: ≥97.5% (ZrCl4) Assay Percent Range: 98% Molecular Formula: Cl4Zr Linear Formula: ZrCl4 Formula Weight: 233.03 MDL Number: MFCD00011306 Synonym: Zirconium tetrachloride
Zirconium powder, Alfa Aesar™
Chemical Name or Material: Zirconium CAS: 7440-67-7 Physical Form: Powder MDL Number: MFCD00011303
Barium zirconium oxide, 99% (metals basis), Alfa Aesar™
Chemical Name or Material: Barium zirconium oxide CAS: 12009-21-1 Physical Form: Powder Assay Percent Range: 99% (Metal basis) Molecular Formula: BaZrO3 Formula Weight: 276.53 MDL Number: MFCD00014189 Synonym: Barium zirconate
Zirconium Oxide, Catalyst Support, Median pore diameter 160/600Å, Alfa Aesar™
Chemical Name or Material: Zirconium Oxide Name Note: Catalyst Support Physical Form: Pellets, 3.2mm (0.125 in.) MDL Number: MFCD00011310
Zirconium Slug, 6.35mm(0.25in.) dia. x 12.7mm(0.50in.)L, 99.95% (Metals basis not Hf), Alfa Aesar™
Chemical Name or Material: Zirconium CAS: 7440-67-7 Physical Form: Slug, 6.35mm (0.25 in.) dia. X 12.7mm (0.50 in.) length Assay Percent Range: 99.95% (Metals basis excluding Hf) MDL Number: MFCD00011303
Zirconium dinitrate oxide hydrate, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Zirconium dinitrate oxide hydrate CAS: 14985-18-3 Physical Form: Crystalline Assay Percent Range: 99.9% (metals basis) Molecular Formula: ZrO(NO3)2·xH2O MDL Number: MFCD00149896 Synonym: Zirconium oxynitrate
Zirconium oxide, Yttria stabilized, Grinding Pieces, 5mm diameter, Alfa Aesar™
Chemical Name or Material: Zirconium oxide MDL Number: MFCD00145546
Zirconium Lump, 3 to 6mm (0.1 to 0.2 in.), 99.8% (Metals basis excluding Hf), Alfa Aesar™
Chemical Name or Material: Zirconium CAS: 7440-67-7 Physical Form: Lump, 3 To 6mm (0.1 to 0.2 in.) Assay Percent Range: 99.8% (Metals basis excluding Hf) MDL Number: MFCD00011303
Zirconium Wire, 0.127mm (0.005 in.) dia., 99.95% (Metals basis excluding Hf), Alfa Aesar™
Chemical Name or Material: Zirconium CAS: 7440-67-7 Physical Form: Wire, 0.127mm (0.005 in.) dia. Assay Percent Range: 99.95% (Metals basis excluding Hf) MDL Number: MFCD00011303
Zirconium(IV) oxide, 98.5%, Acros Organics
Chemical Name or Material: Zirconium(IV) oxide Name Note: 98.5% CAS: 1314-23-4 Assay: ≥98.0% (ZrO2 + HfO2) Assay Percent Range: 98.5% Molecular Formula: O2Zr Linear Formula: ZrO2 Formula Weight: 123.22 MDL Number: MFCD00011310
Zirconium Dichloride Oxide Hydrate, 99.9985% (Metal Basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Zirconium Dichloride Oxide Hydrate CAS: 15461-27-5 Physical Form: Crystalline Aggregates Assay Percent Range: 99.9985%, metal basis Molecular Formula: Cl2OZr·xH2O Formula Weight: 178.13 MDL Number: MFCD00149897
Zirconium sponge, 99.5%, Zr and Hf, Alfa Aesar™
Chemical Name or Material: Zirconium CAS: 7440-67-7 Physical Form: Sponge, 0.8 To 25.4mm (0.03 To 1.0 in.) Assay Percent Range: 99.5% MDL Number: MFCD00011303
Zirconium(IV) Oxide, Yttria Stabilized, 99% (Metal Basis excluding Hf), Alfa Aesar™
Chemical Name or Material: Zirconium(IV) Oxide CAS: 1314-23-4 Physical Form: -325 Mesh Powder Assay Percent Range: 99%, metal basis excluding Hf Molecular Formula: ZrO2 + 10 to 15% Y2O3 MDL Number: MFCD00145546
Zirconium(IV) oxide, 99.7% (metals basis excluding Hf), Hf <75ppm; Alfa Aesar™
Name Note: Hf < 75ppm CAS: 1314-23-4 Physical Form: 3 to 12mm Sintered Lump Assay Percent Range: 99.7% metal basis excluding Hf Molecular Formula: ZrO2 Formula Weight: 123.22 MDL Number: MFCD00011310
Zirconium Dichloride Oxide Octahydrate, 99.9% (Metal Basis), Alfa Aesar™
Chemical Name or Material: Zirconium Dichloride Oxide CAS: 13520-92-8 Physical Form: Lumps Assay Percent Range: 99.9%, metal basis Molecular Formula: ZrOCl2·8H2O Formula Weight: 322.25 (178.13 Anhydrous) MDL Number: MFCD00149898 Synonym: Zirconium oxychloride
Zirconium(IV) Oxide, 18% in H2O, Colloidal Dispersion, Alfa Aesar™
Chemical Name or Material: Zirconium(IV) Oxide Name Note: Colloidal Dispersion CAS: 1314-23-4 Physical Form: Liquid, 0.1μm Particles Molecular Formula: ZrO2/Y2O3 MDL Number: MFCD00011310
  spinner