Stirring Rods
Bel-Art™ SP Scienceware™ Spinplus™ Magnetic Stirring Bars Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Spinplus™ magnetic stirring bars add speed and efficiency to mixing operations
Bel-Art™ SP Scienceware™ Spinbar™ Capsule Magnetic Stirring Bars Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
1.18 x 0.31 in. polygon magnetic stirring bar spins freely inside a stationary capsule, excellent for pH adjustments of small volumes.
United Scientific Supplies Stirring Rods with Rubber Policeman Products for Science Education
Glass stirring rods are supplied with rubber policemen already attached.
Minimixer Stir Bar Products for Science Education
For use with Minimixers
Thermo Scientific™ Nalgene™ Suspended Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Use Nalgene Floating Magnetic Stir Bars for constant low-speed stirring action in tissue culture applications.
Thermo Scientific™ Komet Stir Bars
Generate over three times the torque of conventional stirring bars with Thermo Scientific™ Komet Stir Bars, significantly enhancing stirrer performance and versatility.
Fisherbrand™ PTFE Disposable Stir Bars Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
The multifaceted magnetic stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars
Fisherbrand™ Octagon Magnetic Stir Bars Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape
Fisherbrand™ Soft Glass Rods Products for Science Education
With rounded and fire-polished ends.
Fisherbrand™ Octagon Magnetic Stir Bar Products for Science Education
Octagonal magnetic stirring bars with integral pivot ring are the most commonly used shape
Thermo Scientific™ Nalgene™ StarHead™ Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Greater mixing action at bottom, side, and top of container, even at low stirring speeds
Thermo Scientific™ Nalgene™ Polypropylene Stirring Rod Available on GSA/VA Contract for Federal Government customers only.
Combine functions of a stirring rod and rubber policeman with these polypropylene stirring rods, featuring a flat paddle at one end and a triangular paddle at the other end.
Fisherbrand™ Round Stir Bars with Removable Pivot Ring Available on GSA/VA Contract for Federal Government customers only.
Stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars
Kimble™ Konte™s Round PTFE Micro Magnetic Stir Bar
PTFE-covered round magnetic stirring bar
Bel-Art™ SP Scienceware™ Spinbar™ Octagon Rare Earth Magnetic Stir Bar
Rare Earth/Samarium Cobalt magnet provides strong coupling with drive magnets to reduce frequency of spinout
Bel-Art™ SP Scienceware™ Flea Micro Spinbar™ Magnetic Stirring Bars Available on GSA/VA Contract for Federal Government customers only.
Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
Fisherbrand™ Magnetic Stir Bar Six Packs Available on GSA/VA Contract for Federal Government customers only.
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape
Bel-Art™ SP Scienceware™ Pyrex™ Glass Round Spinbar™ Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Useful for high temperature applications up to 274°C (525°F)
Fisherbrand™ Magnetic Stirring Bars Available on GSA/VA Contract for Federal Government customers only.
Stirbar assortments come in a convenient storage box. Fisherbrand™ Magnetic Stirring Bars contain PTFE encapsulated Alnico magnets and come in various sizes.
Fisherbrand™ Solid Silicone Stoppers
Create a tight, reliable seal in virtually all sealing applications
SP SCIENCEWARE BEL ART SP Scienceware™ Polygon Spinbar™ Magnetic Stirring Bars without Pivot Ring Available on GSA/VA Contract for Federal Government customers only.
Adds turbulence relative to similar smooth size cylindrical bars. SP Scienceware™ Polygon Spinbar™ Magnetic Stirring Bars without Pivot Ring are manufactured in a registered ISO 9001:2008 facility.
Qorpak™ Cylindrical Stir Bars Available on GSA/VA Contract for Federal Government customers only.
With Alinco V magnet cores and Isostatic encapsulation that eliminate cracks and porosity
Fisherbrand™ 8-Agon Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape. Choose from red, yellow or blue.
Bel-Art™ SP Scienceware™ Stir and Add™ Cuvet Mixer Available on GSA/VA Contract for Federal Government customers only.
Made of clear acrylic with a straight handle, this tool is used when reactions are performed directly into cuvettes, to mix added reagents
Fisherbrand™ Magnetic Micro Stirring Bars Available on GSA/VA Contract for Federal Government customers only.
Micro magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes and gradient makers
  spinner