Stirring Rods
United Scientific Supplies Stirring Rods with Rubber Policeman Products for Science Education
Glass stirring rods are supplied with rubber policemen already attached.
Fisherbrand™ Soft Glass Rods Products for Science Education
With rounded and fire-polished ends.
Fisherbrand™ Soft Glass Stirring Rods Available on GSA/VA Contract for Federal Government customers only.
Available in different lengths ranging from 5 to 10 in.
Bel-Art™ SP Scienceware™ Bosshead Rod holder
Adjustable with large flat wing head screws
Bel-Art™ SP Scienceware™ Stir and Add™ Cuvet Mixer Available on GSA/VA Contract for Federal Government customers only.
Made of clear acrylic with a straight handle, this tool is used when reactions are performed directly into cuvettes, to mix added reagents
Kimble™ KIMAX™ Solid Stirring Rods Available on GSA/VA Contract for Federal Government customers only.
Solid construction for repeated use
Saint-Gobain™ Chemware™ Bendable PTFE Stirring Rods
Contain fully enclosed steel insert for stiffness, yet can be bent to various shapes for use as powered stirrers
  spinner