Rubber Stoppers
Fisherbrand™ One-Hole Rubber Stoppers Products for Science Education
Black Natural Rubber/SBR Blend for general purpose uses
Fisherbrand™ Solid Rubber Stoppers Products for Science Education
Black Natural Rubber/SBR Blend for general purpose uses
3B Scientific™ Rubber Stopper Sets Products for Science Education
Available with no hole, one hole and two holes.
Fisherbrand™ Solid Rubber Stoppers
Available in a variety of sizes ranging from 000 through 16
Wheaton™ Ultra Pure Complete Coat Stoppers Available on GSA/VA Contract for Federal Government customers only.
Featuring PTFE-like coatings to prevent reactions due to sample contact with rubber. Wheaton™ Ultra Pure Complete Coat Stoppers are manufactured in a silicone-free environment.
Fisherbrand™ Solid Neoprene Rubber Stoppers
Recommended where rubber would be attacked or deteriorated
Fisherbrand™ Twistit™ Rubber Stoppers
Stoppers made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation
Wheaton™ Ultra Pure Straight Plug Stoppers Available on GSA/VA Contract for Federal Government customers only.
Provides for extremely high chemical purity and low gas permeability. Wheaton™ Ultra Pure Straight Plug Stoppers are manufactured from an ultra pure bromobutyl formulation.
Fisherbrand™ Turnover Septum Stoppers
For narrow-aperture containers
Wheaton™ Straight Plug Style Stoppers for Vials and Serum Bottles
For maximum sealing of vials with straight wall glass finish
Fisherbrand™ Silicone Recessed Septum Stoppers
Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal
Wheaton™ Snap-on Style Stoppers for Vials and Serum Bottles
For maximum sealing of vials with blow-back glass finish
Wheaton™ Flange Style Rubber Stopper Available on GSA/VA Contract for Federal Government customers only.
For research and parenteral drug applications
Wheaton™ Sleeve Style Stoppers for Vials without Crimp Closure
Provides injection site for multiple dose entry
United Scientific Supplies Rubber Stoppers, Solid Version Products for Science Education
Ideal for use with non-aggressive or concentrated solutions. United Scientific Supplies Rubber Stoppers, Solid Version withstand temperatures from -13° to 158°F (-25° to 70°C).
United Scientific Supplies Rubber Stoppers, One Hole Products for Science Education
Ideal for use with non-aggressive or concentrated solutions. United Scientific Supplies Rubber Stoppers, One Hole withstand temperatures from -13° to 158°F (-25° to 70°C).
United Scientific Supplies Rubber Stoppers, Two Hole Products for Science Education
Ideal for use with non-aggressive or concentrated solutions. United Scientific Supplies Rubber Stoppers, Two Hole withstand temperatures from -13° to 158°F (-25° to 70°C).
  spinner