Silicone Stoppers
Fisherbrand™ Solid Silicone Stoppers
Create a tight, reliable seal in virtually all sealing applications
DURAN™ GL 18 Silicone Seal Septa
Suitable for PBT screw cap with aperture. Heat resistance: 130°C (vapor) and 200°C (dry heat). Silicone is peroxide-cured
  spinner