Filter Units
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with Nylon Membrane Available on GSA/VA Contract for Federal Government customers only.
Filter solutions where very low extractables are desired and solutions containing weak solvents with these disposable filter units with nylon membrane.
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with CN Membrane Available on GSA/VA Contract for Federal Government customers only.
Perform sterile filtration, prefiltration or clarification of buffers and other aqueous solutions when protein binding is not a concern with these filter units.
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES Membrane Available on GSA/VA Contract for Federal Government customers only.
Increase filtration efficiency with PES membranes and Rapid-Flow design, providing fast flow rates and high throughput while defending against cell culture contamination.
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with SFCA Membrane Available on GSA/VA Contract for Federal Government customers only.
Perform vacuum filtration and single-use biological work requiring sterile filtration, prefiltration or clarification of aqueous solutions in the lab or field.
EMD Millipore Stericup™ Sterile Vacuum Filter Units
Designed for sterilizing tissue culture media, protein and DNA solutions. EMD Millipore Stericup™ Sterile Vacuum Filter Units have a no-tip design for improved stability during filtration. Steritop bottle top filter units use either a PES or PVDF filter. Available in varied pore sizes and volumes.
EMD Millipore Steriflip™ Sterile Disposable Vacuum Filter Units
Performance and convenience all in one.  Merck Millipore™ Steriflip™ Sterile Disposable Vacuum Filter Units uses vacuum to collect filtrate.
EMD Millipore Nylon-Net Steriflip™ Vacuum Filter Unit
Sterile, disposable filter unit for cell isolation
Whatman™ Glass Vacuum Filtration Devices Available on GSA/VA Contract for Federal Government customers only.
Equipment and accessories for vacuum filtration
Sartolab™ P20 Plus Filter Systems Available on GSA/VA Contract for Federal Government customers only.
Filter tissue culture solutions quickly, economically and reliably. Sartorius™ Sartolab™ P20 Plus Filter Systems are sterile, ready-to-use units. A large filtration area and automatic de-aeration ensures high flow rates. Available with PES or SFCA membrane, different connectors or with a prefilter to process up to 10L.
Sartorius™ Sartolab™ RF/BT Vacuum Filtration Units Available on GSA/VA Contract for Federal Government customers only.
Filter aqueous solutions quickly and easily. Sartorius™ Sartolab™ RF/BT Vacuum Filtration Units are disposable and easy to use filter units. They contain a high quality PES membrane and combine the highest flow-rates and throughput with extremely low protein binding and extractables. Available in a range of volumes.
Sartorius™ Biosart™ 100 Monitors Available on GSA/VA Contract for Federal Government customers only.
Designed specifically for microbiological testing of pharmaceuticals, food, beverages, water and other liquids. Sartorius™ Biosart™ 100 Monitors are sterile disposables with an incorporated membrane filter and cellulose pad are ready to use. The printed grid lines make the counting easier, especially for higher bacteria counts and for micro-colonies, but does not influence the growth.
Sartorius™ Polycarbonate Filtration Holders Available on GSA/VA Contract for Federal Government customers only.
For the clarification or sterile filtration of up to about 200ml volumes of aqueous solutions
Sartorius™ Equipment and Accessories for Vacuum Filter Holders Available on GSA/VA Contract for Federal Government customers only.
Choose the best accessories to support your filtration applications. Sartorius™ Equipment and Accessories for Vacuum Filter Holders include an all-glass vacuum filter holder assembly plus a hose, hand-held pump with gauge, a 3-way valve, a 3-way tap and a stainless steel connector.
Sartorius™ Microsart™ Funnels Available on GSA/VA Contract for Federal Government customers only.
Ready-to-use, sterile, polypropylene funnels with click-fit closure
Sartorius™ In-Line Filter Holders Available on GSA/VA Contract for Federal Government customers only.
Designed for use in pressure applications in batch or continuous filtration processes. Sartorius™ In-Line Filter Holders are available in polycarbonate for compatibility with many simple laboratory filtrations or stainless steel for compatibility with aggressive liquids and gasses.
Sartorius™ Biosart™ 250 Funnels Available on GSA/VA Contract for Federal Government customers only.
Sterile Funnels, ready-to-use
Sartorius™ Syringe Filter Holders Available on GSA/VA Contract for Federal Government customers only.
Achieve leak-free filtration up to 7bar. Sartorius™ In-Line Filter Holders are inexpensive, reusable holders made of clear polycarbonate with silicone gaskets. Auclavable units are available for 25mm and 13mm syringe filters.
  spinner