Extraction Thimbles
Fisherbrand™ Filtration Extraction Thimbles - Grade 645 Cellulose
Ensures complete extraction of samples, prevents contamination and minimize hands-on time
PYREX™ Filter Tubes Available on GSA/VA Contract for Federal Government customers only.
Used to collect filtrate
Fisherbrand™ Filtration Extraction Thimbles - Grade 649 Glass Fibre
Ensures complete extraction of samples, prevents contamination and minimize hands-on time
PYREX™ Glass Extraction Thimbles Available on GSA/VA Contract for Federal Government customers only.
Resist attack by most chemicals
Whatman™ Cellulose Soxhlet Extraction Thimbles, single 1mm wall
Manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Whatman™ Cellulose Soxhlet Extraction Thimbles are intended for applications including use in air and waste gas analysis.
Whatman™ Glass Microfiber Extraction Thimbles, Grade 603G Available on GSA/VA Contract for Federal Government customers only.
Flexibly designed for use in a full range of commercially available Soxhlet apparatus systems. Whatman™ Glass Microfiber Extraction Thimbles are designed for applications including analysis of hot and acidic gases, analysis of pesticide residues, determination of oil/fat content of foods, and analysis of oil and grease in solid wastes.
Whatman™ Quartz Microfiber Extraction Thimbles for Emission Testing
Support accurate, reproducible emission testing by providing low extractable levels and temperature resistance up to 800°C. Whatman™ Quartz Microfiber Extraction Thimbles are tapered for stack sample loading. Applications include emission testing, smoke stack gas sampling, gravimetric calculations in gases, analysis of hot and acidic gases, trace element analysis, and analysis of pesticides and residues.
Whatman™ High Purity Glass Microfiber Soxhlet Extraction Thimbles
Designed to fit most Soxhlet extraction units. Whatman™ High Purity Glass Microfiber Soxhlet Extraction Thimbles are made of 100% borosilicate glass microfiber. They are well-suited to pollution monitoring when analysis of hot and acidic gases is required.
Whatman™ Cellulose Extraction Thimbles Available on GSA/VA Contract for Federal Government customers only.
Ensures optimal fit with its wide range of sizes suitable for use in thimble holders and automated extraction apparatus. Whatman™ Cellulose Extraction Thimbles are constructed of high-quality cellulose for excellent mechanical strength and retention.
Whatman™ Grade 603 Cellulose Extraction Thimbles Available on GSA/VA Contract for Federal Government customers only.
Range in size and length to ensure optimal fit.. Whatman™ Grade 603 Cellulose Extraction Thimbles are for use with extracting pesticides from environmental samples and fat analysis in the food industry
Whatman™ Cellulose Soxhlet Extraction Thimbles, double 2mm wall
Manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Whatman™ Cellulose Soxhlet Extraction Thimbles are intended for applications including use in air and waste gas analysis.
Whatman™ Glass Microfiber Extraction Thimbles, Grade 603G, without binder Available on GSA/VA Contract for Federal Government customers only.
Use for analysis of hot and acidic gases, including emission control and smoke tack gas monitoring. Whatman™ Glass Extraction Thimbles are designed to fit most Soxhlet extraction units and can be used with solvents that are incompatible with cellulose thimbles.
Labconco™ Alundum™ Extraction Thimbles
Goldfisch Fat Extractor replacement part
Whatman™ High-Purity Glass Thimbles Available on GSA/VA Contract for Federal Government customers only.
Use for pollution monitoring when analysis of hot and acidic gases is require. Whatman™ High-Purity Thimbles are designed to fit mosst Soxhlet extraction units and can be used with solvents that are incompatible with cellulose thimbles.
Whatman™ High-Purity Glass Thimbles Available on GSA/VA Contract for Federal Government customers only.
Use for pollution monitoring when analysis of hot and acidic gases is required. Whatman™ High-Purity Thimbles are designed to fit most Soxhlet extraction units and can be used with solvents that are incompatible with cellulose thimbles.
Whatman™ Extraction Thimble for DIONEX ASE 200
Excellent mechanical strength and retention
Saint-Gobain ALUNDUM™ Extraction Thimbles
Wash, ignite, and use repeatedly
Koehler™ Sediment Extraction Apparatus Accessories: Thimble Basket
Determines sediment content of crude oil and fuel oils by extraction with toluene
Ahlstrom™ Cellulose Extraction Thimble
Useful for estimating fats in food and analyzing lacquer and binder in paints
  spinner