Filter Paper
Fisherbrand™ Qualitative Grade Plain Filter Paper Circles - P5 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Fisherbrand™ Qualitative Grade Plain Filter Paper Circles and Sheets - P4 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Fisherbrand™ Qualitative Grade Plain Filter Paper Sheets - P8 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
EISCO Filter Paper Products for Science Education
Student quality filter paper.
Fisherbrand™ Quantitative Grade Filter Paper Circles
Manufactured to reduce silicon dioxide, calcium oxide and ferric oxide levels
Fisherbrand™ Qualitative Grade Plain Filter Paper Circles and Sheets - P2 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Fisherbrand™ Wet-Strengthened Qualitative Filter Paper Circles
Coarse cellulose fiber papers remove particles >25μm
Fisherbrand™ Qualitative Grade Plain Filter Paper Circles - P8 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Fisherbrand™ Qualitative Grade Plain Filter Paper Sheets - P5 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Fisherbrand™ Qualitative Filter Paper: Grade 6 Circles
Filters particles twice as fast as Qualitative Filter Paper Grade 5. Fisherbrand™ Qualitative Grade 6 Filters provide fine particle retention for boiler water analysis.
GE Whatman Quantitative Filter Paper: Grade 40 Sheets
Supports accurate analysis in applications sensitive to very low levels of background contamination. Fisherbrand™ Quantitative Filter Paper: Grade 40 Circles are additionally useful in gravimetric analysis where ash content might affect weight measurements of combusted samples.
Bel-Art™ SP Scienceware™ Filter Paper for Table-Top Buchner Funnels
Fitted specifically for Scienceware table-top Buchner funnels
GE Whatman Quantitative Filter Paper: Grade 41 Sheets
Use for quantitative air pollution analysis as a paper tape for impregnation when determining gaseous compounds at high flow rates or for quantitative air pollution analysis. Fisherbrand™ Quantitative Filter Paper: Grade 41 Sheets are commonly used in preparing samples for metal analysis after nitric acid digestion.
Whatman™ Glass Microfiber Filter Paper Grade 6 Available on GSA/VA Contract for Federal Government customers only.
Captures tiny particles without becoming weak. Whatman™ Glass Microfiber Filter Paper Grade 6 may also be used for determination of filterable substances and residue upon ignition or analysis of aggressive media.
Whatman™ Qualitative Filter Paper: Grade 1574½ Pleated Circles Available on GSA/VA Contract for Federal Government customers only.
Maintains flow rate with reduced contact between filter and funnel. Whatman™ Grade1574½ Pleated Circles are a medium fast filter paper with high wet strength.
Whatman™ Grade 54 Quantitative Filter Paper
Supports collection of coarse and gelatinous precipitates with minimal fiber shredding, even when wet. Whatman™ Grade 54 Quantitative Filter Paper has a hard, smooth surface to sustain vacuum and suction filtration.
Whatman™ Qualitative Filter Paper: Grade 2 Circles
Retains and absorbs more than Grade 1 filters, with a corresponding increase in filtration time. Whatman™ Qualitative Filter Paper: Grade 2 Circles are used for monitoring specific contaminants in the atmosphere and in soil testing.
Whatman™ Qualitative Filter Paper: Grade 1 Sheets
Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Sheets are widely used standard grade filter paper for routine applications with medium retention and flow rates.
Whatman™ Qualitative Filter Paper: Grade 1 Circles
Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Circles are widely used standard grade filter papers for routine applications with medium retention and flow rates.
Whatman™ Grade 2V Folded Qualitative Filter Papers Available on GSA/VA Contract for Federal Government customers only.
Saves time required to fit circles into conical funnels. Whatman™ Grade 2V Folded Qualitative Filter Papers provide medium flow and nominal particle retention rating of 8µm at a higher filtration speed.
Whatman™ Qualitative Filter Papers – Grade 520 B Available on GSA/VA Contract for Federal Government customers only.
Filters particles of 15μm. Whatman™ Qualitative Filter Papers – Grade 520 B with high wet strength and flow rate are designed for technical use.
Whatman™ Quantitative Filter Paper: Grade 540 Circles
Use as a gravimetric filter during analysis of metals in acidic or alkaline solutions and in the collection of hydroxides. Whatman™ Quantitative Filter Paper: Grade 540 Circles acid-treated surface prevents tearing during filtration and allows the user to scrape precipitates from the paper.
Whatman™ Grade 52 Quantitative Filter Paper
Enables efficient precipitate recovery with acid-hardened surface and low ash content. Whatman™ Grade 52 Quantitative Filter Paper prevents fiber shredding during vacuum filtration or precipitate collection.
Whatman™ Qualitative Filter Paper: Grade 595½ Pleated Circles Available on GSA/VA Contract for Federal Government customers only.
Pleated to save time and achieve optimal fit in conical filter funnels. Whatman™ Qualitative Filter Paper: Grade 595½ Pleated Circles provide extra surface area to increase total loading capacity.
  spinner