Quantitative Filter Paper
Fisherbrand™ Qualitative Filter Paper: Grade 6 Circles
Filters particles twice as fast as Qualitative Filter Paper Grade 5. Fisherbrand™ Qualitative Grade 6 Filters provide fine particle retention for boiler water analysis.
Fisherbrand™ Qualitative Grade Plain Filter Paper Circles and Sheets - P4 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Bel-Art™ SP Scienceware™ Filter Paper for Table-Top Buchner Funnels
Fitted specifically for Scienceware table-top Buchner funnels
Fisherbrand™ Wet-Strengthened Qualitative Filter Paper Circles
Coarse cellulose fiber papers remove particles >25μm
Fisherbrand™ Quantitative Grade Filter Paper Circles
Manufactured to reduce silicon dioxide, calcium oxide and ferric oxide levels
Fisherbrand™ Qualitative Grade Plain Filter Paper Circles - P5 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Fisherbrand™ Qualitative Grade Plain Filter Paper Circles - P8 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Fisherbrand™ Qualitative Grade Plain Filter Paper Circles and Sheets - P2 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
EISCO Filter Paper Products for Science Education
Student quality filter paper.
Fisherbrand™ Qualitative Grade Plain Filter Paper Sheets - P5 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Fisherbrand™ Qualitative Grade Plain Filter Paper Sheets - P8 Grade
Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
Fisherbrand™ Quantitative Filter Paper: Grade 41 Sheets
Use for quantitative air pollution analysis as a paper tape for impregnation when determining gaseous compounds at high flow rates or for quantitative air pollution analysis. Fisherbrand™ Quantitative Filter Paper: Grade 41 Sheets are commonly used in preparing samples for metal analysis after nitric acid digestion.
Fisherbrand™ Quantitative Filter Paper: Grade 40 Sheets
Supports accurate analysis in applications sensitive to very low levels of background contamination. Fisherbrand™ Quantitative Filter Paper: Grade 40 Circles are additionally useful in gravimetric analysis where ash content might affect weight measurements of combusted samples.
Whatman™ Quartz Air Sampling Filters, Grade QM-A, Circle Available on GSA/VA Contract for Federal Government customers only.
Used in many air sampling filter applications, including sampling of acidic gases, stacks, flues, and aerosols. Whatman™ Quartz Air Sampling Filters are sequentially numbered according to U.S. Environmental Protection Agency (EPA) standards and are suitable for most applications.
Whatman™ Quantitative Filter Paper: Grade 41 Circles Available on GSA/VA Contract for Federal Government customers only.
Use for quantitative air pollution analysis as a paper tape for impregnation when determining gaseous compounds at high flow rates or for quantitative air pollution analysis. Whatman™ Quantitative Filter Paper: Grade 41 Circles are commonly used in preparing samples for metal analysis after nitric acid digestion.
Whatman™ Qualitative Filter Paper: Grade 1 Sheets
Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Sheets are widely used standard grade filter paper for routine applications with medium retention and flow rates.
Whatman™ Qualitative Filter Paper: Grade 595½ Pleated Circles Available on GSA/VA Contract for Federal Government customers only.
Pleated to save time and achieve optimal fit in conical filter funnels. Whatman™ Qualitative Filter Paper: Grade 595½ Pleated Circles provide extra surface area to increase total loading capacity.
Whatman™ Qualitative Filter Paper: Grade 5 Circles Available on GSA/VA Contract for Federal Government customers only.
Retains the fine precipitates encountered in chemical analysis. Whatman™ Qualitative Filter Paper - Grade 5 Circles are excellent clarifying filters for cloudy suspensions and for water and soil analysis.
Whatman™ Qualitative Filter Paper: Grade 113 Circles
Strengthened with chemically stable resin additive. Whatman™ Qualitative Filter Paper: Grade 113 Circles boast the fastest flow rate of the qualitative grades. Crepe surface promotes ultra high loading capacity.
Whatman™ Reeve Angel 202 Creped Qualitative Circles
For routine, noncritical laboratory filtration applications such as general chemistry, pharmacology, emulsions, petroleum testing, soaps and detergents
Whatman™ Grade 91 Wet Strengthened Qualitative Filter Papers - Circles
High wet strength due to the addition of chemically stable resin
Whatman™ Qualitative Filter Paper: Grade 602EH½ Pleated Circles Available on GSA/VA Contract for Federal Government customers only.
Maintains flow rate with reduced contact between filter and funnel. Whatman™Grade 602EH½ Pleated Circles are slow and dense filter papers used for very fine precipitates.
Whatman™ Qualitative Filter Paper: Grade 4 Circles
Filters and retains course particles for monitoring air pollution. Whatman™ Qualitative Filter Paper: Grade 4 Circles are used when high flow rates in air pollution monitoring are required and the collection of fine particles is not critical.
Whatman™ Grade 44 Quantitative Filter Paper Available on GSA/VA Contract for Federal Government customers only.
Reduces overall ash content and the amount of time required for filtration. Whatman™ Grade 44 Quantitative Filter Paper provides faster gravimetric analysis with filter thinner material.
Whatman™ Qualitative Filter Paper: Grade 2 Circles
Retains and absorbs more than Grade 1 filters, with a corresponding increase in filtration time. Whatman™ Qualitative Filter Paper: Grade 2 Circles are used for monitoring specific contaminants in the atmosphere and in soil testing.
  spinner