Nonsterile Syringe and Syringeless Filters
Thermo Scientific™ Target2™ PVDF Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Ensure cleaner sample extracts by removing interfering materials and fine particles prior to injection with these robust PVDF syringe filters.
Thermo Scientific™ Target2™ Regenerated Cellulose Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Ensure cleaner sample extracts by removing interfering materials and fine particles prior to injection with these robust regenerated cellulose syringe filters.
Thermo Scientific™ Titan3™ PVDF Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Ensure cleaner sample extracts by removing interfering materials and fine particles prior to injection with these robust PVDF syringe filters.
Fisher Scientific™ Basix™ Syringe Filters, Nylon, Non-Sterile
Attach to the end of syringes and filter contents as it is being forced out. Fisher Scientific™ Basix™ Syringe Filters, Nylon, Non-Sterile offers high sample throughput.
Thermo Scientific™ Target2™ Nylon Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Ensure cleaner sample extracts by removing interfering materials and fine particles prior to injection with these robust nylon syringe filters. Ideal for general HPLC use.
Thermo Scientific™ Nalgene™ 4mm Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Filter sample volume sizes of 0.5 to 1.0mL using Nalgene 4mm Syringe Filters.
Thermo Scientific™ Target2™ Polypropylene Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Ensure cleaner sample extracts by removing interfering materials and fine particles prior to injection with these robust polypropylene syringe filters.
Thermo Scientific™ Nalgene™ Syringe Prefilter Plus
Conduct two-stage filtration for sterilization of highly viscous solutions or solutions with high particulate loads.
Thermo Scientific™ Target2™ PTFE Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Ensure cleaner sample extracts by removing interfering materials and fine particles prior to injection with these robust PTFE syringe filters.
Thermo Scientific™ Nalgene™ 25mm Syringe Filters
Filter sample volumes of 10 to 100mL with these nonsterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.
Thermo Scientific™ Target2™ Cellulose Acetate Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Ensure cleaner sample extracts by removing interfering materials and fine particles prior to injection with these robust cellulose acetate (CA) syringe filters.
Thermo Scientific™ Nalgene™ 13mm Syringe Filters
Filter aqueous solutions and media or solvents in sample volumes from 2 to 10mL using Thermo Scientific™ Nalgene™ 13mm Syringe Filters.
Thermo Scientific™ SUN-SRi™ Titan 30mm Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Banded polypropylene housing ensures lab safety preventing the housing from bursting
Thermo Scientific™ Target2™ GMF (Glass MicroFiber) Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Ensure cleaner sample extracts by removing interfering materials and fine particles prior to injection with these robust GMF (glass microfiber) syringe filters.
Fisher Scientific™ Basix™ Syringe Filters, PTFE, Non-Sterile
Attach to the end of syringes and filter contents as it is being forced out. Fisher Scientific™ Basix™ Syringe Filters, PTFE, Non-Sterile offers high sample throughput.
Fisher Scientific™ Basix™ Syringe Filters, PVDF, Non-Sterile
Attach to the end of syringes and filter contents as it is being forced out. Fisher Scientific™ Basix™ Syringe Filters, PVDF, Non-Sterile offers high sample throughput.
Thermo Scientific™ Nalgene™ 25mm Syringe Filters, Nylon, 0.2µm pore, Sterile
Filter culture media and additives (SFCA), buffers (CA) and alcohol or DMSO (Nylon) in sample volumes of 10 to 50mL.
Thermo Scientific™ Target2™ PES (Polyethersulfone) Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Ensure cleaner sample extracts by removing interfering materials and fine particles prior to injection with these robust PES (polyethersulfone) syringe filters.
Thermo Scientific™ Nalgene™ Syringe Prefilter, 25mm diameter, polypropylene housing
Clarify small volumes of highly viscous solutions and remove large particulates from suspensions using this syringe prefilter.
Thermo Scientific™ Nalgene™ 25mm Syringe Filters, CA; 0.8µm pore, Sterile
Filter culture media and additives (SFCA), buffers (CA) and alcohol or DMSO (Nylon) in sample volumes of 10 to 50mL.
Thermo Scientific™ Nalgene™ 25mm Syringe Filters, Nylon, 0.45µm pore, Sterile
Filter culture media and additives (SFCA), buffers (CA) and alcohol or DMSO (Nylon) in sample volumes of 10 to 50mL.
Thermo Scientific™ Target2™ PTFE Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Strong, hydrophobic membrane is resistant to nearly all solvents, acids and bases
EMD Millipore 13mm Nonsterile Millex™ Syringe Filters - Nylon Membrane Available on GSA/VA Contract for Federal Government customers only.
Overmolded polypropylene housing and optimized housing design
EMD Millipore Millex™ Nonsterile Syringe Filters with PE Housing - PES Available on GSA/VA Contract for Federal Government customers only.
Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses
EMD Millipore Millex™ Nonsterile Syringe Filters with PE Housing - Hydrophilic PTFE Available on GSA/VA Contract for Federal Government customers only.
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
EMD Millipore Automation Compatible Filters Available on GSA/VA Contract for Federal Government customers only.
Trouble-free performance on a variety of automated systems
EMD Millipore 13mm Nonsterile Millex™ Syringe Filters - Hydrophobic PTFE Membrane Available on GSA/VA Contract for Federal Government customers only.
Overmolded polypropylene housing and optimized housing design
EMD Millipore Millex™ Nonsterile 25mm Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Achieve up to 50% greater throughput and easier filtration than traditional syringe filters. EMD Millipore HPF Millex™ Nonsterile Syringe Filters feature a choice of nylon or PVDF membranes for optimal, fine filtration. Non-sterile filters are compatible with multiple Zymark workstations.
Fisherbrand™ Syringe Filters: Nylon Membrane Available on GSA/VA Contract for Federal Government customers only.
Low hold-up volume helps you to recover virtually all of your sample after filtering
  spinner