Sterile Syringe and Syringeless Filters
EMD Millipore Millex™ Sterile Syringe Filters: Durapore™ PVFD Membrane Available on GSA/VA Contract for Federal Government customers only.
Designed for sterilizing and clarifying sterile solutions. EMD Millipore Millex™ Sterile Syringe Filters: MCE Membrane are ready-to-use, syringe-driven units that have never been touched by human hands during filter assembly. Feature 0.1, 0.22, 0.45 and 5.0µm pore sizes; medical-grade filters available.
EMD Millipore Aervent-50 Disposable Filters Available on GSA/VA Contract for Federal Government customers only.
Sterilizing-grade filters designed for critical small-scale gas line and air venting applications
EMD Millipore Millex™ Sterile Syringe Filters: MCE Membrane Available on GSA/VA Contract for Federal Government customers only.
Designed for sterilizing and clarifying sterile solutions. EMD Millipore Millex™ Sterile Syringe Filters: MCE Membrane are ready-to-use, syringe-driven units that have never been touched by human hands during filter assembly. Feature 0.22, 0.45 and 0.80µm pore sizes; medical-grade filters available.
Fisher Scientific™ Basix™ Syringe Filters, MCE, Sterile
Attach to the end of syringes and filter contents as it is being forced out. Fisher Scientific™ Basix™ Syringe Filters, MCE, Sterile offers high sample throughput.
EMD Millipore Millex™-GP Sterile Syringe Filters with PES Membrane Available on GSA/VA Contract for Federal Government customers only.
Designed for sterilizing and clarifying sterile solutions. EMD Millipore Millex™-GP Sterile Syringe Filters with PES Membrane are ready-to-use, syringe-driven units with a unique membrane that results in six times less protein binding. Available in a medical grade unit, with a PES or acrylic tip.
Corning™ Nylon Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Recommended for applications requiring very low extractables because they do not contain any wetting agents, detergents or surfactants. Corning™ Nylon Syringe Filters provide excellent chemical resistance for optimum performance in filtering aggressive solutions.
Fisher Scientific™ Basix™ Syringe Filters, PVDF, Sterile
Attach to the end of syringes and filter contents as it is being forced out. Fisher Scientific™ Basix™ Syringe Filters, PVDF, Sterile offers high sample throughput.
Fisherbrand™ Syringe Filters - Sterile Available on GSA/VA Contract for Federal Government customers only.
Low hold-up volume helps you to recover virtually all of your sample after filtering
Corning™ SFCA Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
High burst strength for assured performance
Fisher Scientific™ Basix™ Syringe Filters, PES, Sterile
Attach to the end of syringes and filter contents as it is being forced out. Fisher Scientific™ Basix™ Syringe Filters, PES, Sterile offers high sample throughput.
Fisher Scientific™ Basix™ Syringe Filters, Nylon, Sterile
Attach to the end of syringes and filter contents as it is being forced out. Fisher Scientific™ Basix™ Syringe Filters, Nylon, Sterile offers high sample throughput.
Corning™ Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Manufactured with PTFE for the highest degree of chemical resistance. Corning™ Syringe Filters are used with syringes that serve as both the fluid reservoir and the pressure source. They are 100% integrity tested.
Corning™ RC Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
High burst strength for assured performance
EMD Millipore Millex™ Sterile Syringe Filters: PES Membrane - Clear
Ready-to-use syringe-driven units, designed for sterilizing and clarifying sterile solutions
Corning™ PES Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Manufactured from polyethersulfone (PES) for the fastest flow rates, low protein binding and low extractables. Corning™ PES Syringe Filters are ideal for tissue culture and HPLC applications.
Whatman™ Autovial™ 5mL Syringeless Filters Available on GSA/VA Contract for Federal Government customers only.
Replaces syringe-coupled devices with single filter units for one-time use. Whatman™ Autovial™ 5mL Syringeless Filters are pre-assembled filtration devices for removing particulates from samples.
GE Healthcare, Whatman Anotop 25mm HPLC Syringe Filters, Pore size 0.2μm Available on GSA/VA Contract for Federal Government customers only.
Whatman Anotop HPLC syringe filter products from GE Healthcare’s Life Sciences business are specifically designed for effective UHPLC and HPLC sample preparation.
Whatman™ Anotop™ 10 Plus Sterile Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Hold-up volume: <30μL (10mm size), <200μL (25mm)
GE Healthcare, Whatman Anotop 10mm HPLC Syringe Filters, Pore size 0.2μm Available on GSA/VA Contract for Federal Government customers only.
Whatman Anotop HPLC syringe filter products from GE Healthcare’s Life Sciences business are specifically designed for effective UHPLC and HPLC sample preparation.
Whatman™ 30mm Puradisc™ FP Sterile Cellulose Acetate Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Exhibit very low protein binding, so they are suitable for protein recovery applications. Whatman™ 30mm Puradisc™ FP Sterile Cellulose Acetate Syringe Filters are hydrophilic, making them functional with aqueous and alcoholic media despite having limited solvent resistance.
Whatman™ Autovial™ Syringeless Filters: 12mL Capacity Available on GSA/VA Contract for Federal Government customers only.
Faster sample handling, up to 12mL in volume
Whatman™ Anotop™ 10 Sterile Syringe Filters
Hold-up volume: <30μL (10mm size), <200μL (25mm)
Whatman™ Puradisc™ 4mm Sterile Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Designed for aqueous and organic solutions containing high debris levels. Whatman™ Puradisc DpPP Syringe Filters: Nonsterile tolerates high heat and mechanical stress.
Whatman™ GD/X Sterile Glass Micro Fiber (GMF) Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Processes three to seven times more sample volume than filters without prefilter. Whatman™ GD/X Sterile Glass Micro Fiber (GMF) Syringe Filters are specifically designed for filtration of viscous or otherwise hard-to-filter samples with high solids content.
GE Healthcare, Whatman Puradisc™ FP 13 Polypropylene Syringe Filters, 0.2μm, sterile Available on GSA/VA Contract for Federal Government customers only.
Whatman Puradisc syringe filters from GE Healthcare’s Life Sciences business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.
Whatman™ Anotop™ 25 Plus Sterile Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Remove virtually 100% of particulates larger than pore size
Whatman™ GD/X PVDF Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Processes three to seven times more sample volume than filters without prefilter. Whatman™ GD/X PVDF Syringe Filters are specifically designed for filtration of viscous or otherwise hard-to-filter samples with high solids content.
Whatman™ Anotop™ 10 Sterile Syringe Filters
Hold-up volume: <30μL (10mm size), <200μL (25mm)
  spinner