Digital Burets
BrandTech™ BRAND™ Titrette™ Digital Bottle-Top Burets Available on GSA/VA Contract for Federal Government customers only.
Provides accuracy and precision within the tolerances of Class A glass burettes. BrandTech™ BRAND™ Titrette™ Digital Bottle-top Burets are suitable for most titrants in concentrations up to 1 mol/L. Ideal for general chemistry, wastewater treatment applications, food/beverage analysis, industrial titrations, environmental fieldwork, and more.
Sartorius™ Biohit™ Biotrate Digital Burets
Accurate, precise and convenient bottle-top titration
Fisherbrand™ Digital Burets
Delivers accurate, precise and convenient bottle top titration
Jencons™ Digitrate™ Electronic Burets
Perform accurate volumetric titrations in the lab and field
  spinner