Gas Burners
Simple Bunsen Burner Products for Science Education
Choice of LPG or natural gas burner.
Fisher Scientific™ Adjustable-Flame Burners
Ideal for very slow heating or quick boiling
Fisher Scientific™ accuFlame™ Natural Gas Burner
Stainless-steel grid top for better flame adjustment
EISCO Micro Bunsen Burner with Gas Flow Adjustment Products for Science Education
Eisco micro bunsen burner with adjustable orifice and flame stabilizing burner tip can be use with mixed and natural gas with a heat value of 800 to 1200 BTU.
EISCO Burner Bunsen Meker: High Temperature Products for Science Education
Ideal for lab work requiring a hotter flame than that attainable with a bunsen burner.
EISCO Bunsen Burners, Micro Products for Science Education
Allows air ratio adjustments.
EISCO Bunsen Burners, Basic Products for Science Education
Prevents easy tipping, even with hoses connected.
EISCO Tirrill Bunsen Burner with Gas Control Products for Science Education
Tirrill burners allow greater flexibility in the adjustment of the air-gas mixture
EISCO Simple Bunsen Burner, LPG Products for Science Education
Simple, inexpensive gas burner is suitable for general lab.
EISCO Bunsen Burners with Flame Stabilizer Products for Science Education
Expanded base prevents easy tipping, even with hoses connected.
EISCO Bunsen Burners, Tirrill Products for Science Education
Features wing nut on bottom of tube for gas adjustment and tube screws for fine adjustment of air to fuel ratio.
Fisher Scientific™ Standard Grid-Top Adjustable-Flame Burners
Separate models for artificial and LP gases
Fisher Scientific™ Economy Burners
Nonferrous, rustproof construction
EISCO Micro Bunsen Burner Products for Science Education
Premium burner that is ideal for labs and classrooms
Fisher Scientific™ Small-Size Burners
For natural, artificial, and LP gases
EISCO Bunsen Burner With Flame Stabilizer Products for Science Education
Quality lab gas burner mounted on a die casted zinc alloy base and with a brass flame stabilizer.
EISCO Bunsen Burners with Flame Stabilizer and Gas Adjustment Products for Science Education
Allows both air to fuel ratio adjustments and throttle control via gas adjustment needle valve.
EISCO Basic Bunsen Burner, Natural Gas with Flame Stabilizer Products for Science Education
Basic natural gas burner with flame stabilizer is ideal for lab work
EISCO Bunsen Burners, Meker Products for Science Education
Designed to produce an extremely hot and stable flame.
Butane-Powered Micro Burners Products for Science Education
Provide safe, portable flame applications in a variety of settings.
Self-Contained LPG Burner Accessories: Meker Adapter Products for Science Education
Accessory for self-contained LPG burner.
Self-Contained LPG Burner Products for Science Education
Handy where natural gas is not available.
Meker-Type Burners Products for Science Education
Provide extremely hot flame.
Supertek Scientific Meker Burners Products for Science Education
For use with natural gas.
Argos FireStar™ ST Bunsen Burners
Provides economical solution to Bunsen burner safety with footswitch or on/off button for easy ignition and shut off
Straight-Tube Bunsen Burners Products for Science Education
Noiseless burners allow separate control of both flame temperature and flame height.
Butane Micro Burner Accessories: Butane Fuel Products for Science Education
Butane fuel for butane micro burner.
Humboldt Meker-Type Burner
Provides extremely hot flame
Butane Micro Burner Accessories: Tripod Stand Products for Science Education
Tripod stand for butane micro burner.
  spinner