FIS: Metal Crucibles
Fisherbrand™ Nickel Crucibles
Withstand temperatures up to 800°C (1472°F)
Fisherbrand™ Zirconium Crucibles
Withstand about 100 two- to five-minute fusions between 460°C (860°F) and 900°C (1652°F)
Fisherbrand™ Steel Crucibles
Withstand temperatures up to 500°C (932°F)
Fisherbrand™ Nickel Chromium Crucible Cover
For Nickel Chromium Crucible
Fisherbrand™ Nickel Chromium Crucible
Withstands temperatures up to 1000°C (1832°F)
  spinner