FIS: Porcelain Crucibles
Fisherbrand™ Crucibles Products for Science Education
Easy to clean smooth, glazed surface.
Fisherbrand™ Crucible Covers Products for Science Education
For use with Fisherbrand Crucibles.
PYREX™ High-Form Glass Filtering Crucibles Products for Science Education
Work with higher temperatures, such as in an electric oven.
Fisherbrand™ Porcelain Flat Capsule Crucibles
For use in laboratories of educational institutions, scientific development and industrial research, chemical, fertilizer, petro-chemical establishments
Fisherbrand™ Wide-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
Fisherbrand™ Porcelain Bitumen Filtering Crucible
Filtration device for general laboratory use
Fisherbrand™ Porcelain 25mL Crucibles
For laboratory and industrial purpose
Kimble™ KIMAX™ Gooch Filtering Crucibles Available on GSA/VA Contract for Federal Government customers only.
Dry precipitates to constant weight at 110°C (230°F)
Fisherbrand™ High-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
Fisherbrand™ Porcelain Crucible Covers
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
Fisherbrand™ Porcelain Gooch Filtering Crucibles
For total suspended solids determinations
DURAN™ Filter Crucible
Thermal shock and chemical resistant
PYREX™ Gooch Type Filtering Crucibles Available on GSA/VA Contract for Federal Government customers only.
For drying precipitates to constant weight at 110°C (230°F)
Fisherbrand™ Porcelain Capsule-Form Crucibles
Glazed inside and out except for rim
Fisherbrand™ Porcelain Rose Crucible
For use in very high temperatures
Fisherbrand™ Porcelain Casserole Crucibles
For use in laboratory studies
Fisherbrand™ Porcelain 90mL Dish Crucible
For laboratory and industrial purpose
GSC International Low Form Crucibles with Lid Products for Science Education
Uniform density provides stable mechanical strength and low permeability.
Walters Crucible Holder Products for Science Education
High-quality holder.
GSC International Low Form Porcelain Crucibles with Lids Products for Science Education
Constructed with uniform thickness and density for stable mechanical strength. GSC International Low Form Porcelain Crucibles with Lids can withstand temperatures of up to 1050°C.
Plasticoid Crucible Holders
Holders for glass crucibles
Fused-Quartz Crucibles
Withstand temperatures up to 1250°C (2282°F)
GSC International High Form Porcelain Crucibles with Lids Products for Science Education
Constructed with uniform thickness and density for stable mechanical strength. GSC International High Form Porcelain Crucibles with Lids can withstand temperatures of up to 1050°C.
Plasticoid Walter Crucible Holder
Rubber with removable glass stem ground at 45° angle
Fused Quartz Crucible Covers
For 08-072 Series Crucibles
CoorsTek™ Porcelain Gooch Filtering Crucibles
Glazed inside and out except for outside bottom surface
CoorsTek™ Porcelain Coal Moisture Testing Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
CoorsTek™ Wide-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
  spinner