Desiccator Accessories
Fisherbrand™ Humidity Sponge™ Products for Science Education
Turns any container into a desiccator.
Thermo Scientific™ Nalgene™ Ceramic-Metal Desiccator Plate Products for Science Education
With numbered quadrants for easy location of crucibles and other containers
Thermo Scientific™ Nalgene™ 230mm Ceramic Metal Desiccator Plate
Ensure greater resistance to thermal shock than porcelain using this guaranteed unbreakable and corrosion-resistant ceramic metal desiccator plate.
Bel-Art™ SP Scienceware™ Reusable Desiccant Cartridges Available on GSA/VA Contract for Federal Government customers only.
Convenient, easy-to-handle cartridges with Natrasorb™ TI indicating silica gel beads. Bel Art SP Scienceware™ Reusable Desiccant Cartridges can also be used with other plastic desiccators of similar size.
Bel-Art™ SP Scienceware™ Secador™ Desiccator Replacement Shelves Available on GSA/VA Contract for Federal Government customers only.
Convert 4.0 units from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
PYREX™ Desiccator Replacement Covers Available on GSA/VA Contract for Federal Government customers only.
Various covers dependent on desiccator size and style
PYREX™ Desiccator Replacement Bowls Available on GSA/VA Contract for Federal Government customers only.
Strong, easily cleaned, borosilicate glass bowls
Bel-Art™ SP Scienceware™ Reuseable/Disposable Desiccant Cartridges Available on GSA/VA Contract for Federal Government customers only.
Use anywhere dessiccation is needed, including analytical balance cases and precision laboratory instruments. SP Scienceware™ Reuseable/Disposable Desiccant Cartridges fit “Space Saver” desiccators, in addition to other plastic desiccators of similar size.
Fisherbrand™ Reusable Desiccant Canister Available on GSA/VA Contract for Federal Government customers only.
Ideal for desiccator cabinets and storage areas subject to moisture damage
PYREX™ 24/29 Standard Taper Desiccator Stopcock with Hose Connector Available on GSA/VA Contract for Federal Government customers only.
Replacement 24/29 standard taper desiccator stopcock with a tooled #10 hose connection
Bel-Art™ SP Scienceware™ Vacuum Pressure Indicator Available on GSA/VA Contract for Federal Government customers only.
Indicates if desiccator is under vacuum.
Bel-Art™ SP Scienceware™ Secador™ Auto-Desiccator Module Desiccant Replacement Beads
Color-indicating silica gel beads for use in refilling the Secador auto-desiccator module
Bel-Art™ SP Scienceware™ Desi-Can™ Reusable Desiccant Canister
For use in closets, containers, tool boxes, camera cases and other storage areas to prevent rust and moisture, these canisters contain silica gel to effectively absorb moisture through perforations in the aluminum case
Fisherbrand™ Replacement Covers for Nonvacuum Desiccator
For 08-595 Series and 08-615 Series desiccators
Bel-Art™ SP Scienceware™ Under-Shelf Mounting Kit Available on GSA/VA Contract for Federal Government customers only.
For use with Secador Mini Desiccator Cabinet.
Drierite™ Desiccant in Cylinder
Efficient drying unit for analytical balances
W.A. Hammond Drierite™ Gas Purifier
Designed for gas chromatography and other applications requiring dry, pure gas. W.A. Hammond Drierite™ Gas Purifier dries, purifies, and filters gases used for chromatography and spectrometry.
W.A. Hammond Drierite™ Gas Drying Unit with Hose Barbs
Dries air and gases saturated with moisture to dew point. W A Hammond Drierite™Gas Drying Unit with Hose Barbs can be regenerated by removing from column and heating granules on tray in single layer
  spinner