Stir Bars
Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Sink Strainers Products for Science Education
Prevents Spinbar Magnetic Stirring Bars from going down the sink drain.
Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Stirring Bar Restrainers Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Fits ergonomically in one hand and when placed alongside the outer wall of a mixing vessel, it draws in and secures the stirring bar in situ.
Bel-Art™ SP Scienceware™ Spinplus™ Magnetic Stirring Bars Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Spinplus™ magnetic stirring bars add speed and efficiency to mixing operations
Bel-Art™ SP Scienceware™ Spinbar™ Capsule Magnetic Stirring Bars Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
1.18 x 0.31 in. polygon magnetic stirring bar spins freely inside a stationary capsule, excellent for pH adjustments of small volumes.
Bel-Art™ SP Scienceware™ Octagon Spinbar™ Magnetic Stir Bars Products for Science Education
Octagon magnetic stirring bars with integral pivot ring
Minimixer Stir Bar Products for Science Education
For use with Minimixers
Thermo Scientific™ Nalgene™ StarHead™ Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Greater mixing action at bottom, side, and top of container, even at low stirring speeds
Thermo Scientific™ Nalgene™ Polypropylene Stirring Rod Available on GSA/VA Contract for Federal Government customers only.
Combine functions of a stirring rod and rubber policeman with these polypropylene stirring rods, featuring a flat paddle at one end and a triangular paddle at the other end.
Fisherbrand™ Octagon Magnetic Stir Bars Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape
Thermo Scientific™ Komet Stir Bars
Generate over three times the torque of conventional stirring bars with Thermo Scientific™ Komet Stir Bars, significantly enhancing stirrer performance and versatility.
Thermo Scientific™ Nalgene™ Suspended Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Use Nalgene Floating Magnetic Stir Bars for constant low-speed stirring action in tissue culture applications.
Fisherbrand™ Octagon Magnetic Stir Bar Products for Science Education
Octagonal magnetic stirring bars with integral pivot ring are the most commonly used shape
Fisherbrand™ PTFE Disposable Stir Bars Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
The multifaceted magnetic stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars
Bel-Art™ SP Scienceware™ Octagon Spinbar™ Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Octagon magnetic stirring bars with integral pivot ring
Bel-Art™ SP Scienceware™ Spinfin™ Magnetic Stirring Bars
Agitate liquids in round bottom vessels like test tubes, cylinders and flasks
Fisherbrand™ TFE Starburst Stirring Heads Individual Heads Available on GSA/VA Contract for Federal Government customers only.
Provide better mixing at bottom of vessel with less splashing
Fisherbrand™ Octagonal Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape
Bel-Art™ SP Scienceware™ Flea Micro Spinbar™ Magnetic Stirring Bars Available on GSA/VA Contract for Federal Government customers only.
Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
Fisherbrand™ Color-Coded Dumbbell-Style Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Provide fast mixing action, strong turbulence, greater stability, and less drifting at higher speeds
Bel-Art™ SP Scienceware™ Spinbar™ Round Magnetic Stirring Bars Available on GSA/VA Contract for Federal Government customers only.
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics
Corning™ PTFE-Coated Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.
For use in Corning Hotplate Stirrers
Bel-Art™ SP Scienceware™ Spinbar™ Octagon Rare Earth Magnetic Stir Bar
Rare Earth/Samarium Cobalt magnet provides strong coupling with drive magnets to reduce frequency of spinout
Bel-Art™ SP Scienceware™ Polygon Spinbar™ Magnetic Stirring Bars Available on GSA/VA Contract for Federal Government customers only.
Center molded pivot ring minimizes friction and chattering
Fisherbrand™ Magnetic Stir Bar Six Packs Available on GSA/VA Contract for Federal Government customers only.
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape
Bel-Art™ SP Scienceware™ Spinstar™ Magnetic Stirring Bars
Star-shape creates deep vortex at relatively slow speed extending to the bottom of vessel
Fisherbrand™ Round Stir Bars with Removable Pivot Ring Available on GSA/VA Contract for Federal Government customers only.
Stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars
  spinner