Magnetic Stir Bar Retrievers
Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Sink Strainers Products for Science Education
Prevents Spinbar Magnetic Stirring Bars from going down the sink drain.
Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Flexible Retrievers Available on GSA/VA Contract for Federal Government customers only.
For retrieving magnetic stirring bars from containers or nonmagnetic sink drains and traps
Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Stirring Bar Restrainers Products for Science Education
Fits ergonomically in one hand. Bel Art™ SP Scienceware™ Spinbar™ Magnetic Stirring Bar Restrainers retains and secures the stirring bar in position.
Fisherbrand™ PTFE Stirring Bar Retriever
13 in. long, for deep vessels
Bel-Art™ SP Scienceware™ Magnetic Stirring Bar Arrester Available on GSA/VA Contract for Federal Government customers only.
Holds stirring bar in place when pouring from a beaker or flask by placing the arrester against the bottom of the container
Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Stirring Bar Restrainers Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Fits ergonomically in one hand and when placed alongside the outer wall of a mixing vessel, it draws in and secures the stirring bar in situ.
Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Sink Strainer
Resistant to chemicals sink strainer has four strong magnets in each corner to attract and hold Spinbar magnetic stirring bars. Bel Art™ SP Scienceware™ Spinbar™ Magnetic Sink Strainer is a simple solution to prevent your Spinbar® Magnetic Stirring Bars from going down the sink drain.
Fisherbrand™ Stir Bar Retrievers Available on GSA/VA Contract for Federal Government customers only.
Pull stirring bars out of containers easily with a magnet
  spinner