Magnetic Stir Bars
Bel-Art™ SP Scienceware™ Spinbar™ Capsule Magnetic Stirring Bars Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
1.18 x 0.31 in. polygon magnetic stirring bar spins freely inside a stationary capsule, excellent for pH adjustments of small volumes.
Bel-Art™ SP Scienceware™ Octagon Spinbar™ Magnetic Stir Bars Products for Science Education
Octagon magnetic stirring bars with integral pivot ring
Bel-Art™ SP Scienceware™ Spinplus™ Magnetic Stirring Bars Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Spinplus™ magnetic stirring bars add speed and efficiency to mixing operations
Minimixer Stir Bar Products for Science Education
For use with Minimixers
Thermo Scientific™ Nalgene™ Suspended Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Use Nalgene Floating Magnetic Stir Bars for constant low-speed stirring action in tissue culture applications.
Thermo Scientific™ Komet Stir Bars
Generate over three times the torque of conventional stirring bars with Thermo Scientific™ Komet Stir Bars, significantly enhancing stirrer performance and versatility.
Fisherbrand™ Octagon Magnetic Stir Bars Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape
Fisherbrand™ PTFE Disposable Stir Bars Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
The multifaceted magnetic stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars
Thermo Scientific™ Nalgene™ StarHead™ Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Greater mixing action at bottom, side, and top of container, even at low stirring speeds
Thermo Scientific™ Nalgene™ Polypropylene Stirring Rod Available on GSA/VA Contract for Federal Government customers only.
Combine functions of a stirring rod and rubber policeman with these polypropylene stirring rods, featuring a flat paddle at one end and a triangular paddle at the other end.
Fisherbrand™ Octagon Magnetic Stir Bar Products for Science Education
Octagonal magnetic stirring bars with integral pivot ring are the most commonly used shape
Bel-Art™ SP Scienceware™ Spinbar™ Capsule Magnetic Stirring Bar
Capsule prevents stirring bar from spinning off center and damaging pH probes and gels.
Bel-Art™ SP Scienceware™ Octagon Spinbar™ Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Octagon magnetic stirring bars with integral pivot ring
Bel-Art™ SP Scienceware™ Spinbar™ Round Magnetic Stirring Bars Available on GSA/VA Contract for Federal Government customers only.
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics
Fisherbrand™ Cross-Type Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Agitate liquids in round-bottom test tubes and flasks, where other stir bars cannot be used
Fisherbrand™ Polygon Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars
SP SCIENCEWARE BEL ART SP Scienceware™ Polygon Spinbar™ Magnetic Stirring Bars without Pivot Ring Available on GSA/VA Contract for Federal Government customers only.
Adds turbulence relative to similar smooth size cylindrical bars. SP Scienceware™ Polygon Spinbar™ Magnetic Stirring Bars without Pivot Ring are manufactured in a registered ISO 9001:2008 facility.
Bel-Art™ SP Scienceware™ Flea Micro Spinbar™ Magnetic Stirring Bars Available on GSA/VA Contract for Federal Government customers only.
Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
Fisherbrand™ Magnetic Stir Bars with Vanes Available on GSA/VA Contract for Federal Government customers only.
Ensure thorough sample mixing in test tubes, micro vials and conical-bottom centrifuge tubes
Fisherbrand™ Magnetic Micro Stirring Bars Available on GSA/VA Contract for Federal Government customers only.
Micro magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes and gradient makers
Fisherbrand™ PTFE Stirring Rods Available on GSA/VA Contract for Federal Government customers only.
Pure PTFE rods are available either solid or with a milled steel core
Kimble™ Konte™s Round PTFE Micro Magnetic Stir Bar
PTFE-covered round magnetic stirring bar
Bel-Art™ SP Scienceware™ Double Spinfin™ Magnetic Stirring Bars
To agitate liquids in round bottom vessels where other stirring bars cannot be used.
Bel-Art™ SP Scienceware™ Spinwedge™ Magnetic Stirring Bars Available on GSA/VA Contract for Federal Government customers only.
Provide strong tubulence at fairly low speeds and are well suited for churning sediment or dissolving salts
Fisherbrand™ High-Energy Octagon Stir Bar Available on GSA/VA Contract for Federal Government customers only.
Composition and octagonal shape produce exceptional power suitable for viscous solutions and high-speed stirring
Fisherbrand™ Magnetic Stir Bar Six Packs Available on GSA/VA Contract for Federal Government customers only.
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape
Bel-Art™ SP Scienceware™ Pyrex™ Glass Round Spinbar™ Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.
Useful for high temperature applications up to 274°C (525°F)
  spinner