Non-Electronic Hygrometers
Fisher Scientific™ Mason's Hygrometer Products for Science Education
Use the dual thermometers to calculate relative humidity. Fisher Scientific Mason's Hygrometer has one dry thermometer and one wet thermometer which provide temperature readings for determining the relative humidity.
Fisher Scientific™ Traceable™ Humidity-On-A-Card™ Humidity Monitor Products for Science Education
Records current humidity and maximum humidity attained using color-change reactions.
Fisherbrand™ Traceable™ Humidity-On-A-Card™ Humidity Monitor
Ideal for use with bottles, chemicals, plastic bags, electronics, desiccators and storage containers
Wall-Mount Thermohygrometers Products for Science Education
Easy-to-read dial indicators for both humidity and temperature.
H-B Instrument™ Durac™ Mason's Hygrometer
Designed with wet and dry bulb thermometers that displays relative humidity and dew point
H-B Instrument™ Durac™ Thermometer-Hygrometers
For measurement of temperature and humidity
Fisher Scientific™ Dial Hygrometer/Thermometer
Ideal for use in labs, plant areas, and storage facilities
  spinner