Moisture Meters
Fisherbrand™ Precision Specific Gravity Hydrometers, 150mm Long Available on GSA/VA Contract for Federal Government customers only.
Requires 140mL liquid volume for submersion
Fisherbrand™ PVC Hydrometer Carrying Cases
Rugged hydrometer case protects glass hydrometers during storage and transport
Fisher Scientific™ Baume Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Baume “heavy” scale for liquids heavier than water
Fisher Scientific™ Specific Gravity Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Double scales can be read without turning hydrometer
Fisher Science Education™ Wall-Mountable Masons Hygrometer Products for Science Education
Measures relative humidity and dew point using wet and dry bulb thermometers.
Fisher Scientific™ Twaddell Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Use with liquids heavier than water such as milk, sulfuric acid, tanning solutions
Fisherbrand™ Precision Specific Gravity Hydrometers Products for Science Education
This set includes a thermometer with 0° to 220°F range and eight precision hydrometers.
Fisher Scientific™ Specific Gravity/Baume Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Calibrated according to ASTM™ E 126 specifications
Fisher Scientific™ API ASTM™ Hydrometers, Plain Form Available on GSA/VA Contract for Federal Government customers only.
API hydrometers are available in regular and ASTM™ recommended forms
Fisherbrand™ Brix Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Use to measure the concentration of sugar solutions in refineries
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water Available on GSA/VA Contract for Federal Government customers only.
Accuracy meets or exceeds ASTM™ E 100 specifications
Fisher Scientific™ Traceable™ Digital Humidity/Temperature/Dew Point Meter
Convenient triple memories recall minimum/maximum readouts for relative humidity, dew point and temperature.
Fisher Scientific™ Memory Humidity/ Temperature Pen Products for Science Education
Memory displays highest and lowest humidity/temperature reading achieved overnight or during any time period. Fisher Scientific Memory Humidity/Temperature Pen continuously and simultaneously displays both relative humidity and °F/°C with dual recall memories.
Fisherbrand™ API Petroleum Hydrometers Temperature Range 20° to 130°F, Plain Form Available on GSA/VA Contract for Federal Government customers only.
API hydrometers are available in regular and ASTM™ recommended forms
Fisher Scientific™ Temperature/Humidity Meter Products for Science Education
Capture, display, and store readings on your computer. Fisher Scientific Temperature/Humidity Meter is used to monitor temperature and humidity outside your class, or in a controlled environment.
Fisher Scientific™ Digital Relative Humidity/Temperature Meter Products for Science Education
Simultaneously displays relative humidity and temperature once per second.
Fisher Science Education™ Thermometer / Clock / Humidity Monitor Products for Science Education
Ambient unit displays temperature, humidity and time. Fisher Science Education Thermometer/Clock/Humidity Monitor have internal sensors and can be used in hoods, storerooms, incubators and environmental cabinets.
Fisher Scientific™ Traceable™ Humidity Controller
Ideal for switching your electrical equipment on or off at any 1% increment from 30 to 90% relative humidity.
Fisher Scientific™ Midget Urinometer Available on GSA/VA Contract for Federal Government customers only.
Accuracy guaranteed to ±0.002
Fisherbrand™ Soil Analysis ASTM Hydrometers
These hydrometers are used to determine the specific gravity of soil in the range of 0.995 to 1.038 or -5 to 60g/L (using a weight by volume scale)
Fisherbrand™ Traceable™ Memory Hygrometer/Thermometer
Simultaneously displays both temperature and relative humidity on dual 1.12 in. high LCD.
Kimble-Chase Precision Specific Gravity Hydrometer Set
Set of eight Precision Specific Gravity Hydrometers
  spinner