Electronic Hygrometers
H-B Instrument™ Durac™ Bluetooth Hygro-Thermometer with 30-Day Data Logging Products for Science Education
Broadcast readings to any apple or android smartphone or device with Bluetooth 4.0 or better, utilizing the free THERMSmart app. H-B Instrument Durac Bluetooth Hygro-Thermometer records hourly data for current seven days and then temperatures and humidity are recorded for next 23 days.
Sper Scientific™ RH/Temperature SD Card Logger Products for Science Education
Reads relative humidity and ambient temperature dew point and wet bulb, and functions as a type K or J thermometer.
Sper Scientific™ Mini Environmental Quality Meter Products for Science Education
Ideal for environmental testing anywhere.
Fisher Scientific™ Digital Relative Humidity/Temperature Meter Products for Science Education
Simultaneously displays relative humidity and temperature once per second.
Fisher Scientific™ Traceable™ Digital Humidity/Temperature/Dew Point Meter
Convenient triple memories recall minimum/maximum readouts for relative humidity, dew point and temperature.
Fisher Science Education™ Wall-Mountable Masons Hygrometer Products for Science Education
Measures relative humidity and dew point using wet and dry bulb thermometers.
Fisher Scientific™ Memory Humidity/ Temperature Pen Products for Science Education
Memory displays highest and lowest humidity/temperature reading achieved overnight or during any time period. Fisher Scientific Memory Humidity/Temperature Pen continuously and simultaneously displays both relative humidity and °F/°C with dual recall memories.
Fisher Science Education™ Thermometer / Clock / Humidity Monitor Products for Science Education
Ambient unit displays temperature, humidity and time. Fisher Science Education Thermometer/Clock/Humidity Monitor have internal sensors and can be used in hoods, storerooms, incubators and environmental cabinets.
Fisher Scientific™ Temperature/Humidity Meter Products for Science Education
Capture, display, and store readings on your computer. Fisher Scientific Temperature/Humidity Meter is used to monitor temperature and humidity outside your class, or in a controlled environment.
Fisher Scientific™ Traceable™ Jumbo Thermo-Humidity Meter
Fast-response, solid-state sensors show changes in relative humidity, ambient temperature, and (with probe) external temperature in under 30 seconds
Fisher Scientific™ Traceable™ Humidity/Temperature Meter/Recorder
Ideal for monitoring conditions in chambers, cleanrooms, food processing facilities, scientific laboratories, and other humidity-sensitive areas
Fisher Scientific™ Traceable™ Temperature/Humidity Meter
Simultaneously displays humidity, dew point and temperature
Fisher Scientific™ Traceable™ Relative Humidity/Temperature Meters
Monitor air conditions in environmental chambers, chemical storage areas, cleanrooms, incubators and stockrooms
Fisher Scientific™ Traceable™ Clock/Thermometer/Humidity Meter
Battery-operated, wall-mounted unit has 9.75 in. dial
Fisher Scientific™ Traceable™ Radio-Signal Hygrometer/Thermometer Products for Science Education
Monitor humidity and temperature from up to 100 ft. away—even outdoors. Remote sensor displays current temperature and humidity.
Fisher Scientific™ Traceable™ Radio-Signal Hygrometer/Thermometer
Monitors humidity and temperature from up to 100 ft. away—even outdoors. Remote sensor displays current temperature and humidity
Fisher Scientific™ Traceable™ Digital Humidity/Temperature Meter Available on GSA/VA Contract for Federal Government customers only.
Displays relative humidity from 10.0 to 95.0% with resolution of 0.1% and accuracy of 1.5%
Fisher Scientific™ Certified Traceable™ Digital Hygrometer/Thermometers Available on GSA/VA Contract for Federal Government customers only.
Meet critical experimental requirements, perform routine measurements, and fulfill all quality control needs.
Fisher Scientific™ Traceable™ Humidity Meter
Retains highest and lowest readings recorded over selected period
Fisher Scientific™ Traceable™ Remote Alarm RH/Temperature Monitor
Triple-display and remote control unit offers safety and convenience
Fisher Scientific™ Traceable™ Jumbo Thermo-Humidity Meter Products for Science Education
Fast-response, solid-state sensors show changes in relative humidity, ambient temperature, and (with probe) external temperature in under 30 seconds.
Fisherbrand™ Traceable™ Memory Hygrometer/Thermometer
Simultaneously displays both temperature and relative humidity on dual 1.12 in. high LCD.
Fisher Scientific™ Remote Sensor Module for Traceable™ Radio-Signal Remote Hygrometer/Thermometer
Remote sensor module for Traceable radio-signal remote hygrometer/thermometer
Fisher Scientific™ Traceable™ Humidity Controller
Ideal for switching your electrical equipment on or off at any 1% increment from 30 to 90% relative humidity.
Fisher Scientific™ Certified Traceable™ Digital Hygrometer/Thermometer
Designed for cleanrooms, hoods, process control, and air monitoring.
Fisher Scientific™ Traceable™ Digital Hygrometer/Thermometer
Hang on the wall or use included plastic stand; housed in rugged ABS case
  spinner