Hydrometers
H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm&superscript_3;) Plain Form Hydrometers: for Liquids Lighter Than Water Products for Science Education
Available in a variety of models measuring specific gravity/relative density (g/cm&superscript_3;) of liquids lighter than water in various ranges from 0.690 to 3.000.
H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm&superscript_3;) Plain Form Hydrometers: for Liquids Heavier Than Water Products for Science Education
Available in a variety of models measuring specific gravity/relative density (g/cm&superscript_3;) of liquids heavier than water in various ranges from 0.690 to 3.000.
Durac™ Soil Analysis ASTM Hydrometers Products for Science Education
For determining the specific gravity of soil.
H-B Instrument™ Durac™ Salt Brine Plain Form Hydrometers Products for Science Education
Choose from two plain form models with ranges from 0 to 100% or 0 to 26.5% to measure sodium chloride saturation of salt brines
H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm3) Hydrometer Set with EnviroSafe Thermometer Products for Science Education
Set consists of eight specific gravity/relative density plain form hydrometers and one total immersion thermometer to reference liquid temperature for accurate hydrometer readings.
H-B Instrument™ Durac™ Safety Brix Sugar Scale Combined Form Thermo-Hydrometers Products for Science Education
These combined-form hydrometers are used to measure % sucrose by weight in various ranges from 0° to 101° Brix, and include a convenient, built-in thermometer with a temperature scale of 0/50°C.
H-B Instrument™ Durac™ Battery Hydrometer with Siphon Set Products for Science Education
Used to determine the specific gravity/relative density (g/cm3) of battery acid in the range of 1.150 to 1.300
H-B Instrument™ Durac™ API ASTM Hydrometers; Traceable to NIST
Measure the relative density of oil and petroleum products in ranges from -1° to 101° API with these hydrometers. H-B Instrument Durac™ API ASTM Hydrometers conform to American Petroleum Institute (API) and the American Society of Testing Materials (ASTM) E100 standards. Traceable to NIST.
Fisher Scientific™ Specific Gravity/Baume Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Calibrated according to ASTM™ E 126 specifications
Fisher Scientific™ API ASTM™ Hydrometers, Plain Form Available on GSA/VA Contract for Federal Government customers only.
API hydrometers are available in regular and ASTM™ recommended forms
Fisherbrand™ API Petroleum Hydrometers Temperature Range 20° to 130°F, Plain Form Available on GSA/VA Contract for Federal Government customers only.
API hydrometers are available in regular and ASTM™ recommended forms
Fisherbrand™ Brix Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Use to measure the concentration of sugar solutions in refineries
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water Available on GSA/VA Contract for Federal Government customers only.
Accuracy meets or exceeds ASTM™ E 100 specifications
Fisher Scientific™ Specific Gravity Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Double scales can be read without turning hydrometer
Fisherbrand™ Precision Specific Gravity Hydrometers Products for Science Education
This set includes a thermometer with 0° to 220°F range and eight precision hydrometers.
Fisherbrand™ PVC Hydrometer Carrying Cases
Rugged hydrometer case protects glass hydrometers during storage and transport
Fisherbrand™ Precision Specific Gravity Hydrometers, 150mm Long Available on GSA/VA Contract for Federal Government customers only.
Requires 140mL liquid volume for submersion
Fisher Scientific™ Baume Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Baume “heavy” scale for liquids heavier than water
Fisher Scientific™ Twaddell Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Use with liquids heavier than water such as milk, sulfuric acid, tanning solutions
Fisherbrand™ Soil Analysis ASTM Hydrometers
These hydrometers are used to determine the specific gravity of soil in the range of 0.995 to 1.038 or -5 to 60g/L (using a weight by volume scale)
Kimble-Chase Ethanol Hydrometer
With dual graduations in 0 to 100% and 0 to 200 proof scales
  spinner