Temperature Control
Fisherbrand™ Ice Buckets Products for Science Education
Ideal for use with dry ice, water ice and salt solutions.
Thermo Scientific™ Benchtop Coolers Available on GSA/VA Contract for Federal Government customers only.
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Thermo Scientific™ Mr. Frosty™ Freezing Container Available on GSA/VA Contract for Federal Government customers only.
Freeze cells in tubes from 1 to 5mL using the Thermo Scientific™ Mr. Frosty™ Container, designed to achieve a rate of cooling very close to -1°C/minute.
Thermo Scientific™ Benchtop Flasks Available on GSA/VA Contract for Federal Government customers only.
Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with Thermo Scientific™ Benchtop Flasks.
Thermo Scientific™ Dewar Flasks Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Chemical resistant and withstand a wide temperature range.
Fisherbrand™ Metal-Cased Cylindrical Dewar Flasks
Borosilicate glass with silvered double walls
Bel-Art™ SP Scienceware™ Magic Touch™ Iceware Ice Buckets Available on GSA/VA Contract for Federal Government customers only.
Lightweight ice buckets have excellent insulating properties for keeping temperature sensitive samples in an optimal environment
Fisherbrand™ Polyurethane Ice Buckets
For use with liquid nitrogen, dry ice, water ice, and salt solutions
Bel-Art™ SP Scienceware™ Cryo-Safe™ Microcentrifugre Tube and Cryogenic Vial Coolers
Holds thirty-two 0.5, 1.5 or 2.0ml microcentrifuge tubes or cryogenic vials at a temperature below -15ºC for approximately 1½ hours. Bel Art SP Scienceware™ Cryo-Safe™ Microcentrifugre Tube and Cryogenic Vial Coolers use the included insert to hold 0.5ml microcentrifuge tubes.
Fisherbrand™ Low-Form Dewar Flasks
Slightly curved bottom increases coolant capacity
Bel-Art™ SP Scienceware™ Magic Touch™ Insulated Lab Pans Available on GSA/VA Contract for Federal Government customers only.
Keep temperature sensitive samples in an optimal environment. Bel Art™ SP Scienceware™ Magic Touch™ Insulated Lab Pans are the lightest icewares on the market.
Bel-Art™ SP Scienceware™ Cryo-Safe™ Mini Quick-Freeze Microcentrifuge Tube Cooler
Portable unit rapidly and repeatedly chills samples for molecular biology
Bel-Art™ SP Scienceware™ Cryo-Safe™ Cold Box
Maintains the low temperature required for safely storing and transporting enzymes and other critical samples
Sonoco™ ThermoSafe Insulated Storage and Transport Chests Available on GSA/VA Contract for Federal Government customers only.
Ideal for dry ice and wet ice storage, short-term frozen and hot storage.
GE Healthcare MultiTemp™ IV Thermostatic Circulator Replacement Hose Clamp Available on GSA/VA Contract for Federal Government customers only.
For use with GE Healthcare MultiTemp IV Thermostatic Circulator
Heidolph™ Storm Heating/Cooling Work Station Available on GSA/VA Contract for Federal Government customers only.
Suitable thermoregulator help to provide controlled steady state heating and cooling
Labconco™ Universal Tube Bath
Maintain a constant temperature for tube incubations
Sonoco™ ThermoSafe Insulated Shipping Container Available on GSA/VA Contract for Federal Government customers only.
Polyurethane foam insulated container with plug
GE Healthcare MultiTemp™ IV Thermostatic Circulator
Specially designed for electrophoresis applications
Sonoco™ ThermoSafe Rugged Dry Ice Storage and Transport Chest
For low-temperature storage and short term high-temperature storage.
Sonoco™ ThermoSafe Nylon Carriers Available on GSA/VA Contract for Federal Government customers only.
Lightweight, nylon clad, durable units ideal for on-site collection, storage and transportation of temperature-sensitive specimens, medications, blood products, medical devices, and organs
Sonoco™ ThermoSafe Chest-Style Containers Available on GSA/VA Contract for Federal Government customers only.
Maintains product quality for your temperature sensitive products and critical low-temperatures for laboratory specific materials
Heidolph™ Breeze Heating/Cooling Work Station and Stand Available on GSA/VA Contract for Federal Government customers only.
Lightweight and compact equipment for applications requiring control by solution temperature
Therapak™ Coleman™ Wheeled Coolers
With rustproof, leak resistant no-tilt drain
  spinner