Rubber Tubing
GSC International Latex Tubing Products for Science Education
Latex tubing with 1.6mm thick wall.
Supertek Scientific Rubber Tubing Products for Science Education
Red colored 10m long tubing for general laboratory use.
Thermo Scientific™ Nalgene™ 980 Braided Clear PVC Tubing Available on GSA/VA Contract for Federal Government customers only.
Easily bend this braided PVC tubing into place and allow tighter compression of crimp fittings for a secure fit. It is designed for high-pressure applications.
Thermo Scientific™ Nalgene™ 180 Clear Plastic PVC Tubing Available on GSA/VA Contract for Federal Government customers only.
Perform a range of non-medical applications with Thermo Scientific™ Nalgene™ 180 Clear Plastic PVC Tubing, which resists hardening and discoloration.
Thermo Scientific™ Nalgene™ 50 Platinum-Cured Silicone Tubing Available on GSA/VA Contract for Federal Government customers only.
Minimize absorption of proteins and food products. A low-binding surface makes this flexible, durable tubing ideal for a variety of pump and transfer applications.
Thermo Scientific™ Nalgene™ 870 Tubing Available on GSA/VA Contract for Federal Government customers only.
Utilize translucent Thermo Scientific™ Nalgene™ 870 Tubing for transferring aggressive and corrosive chemical fluids.
Thermo Scientific™ Nalgene™ 180 Clear Plastic PVC Metric Tubing Available on GSA/VA Contract for Federal Government customers only.
Establish a leak-tight fit with metric fittings by using Nalgene 180 PVC Metric Tubing, manufactured from the highest-grade resins and special plasticizers.
Thermo Scientific™ Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
Ensure consistently reliable and accurate fluid transfers. This durable high-purity tubing is ideal for pharmaceutical, laboratory and bioprocess manufacturing.
Thermo Scientific™ Nalgene™ 50 Platinum-Cured Silicone Tubing for Peristaltic Pumps
Minimize absorption of proteins and food products with this flexible, durable high-purity translucent tubing, ideal for pharmaceutical, laboratory, bioprocess manufacturing and food and beverage applications.
Thermo Scientific™ Nalgene™ 280 Polyurethane Tubing Available on GSA/VA Contract for Federal Government customers only.
Perform high-purity work with this ester-grade tubing, which exhibits excellent abrasion and flex-fatigue resistance, as well as low-temperature resilience.
Thermo Scientific™ Nalgene™ 890 Tubing Available on GSA/VA Contract for Federal Government customers only.
Conduct demanding lab applications with Thermo Scientific™ Nalgene™ 890 Tubing, which resists most known chemicals at room temperature and offers high temperature resistance.
Fisherbrand™ Clear Vinyl Tubing Products for Science Education
Nontoxic, chemical-resistant, colorless and autoclavable.
Thermo Scientific™ Nalgene™ 65 Braided Platinum-Cured Silicone Tubing Available on GSA/VA Contract for Federal Government customers only.
Use this durable silicone tubing for a variety of pump and transfer applications, including pharmaceutical, laboratory, bioprocess manufacturing and food and beverage applications. 
Thermo Scientific™ Nalgene™ 489 Linear LDPE Tubing Available on GSA/VA Contract for Federal Government customers only.
Easily view fluid levels with translucent Thermo Scientific™ Nalgene™ 489 Linear LDPE Tubing, economically priced and suitable for industrial use.
Thermo Scientific™ Nalgene™ 180 Clear Plastic PVC Vacuum Tubing Available on GSA/VA Contract for Federal Government customers only.
Make practical and efficient use of this Nalgene 180 vacuum tubing, thick-walled PVC which can withstand a full vacuum (80mm Hg) at room temperature.
Thermo Scientific™ Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing for Peristaltic Pumps
Ensure consistently reliable and accurate fluid transfers. This durable, high-purity tubing is ideal for pharmaceutical, laboratory and bioprocess manufacturing.
Thermo Scientific™ Nalgene™ 689 Polypropylene Tubing Available on GSA/VA Contract for Federal Government customers only.
Perform a variety of applications with the excellent chemical resistance to several acids and alkalis of Thermo Scientific™ Nalgene™ 689 Polypropylene Tubing.
Fisherbrand™ General-Purpose Rubber Tubing in 12 ft. (3.7m) Lengths
Seamless rubber tubing with nonslip surface and flexibility
Fisherbrand™ Tygon S3™ E-3603 Flexible Tubings
Formulated for resistance to flex-fatigue and abrasion
Cole Parmer™ Masterflex™ Tygon™ Fuel & Lubricant L/S™ Precision Pump Tubing
Use this Tygon™ tubing for applications involving fuels and industrial lubricants. Cole Parmer™ Masterflex™ Tygon™ Fuel & Lubricant L/S™ High Performance Precision Pump Tubing is ideal for transferring hydrocarbons, gasoline, kerosene, heating oils, cutting compounds and glycol based coolants.
Fisherbrand™ Hema Stain Platen Tubing
Highly resistant to oil, gasoline, kerosene
Fisherbrand™ Clear PVC Tubing Available on GSA/VA Contract for Federal Government customers only.
General-purpose polyvinyl chloride tubing for rinse and drain tubes
Cole Parmer C-Flex Tubing
Excellent biocompatibility
Fisherbrand™ Vinyl Tubing and Connector Kits
Resist fruit acids, lye, alcohol, greases, and oils
Kimble™ Kontes™ PTFE Tubing Available on GSA/VA Contract for Federal Government customers only.
Chemically inert; suitable for low pressure applications
Fisherbrand™ Butyl Rubber Tubing
For use with air and carbon dioxide
  spinner